1 - 30 of 30 products displayed
  • GRID2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536405
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, delta-2, GluD2, GluR delta-2 subunit, GRID2, GLURD2, GluR delta2, GluR delta2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536405 309.00
Check your price
  • CD82 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519807
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:80), Western Blot (1 µg/ml)
Alternate names:
Tetraspanin-27; Suppressor of tumorigenicity 6 protein; Metastasis suppressor Kangai-1; Inducible membrane protein R2; IA4; C33 antigen; Metastasis suppressor Kangai-1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519807 349.00
Check your price
  • NCDN Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544254
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Norbin; M-Sema F-associating protein of 75 kDa; Neurite outgrowth-related protein from the rat brain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544254 309.00
Check your price
  • Phospho-MUNC18 (Ser313) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538296
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein unc-18 homolog A; Protein unc-18 homolog 1; rbSec1; p67; N-Sec1; MUNC18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538296 309.00
Check your price
  • MUNC18 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1742
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Protein unc-18 homolog A; Protein unc-18 homolog 1; rbSec1; p67; N-Sec1; MUNC18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1742 375.00
Check your price
  • MUNC18 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518212
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-10ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Protein unc-18 homolog A; Protein unc-18 homolog 1; p67; N-Sec1; MUNC18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518212 349.00
Check your price
  • Phospho-MUNC18 (Ser241) Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA535398
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Protein unc-18 homolog A; Protein unc-18 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535398 349.00
Check your price
  • Lactoferrin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA535355
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Talalactoferrin; Growth-inhibiting protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535355 349.00
Check your price
  • Collagen III Antibody, Biotin
  • Host Rabbit
  • Target Species Bovine, Human, Mammal
  • Conjugate Biotin
  • Catalog # PA128532
  • Size 50 µg
Applications
ELISA (1:10,000-1:50,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunoprecipitation (Assay dependent), Western Blot (1:5000-1:10,000)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128532 349.00
Check your price
  • CD82 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520356
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Tetraspanin-27; Suppressor of tumorigenicity 6 protein; Metastasis suppressor Kangai-1; Inducible membrane protein R2; IA4; C33 antigen; Metastasis suppressor Kangai-1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520356 349.00
Check your price
  • CD82 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513228
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Tetraspanin-27; Suppressor of tumorigenicity 6 protein; Metastasis suppressor Kangai-1; Inducible membrane protein R2; IA4; C33 antigen; Metastasis suppressor Kangai-1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513228 349.00
Check your price
  • CD82 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527233
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Tetraspanin-27; Suppressor of tumorigenicity 6 protein; Metastasis suppressor Kangai-1; Inducible membrane protein R2; IA4; C33 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527233 349.00
Check your price
  • Collagen III Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527828
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, alpha1 (III) collagen, collagen alpha-1(III) chain, collagen, fetal
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527828 349.00
Check your price
  • MUNC18 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530183
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Protein unc-18 homolog A; Protein unc-18 homolog 1; p67; N-Sec1; MUNC18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530183 349.00
Check your price
  • MUNC18 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530184
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Protein unc-18 homolog A; Protein unc-18 homolog 1; p67; N-Sec1; MUNC18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530184 349.00
Check your price
  • Phosphotyrosine Antibody, HRP
  • Host Rabbit
  • Target Species Chemical
  • Conjugate HRP
  • Catalog # 615820
  • Size 100 µg
Applications
ELISA (1:2000), Western Blot (1:1000-2000)
Alternate names:
Tyrosine, pTyr
Catalog # Size Price (AUD) Your Price (AUD) Qty
615820 303.00
Check your price
  • Carbonic Anhydrase IX Antibody (Monoclonal, GT12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516318
  • Size 100 µL
Applications
Flow Cytometry (1:100-1:1000 (1x10^6 cells)), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (2-10 µg/mg of lysate), Western Blot (1:100-1:1000)
Alternate names:
pMW1; Renal cell carcinoma-associated antigen G250; P54/58N; Membrane antigen MN; Carbonic anhydrase IX; Carbonate dehydratase IX
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516318 349.00
Check your price
  • KCNA3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112654
  • Size 100 µg
Applications
Western Blot (5-10 µg/ml)
Alternate names:
Voltage-gated potassium channel subunit Kv3; Voltage-gated potassium channel subunit Kv1.3; RGK5; RCK3; MK3; Voltage-gated K(+) channel HuKIII; HPCN3; HLK3; HGK5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112654 309.00
Check your price
  • TRIM33 Antibody (Monoclonal, 16G9)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1801
  • Size 100 µg
Applications
Immunocytochemistry (4 µg/ml), Immunofluorescence (4 ug/mL), Immunoprecipitation (Assay dependent), Western Blot (2 ug/mL)
Alternate names:
Tripartite motif-containing protein 33; Transcription intermediary factor 1-gamma; RET-fused gene 7 protein; Ectodermin homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1801 349.00
Check your price
  • GAPDH Loading Control Antibody, Biotin (Monoclonal, GA1R)
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Biotin
  • Catalog # MA515738BTIN
  • Size 50 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738BTIN 159.00
Check your price
  • GLT-1 Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA3040A
  • Size 100 µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
Solute carrier family 1 member 2; Sodium-dependent glutamate/aspartate transporter 2; GLT-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3040A 375.00
Check your price
  • GCAP2 Antibody (Monoclonal, A1)
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # MA1725
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1 ug/ml)
Alternate names:
Retinal guanylyl cyclase activator protein p24; Guanylate cyclase activator 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1725 275.00
Check your price
  • TRPM8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14282
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Transient receptor potential p8; Long transient receptor potential channel 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14282 309.00
Check your price
  • HA Tag Antibody, Biotin (Monoclonal, 2-2.2.14)
  • Host Mouse
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # 26183BTIN
  • Size 50 µL
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:1000)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
26183BTIN 159.00
Check your price
  • HA Tag Antibody (Monoclonal, 2-2.2.14)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 261831MG
  • Size 1 mg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (0.1 µg/ml)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
261831MG 928.00
Check your price
  • HA Tag Antibody
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1985
  • Size 100 µg
Applications
Western Blot (1:500-1:5000)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1985 249.00
Check your price
  • SLC9A8 Antibody (Monoclonal, 7A11)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA146357
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (2 µg/ml)
Alternate names:
Solute carrier family 9 member 8; Na(+)/H(+) exchanger 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146357 309.00
Check your price
  • HA Tag Antibody (Monoclonal, 2-2.2.14)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 26183
  • Size 100 µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (0.1 µg/ml)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
26183 159.00
Check your price
  • 6x-His Tag Antibody (Monoclonal, 4E3D10H2/E3)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1135
  • Size 100 ug
Applications
ELISA (25-500 ng/ml), Immunoprecipitation (2 ug), Western Blot (1:250-1:1000)
Alternate names:
HIS Tag, Histidine tag, Poly histidine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135 159.00
Check your price
  • beta Tubulin Loading Control Antibody, Biotin (Monoclonal, BT7R)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA516308BTIN
  • Size 50 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308BTIN 159.00
Check your price