1 - 30 of 30 products displayed
  • Maltose Binding Protein Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA3045
  • Size 50µl
Applications
Western Blot (1:1000-1:2000)
Alternate names:
MBP, Maltose Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3045 309.00
Check your price
  • RALBP1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA1035X
  • Size 20µL
Applications
Immunoprecipitation (2 µg), Western Blot (1:1000)
Alternate names:
Ral-interacting protein 1; Dinitrophenyl S-glutathione ATPase; 76 kDa Ral-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1035X 119.00
Check your price
  • 14-3-3 Pan Polyclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Rabbit
  • Target Species Bovine, C. elegans, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 510700
  • Size 200µg
Applications
ELISA (0.1-1.0 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Protein HS1; 14-3-3 protein tau; 14-3-3 protein T-cell
Catalog # Size Price (AUD) Your Price (AUD) Qty
510700 309.00
Check your price
  • TTK Monoclonal Antibody for Western Blot, IF, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 359100
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Phosphotyrosine picked threonine-protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
359100 303.00
Check your price
  • RPL17 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA524323
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
PD-1; 60S ribosomal protein L23
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524323 275.00
Check your price
  • RBP4 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524129
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Plasma retinol-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524129 275.00
Check your price
  • RBP4 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA524158
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.05-2 µg/ml)
Alternate names:
Plasma retinol-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524158 275.00
Check your price
  • RhoD Monoclonal Antibody for AGG
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17663
  • Size 100µg
Applications
Agglutination (Assay Dependent)
Alternate names:
Rho-related protein HP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17663 275.00
Check your price
  • M13 phage coat protein g8p Monoclonal Antibody for Western Blot, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Bacteria, Virus
  • Conjugate Unconjugated
  • Catalog # MA106604
  • Size 100µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1:25-1:200), Immunocytochemistry (1:25-1:200), Western Blot (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106604 375.00
Check your price
  • Bax Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA514000
  • Size 500µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:10-1:100)
Alternate names:
Bcl-2-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514000 399.00
Check your price
  • TRIM28 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 730029
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
Tripartite motif-containing protein 28; RING finger protein 96; Nuclear corepressor KAP-1; KRAB-interacting protein 1; KRAB-associated protein 1; E3 SUMO-protein ligase TRIM28
Catalog # Size Price (AUD) Your Price (AUD) Qty
730029 309.00
Check your price
  • AGR2/AGR3 Monoclonal Antibody for Western Blot, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110047
  • Size 100ug
Applications
Immunocytochemistry (1 ug/ml), Immunohistochemistry (Paraffin) (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Protein disulfide isomerase family A, member 18; Breast cancer membrane protein 11; Anterior gradient 3 homolog; Secreted cement gland protein XAG-2 homolog; HPC8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110047 309.00
Check your price
  • E6AP Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3843
  • Size 100µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:2000)
Alternate names:
Renal carcinoma antigen NY-REN-54; Oncogenic protein-associated protein E6-AP; Human papillomavirus E6-associated protein; E6AP ubiquitin-protein ligase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3843 349.00
Check your price
  • MTA1 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 390700
  • Size 1mL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MTA1, metastasis associated 1, metastasis associated gene 1, metastasis associated gene 1 protein, metastasis associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
390700 309.00
Check your price
  • Actin Monoclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Dog, Chicken, Cat, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA110046
  • Size 100ug
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1 ug/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Gamma-actin; Alpha-cardiac actin; Alpha-actin; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110046 249.00
Check your price
  • RALBP1 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA1035
  • Size 100µg
Applications
Immunoprecipitation (2 µg), Western Blot (1:1000)
Alternate names:
Ral-interacting protein 1; Dinitrophenyl S-glutathione ATPase; 76 kDa Ral-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1035 349.00
Check your price
  • SKP2 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 323300
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (5 ug), Western Blot (1-2 ug/ml)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45
Catalog # Size Price (AUD) Your Price (AUD) Qty
323300 304.00
Check your price
  • APPL1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA15890
  • Size 50µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Adapter protein containing PH domain, PTB domain and leucine zipper motif 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15890 550.00
Check your price
  • Fyn Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112607
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay Dependent)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112607 499.00
Check your price
  • VRK1 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1828
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (1:100-1:500), Western Blot (1:500-1:2000)
Alternate names:
Vaccinia-related kinase 1; Serine/threonine-protein kinase 51PK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1828 309.00
Check your price
  • PrP Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110152
  • Size 100ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
AltPrP; PrP33-35C; PrP27-30; ASCR
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110152 275.00
Check your price
  • Maltose Binding Protein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1989
  • Size 100µg
Applications
Western Blot (2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1989 159.00
Check your price
  • BRG1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110062
  • Size 100ug
Applications
Western Blot (10 ug/ml)
Alternate names:
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SNF2-beta; Protein brahma homolog 1; Protein BRG-1; Mitotic growth and transcription activator; BRG1-associated factor 190A; ATP-dependent helicase SMARCA4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110062 249.00
Check your price
  • BRG1 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 730011
  • Size 100µg
Applications
Immunofluorescence (1:250), Immunoprecipitation (2 ug), Western Blot (1:2000)
Alternate names:
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SNF2-beta; Protein brahma homolog 1; BRG1-associated factor 190A; ATP-dependent helicase SMARCA4; Protein BRG-1; Mitotic growth and transcription activator
Catalog # Size Price (AUD) Your Price (AUD) Qty
730011 309.00
Check your price
  • SMARCA2 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 730025
  • Size 100µg
Applications
Immunofluorescence (1:250), Immunoprecipitation (Assay Dependent), Western Blot (1:2000)
Alternate names:
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2; SNF2-alpha; Protein brahma homolog; BRG1-associated factor 190B; ATP-dependent helicase SMARCA2
Catalog # Size Price (AUD) Your Price (AUD) Qty
730025 309.00
Check your price
  • Caspase 8 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511558
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunoprecipitation (2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511558 425.00
Check your price
  • Human Serum Albumin Monoclonal Antibody, Biotin conjugate for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110051
  • Size 100ug
Applications
Western Blot (0.5µg/ml)
Alternate names:
GIG20, DKFZp779N1935, PRO0883, PRO0903, PRO1341, cell growth inhibiting protein 42, growth-inhibiting protein 20, serum albumin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110051 269.00
Check your price
  • NEDD8 Monoclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515028
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:300), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Ubiquitin-like protein Nedd8; Neural precursor cell expressed developmentally down-regulated protein 8; Neddylin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515028 275.00
Check your price
  • NEDD8 Polyclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Mouse, Non-human primate, Rat, Xenopus, Zebrafish, Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA517476
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Ubiquitin-like protein Nedd8; Neural precursor cell expressed developmentally down-regulated protein 8; Neddylin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517476 275.00
Check your price
  • PTEN Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 325800
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
325800 305.00
Check your price