1 - 2 of 2 products displayed
  • BAZ1B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA541631
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Williams-Beuren syndrome chromosomal region 9 protein homolog; Williams syndrome transcription factor homolog; Bromodomain adjacent to zinc finger domain protein 1B; hWALp2; Williams-Beuren syndrome chromosomal region 9 protein; Williams-Beuren syndrome chromosomal region 10 protein; Williams syndrome transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541631 309.00
Check your price
  • BAZ1B Monoclonal Antibody, Biotin conjugate for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 730068
  • Size 100 µg
Applications
Immunoprecipitation (2 ug)
Alternate names:
hWALp2; Williams-Beuren syndrome chromosomal region 9 protein; Williams-Beuren syndrome chromosomal region 10 protein; Williams syndrome transcription factor; Bromodomain adjacent to zinc finger domain protein 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
730068 309.00
Check your price