1 - 3 of 3 products displayed
  • FOXP4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530908
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
Fork head-related protein-like A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530908 349.00
Check your price
  • FOXP4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518287
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Fork head-related protein-like A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518287 349.00
Check your price
  • FOXP4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532415
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Fork head-related protein-like A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532415 349.00
Check your price