1 - 2 of 2 products displayed
  • ZNF44 Antibody (Monoclonal, 4E2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521976
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunohistochemistry (PFA fixed) (3 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Zinc finger protein KOX7; Zinc finger protein 58; Zinc finger protein 55; Gonadotropin-inducible ovary transcription repressor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521976 309.00
Check your price
  • ZNF44 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA544858
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Zinc finger protein KOX7; Zinc finger protein 58; Zinc finger protein 55; Gonadotropin-inducible ovary transcription repressor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544858 309.00
Check your price