Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 100 µL of distilled water.
Centrifuge to remove any insoluble material.
ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid sensing (proton gated) ion channel 3; acid sensing ion channel 3; acid-sensing (proton-gated) ion channel 3; Acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; ASIC3; hASIC3; hTNaC1; modulatory subunit of ASIC2a; Neuronal amiloride-sensitive cation channel 3; proton-gated cation channel subunit; Testis sodium channel 1
Gene Aliases: ACCN3; ASIC3; SLNAC1; TNAC1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support