Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD16 (clone J5511, isotype IgG1).
CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells. CD16 is a 50-65kD cell surface molecule which exists in two forms - a transmembranous form expressed by NK cells and some T cells and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD16-I; CD16-II; CD16/CD8; CD16a; CD16a antigen; CD16b; CD8; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8a; CD8b; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor (CD16a); Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor III-A; Fc gamma receptor IIIa; Fc gamma receptor IIIb; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD 16); Fc-gamma receptor IIIb (CD16); Fc-gamma RIII; Fc-gamma RIII-alpha; Fc-gamma RIII-beta; Fc-gamma RIIIa; fc-gamma RIIIb; FcgammaRIIIA; FcR-10; IgG Fc receptor III-1; IgG Fc receptor III-2; IgG Fc receptor III-A; immunoglobulin G Fc receptor III; Leu2 T-lymphocyte antigen; Low affinity immunoglobulin gamma Fc region receptor III-A; Low affinity immunoglobulin gamma Fc region receptor III-B; neutrophil-specific antigen NA; OKT8 T-cell antigen; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T-cell antigen Leu2; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-lymphocyte differentiation antigen T8/Leu-2; T8 T-cell antigen
Gene Aliases: CD16A; CD16B; CD8A; CD8B; CD8B1; FCG3; FCGR3; FCGR3A; FCGR3B; IGFR3; MAL
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support