Search Thermo Fisher Scientific
PA1-24899 detects Compound S (11 Deoxycortisol) from human samples.
PA1-24899 has been successfully used in radioimmune assay procedures.
The PA1-24899 immunogen is Compound S-3-(O)-carboxymethyl oxime-BSA.
For long term storage, keep at -70°C or below. Avoid freeze/thaw cycles.