Invitrogen
MA1-90934 detects Luciferase from Bacteria, Invertebrates and Firefly samples.
MA1-90934 has been successfully used in Western blot applications.
The MA1-90934 immunogen is: Luciferase protein from Photinus pyralis
Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.
Protein Aliases: Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase