Product References
A microengineered Brain-Chip to model neuroinflammation in humans.
iScience
Pediaditakis I,Kodella KR,Manatakis DV,Le CY,Barthakur S,Sorets A,Gravanis A,Ewart L,Rubin LL,Manolakos ES,Hinojosa CD,Karalis K
Published figure using CD11b monoclonal antibody (Product # MA1-80091) in Flow Cytometry
Fri Aug 19 00:00:00 EDT 2022
Sourcing of human peripheral blood-derived myeloid angiogenic cells under xeno-free conditions for the treatment of critical limb ischemia.
Stem cell research & therapy
Wong CWT,Sawhney A,Wu Y,Mak YW,Tian XY,Chan HF,Blocki A
Published figure using CD11b monoclonal antibody (Product # MA1-80091) in Immunohistochemistry
Sat Aug 13 00:00:00 EDT 2022
ADAM8 affects glioblastoma progression by regulating osteopontin-mediated angiogenesis.
Biological chemistry
Li Y,Guo S,Zhao K,Conrad C,Driescher C,Rothbart V,Schlomann U,Guerreiro H,Bopp MH,König A,Carl B,Pagenstecher A,Nimsky C,Bartsch JW
MA1-80091 was used in Immunocytochemistry to demonstrate that ADAM8 regulates OPN via JAK/STAT3 pathway in U87 cells and in primary macrophages.
Wed Jan 27 00:00:00 EST 2021
TGFβ1 and TGFβ2 proteins in corneas with and without stromal fibrosis: Delayed regeneration of apical epithelial growth factor barrier and the epithelial basement membrane in corneas with stromal fibrosis.
Experimental eye research
de Oliveira RC,Tye G,Sampaio LP,Shiju TM,DeDreu J,Menko AS,Santhiago MR,Wilson SE
MA1-80091 was used in Immunohistochemistry to investigate the expression and localization of transforming growth factor (TGF) β1 and TGFβ2 in rabbit corneas that healed with and without stromal fibrosis, and to further study defective perlecan incorporation in the epithelial basement membrane (EBM) in corneas with scarring fibrosis.
Fri Jan 01 00:00:00 EST 2021
Octyl Gallate Induces Pancreatic Ductal Adenocarcinoma Cell Apoptosis and Suppresses Endothelial-Mesenchymal Transition-Promoted M2-Macrophages, HSP90α Secretion, and Tumor Growth.
Cells
Chua KV,Fan CS,Chen CC,Chen LL,Hsieh SC,Huang TS
Published figure using CD11b monoclonal antibody (Product # MA1-80091) in Immunohistochemistry
Mon Dec 30 00:00:00 EST 2019
Epithelial basement membrane injury and regeneration modulates corneal fibrosis after pseudomonas corneal ulcers in rabbits.
Experimental eye research
Marino GK,Santhiago MR,Santhanam A,Lassance L,Thangavadivel S,Medeiros CS,Bose K,Tam KP,Wilson SE
Published figure using CD11b monoclonal antibody (Product # MA1-80091) in Immunofluorescence
Tue Aug 01 00:00:00 EDT 2017
Combined toll-like receptor 3/7/9 deficiency on host cells results in T-cell-dependent control of tumour growth.
Nature communications
Klein JC,Moses K,Zelinskyy G,Sody S,Buer J,Lang S,Helfrich I,Dittmer U,Kirschning CJ,Brandau S
MA180091 was used in immunohistochemistry - frozen section to find that TLR3, 7 and 9 deficiencies on host cells result in complete tumor regression and induction of anti-tumor immunity
Thu Mar 16 00:00:00 EDT 2017
Adipocyte progenitor cells initiate monocyte chemoattractant protein-1-mediated macrophage accumulation in visceral adipose tissue.
Molecular metabolism
Kaplan JL,Marshall MA,C McSkimming C,Harmon DB,Garmey JC,Oldham SN,Hallowell P,McNamara CA
MA1-80091 was used in flow cytometry to identify the source of obesity-induced MCP-1 and identify molecular regulators mediating MCP-1 production
Sun Nov 01 00:00:00 EDT 2015
In vivo transcriptional profiling of Yersinia pestis reveals a novel bacterial mediator of pulmonary inflammation.
mBio
Pechous RD,Broberg CA,Stasulli NM,Miller VL,Goldman WE
MA1-80091 was used in flow cytometry to characterize Yersinia pestis as a novel bacterial mediator of pulmonary inflammation due to in vivo transcriptional profiling
Tue Feb 17 00:00:00 EST 2015
Spatially distinct neutrophil responses within the inflammatory lesions of pneumonic plague.
mBio
Stasulli NM,Eichelberger KR,Price PA,Pechous RD,Montgomery SA,Parker JS,Goldman WE
MA1-80091 was used in flow cytometry to study inflammatory lesions of pneumonic plague and spatially distinct neutrophil responses
Tue Oct 13 00:00:00 EDT 2015
M27 expressed by cytomegalovirus counteracts effective type I interferon induction of myeloid cells but not of plasmacytoid dendritic cells.
Journal of virology
Döring M,Lessin I,Frenz T,Spanier J,Kessler A,Tegtmeyer P,Dağ F,Thiel N,Trilling M,Lienenklaus S,Weiss S,Scheu S,Messerle M,Cicin-Sain L,Hengel H,Kalinke U
MA1-80091 was used in flow cytometry to test if professional antigen-presenting cell subsets and MCMV-encoded evasins alter IFN-I responses
Mon Dec 01 00:00:00 EST 2014
Maternal immune stimulation during pregnancy shapes the immunological phenotype of offspring.
Brain, behavior, and immunity
Mandal M,Donnelly R,Elkabes S,Zhang P,Davini D,David BT,Ponzio NM
MA1-80091 was used in flow cytometry to use poly(I:C)-injected dams to study the effects of inflammation on fetal development.
Tue Oct 01 00:00:00 EDT 2013
Early host cell targets of Yersinia pestis during primary pneumonic plague.
PLoS pathogens
Pechous RD,Sivaraman V,Price PA,Stasulli NM,Goldman WE
MA1-80091 was used in flow cytometry to determine when in infection Yersinia pestis infects macrophages and neutrophils
Mon May 26 00:00:00 EDT 2014
The stimulated innate resistance event in Bordetella pertussis infection is dependent on reactive oxygen species production.
Infection and immunity
Zurita E,Moreno G,Errea A,Ormazabal M,Rumbo M,Hozbor D
MA1-80091 was used in flow cytometry to study the effect of lipopolysaccharide on StIR.
Mon Jul 01 00:00:00 EDT 2013
A2A adenosine receptor signaling in lymphocytes and the central nervous system regulates inflammation during experimental autoimmune encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
Mills JH,Kim DG,Krenz A,Chen JF,Bynoe MS
MA1-80091 was used in immunocytochemistry to study the importance of A2AAR signaling during experimental autoimmune encephalomyelitis.
Fri Jun 01 00:00:00 EDT 2012
CD40-activated B cells can efficiently prime antigen-specific naïve CD8+ T cells to generate effector but not memory T cells.
PloS one
Mathieu M,Cotta-Grand N,Daudelin JF,Boulet S,Lapointe R,Labrecque N
MA1-80091 was used in flow cytometry to test the ability of different types of antigen presenting cells to induce a CD8(+) T cell response.
Mon Jun 04 00:00:00 EDT 2012
NK cells are effectors for resolvin E1 in the timely resolution of allergic airway inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Haworth O,Cernadas M,Levy BD
MA1-80091 was used in flow cytometry to investigate how inflammation is resolved using a self-limited allergic airway inflammation model.
Wed Jun 01 00:00:00 EDT 2011
Outer membrane vesicles obtained from Bordetella pertussis Tohama expressing the lipid A deacylase PagL as a novel acellular vaccine candidate.
Vaccine
Asensio CJ,Gaillard ME,Moreno G,Bottero D,Zurita E,Rumbo M,van der Ley P,van der Ark A,Hozbor D
MA1-80091 was used in flow cytometry to create a safer and more effective pertussis acellular vaccine.
Fri Feb 11 00:00:00 EST 2011
Production of IFN-β during Listeria monocytogenes infection is restricted to monocyte/macrophage lineage.
PloS one
Solodova E,Jablonska J,Weiss S,Lienenklaus S
MA1-80091 was used in flow cytometry to determine which cells are responsible for IFN-beta expression after infection with Listeria monocytogenes
Mon Apr 11 00:00:00 EDT 2011
Conventional bone marrow-derived dendritic cells contribute to toll-like receptor-independent production of alpha/beta interferon in response to inactivated parapoxvirus ovis.
Journal of virology
Siegemund S,Hartl A,von Buttlar H,Dautel F,Raue R,Freudenberg MA,Fejer G,Büttner M,Köhler G,Kirschning CJ,Sparwasser T,Alber G
MA1-80091 was used in flow cytometry to study the effect of inactivated Parapoxvirus ovis on dendritic cells
Tue Sep 01 00:00:00 EDT 2009
A defect in Marco expression contributes to systemic lupus erythematosus development via failure to clear apoptotic cells.
Journal of immunology (Baltimore, Md. : 1950)
Rogers NJ,Lees MJ,Gabriel L,Maniati E,Rose SJ,Potter PK,Morley BJ
MA1-80091 was used in flow cytometry to describe a role for Marco in the development of disease in BXSB mice
Sun Feb 15 00:00:00 EST 2009
Chemokine receptor CXCR5 supports solitary intestinal lymphoid tissue formation, B cell homing, and induction of intestinal IgA responses.
Journal of immunology (Baltimore, Md. : 1950)
Velaga S,Herbrand H,Friedrichsen M,Jiong T,Dorsch M,Hoffmann MW,Förster R,Pabst O
MA1-80091 was used in flow cytometry to characterize and track solitary intestinal lymphoid tissues
Sun Mar 01 00:00:00 EST 2009
Myeloid cells contribute to tumor lymphangiogenesis.
PloS one
Zumsteg A,Baeriswyl V,Imaizumi N,Schwendener R,Rüegg C,Christofori G
MA1-80091 was used in flow cytometry to study the role of bone marrow-derived cells in lymphangiogenesis
Thu Sep 17 00:00:00 EDT 2009
Effective induction of anti-tumor immune responses with oligomannose-coated liposome targeting to intraperitoneal phagocytic cells.
Cancer letters
Ikehara Y,Shiuchi N,Kabata-Ikehara S,Nakanishi H,Yokoyama N,Takagi H,Nagata T,Koide Y,Kuzushima K,Takahashi T,Tsujimura K,Kojima N
MA1-80091 was used in flow cytometry to employ a intraperitoneal drug delivery system for systemic cancer immunotherapy employing ovalbumin as a model antigen
Mon Feb 18 00:00:00 EST 2008
A gene-dosage effect for interleukin-4 receptor alpha-chain expression has an impact on Th2-mediated allergic inflammation during bronchopulmonary mycosis.
The Journal of infectious diseases
Müller U,Stenzel W,Köhler G,Polte T,Blessing M,Mann A,Piehler D,Brombacher F,Alber G
MA1-80091 was used in flow cytometry to determine the role of IL-4Ralpha to pulmonary C. neoformans infection
Mon Dec 01 00:00:00 EST 2008
Endogenous IL-17 does not play a significant role in the development of experimental murine allergic conjunctivitis.
International archives of allergy and immunology
Fukushima A,Sumi T,Ishida W,Yamada J,Iwakura Y,Ueno H
MA1-80091 was used in immunohistochemistry - frozen section to assess the contribution of IL-17 to the development of murine experimental allergic conjunctivitis
Mon Nov 10 00:00:00 EST 2008
IL-13 induces disease-promoting type 2 cytokines, alternatively activated macrophages and allergic inflammation during pulmonary infection of mice with Cryptococcus neoformans.
Journal of immunology (Baltimore, Md. : 1950)
Müller U,Stenzel W,Köhler G,Werner C,Polte T,Hansen G,Schütze N,Straubinger RK,Blessing M,McKenzie AN,Brombacher F,Alber G
MA1-80091 was used in flow cytometry to determine the role of the Th2 cytokine IL-13 during pulmonary infection with C. neoformans
Mon Oct 15 00:00:00 EDT 2007
Selective labeling and isolation of functional classes of interstitial cells of Cajal of human and murine small intestine.
American journal of physiology. Cell physiology
Chen H,Redelman D,Ro S,Ward SM,Ordög T,Sanders KM
MA1-80091 was used in flow cytometry to discuss how to isolate and purify interstitial cells of Cajal subsets
Mon Jan 01 00:00:00 EST 2007
Differential gene expression in functional classes of interstitial cells of Cajal in murine small intestine.
Physiological genomics
Chen H,Ordög T,Chen J,Young DL,Bardsley MR,Redelman D,Ward SM,Sanders KM
MA1-80091 was used in flow cytometry to design selective techniques to isolate the two functional interstitial cells of Cajal classes from enzymatically dispersed intestinal muscles
Wed Nov 14 00:00:00 EST 2007
Postburn monocytes are the major producers of TNF-alpha in the heterogeneous splenic macrophage population.
Shock (Augusta, Ga.)
Noel G,Guo X,Wang Q,Schwemberger S,Byrum D,Ogle C
MA1-80091 was used in flow cytometry to assess the effects of scald injury on spleen myeloid cells
Thu Mar 01 00:00:00 EST 2007
Role of T cells in innate and adaptive immunity against murine Burkholderia pseudomallei infection.
The Journal of infectious diseases
Haque A,Easton A,Smith D,O'Garra A,Van Rooijen N,Lertmemongkolchai G,Titball RW,Bancroft GJ
MA1-80091 was used in flow cytometry to study the in vivo role of cytokine- and antigen-mediated T cell activation in the resistance to Burkholderia pseudomallei
Wed Feb 01 00:00:00 EST 2006
TLR4, TLR9 and MyD88 are not required for the positive selection of autoreactive B cells into the primary repertoire.
European journal of immunology
Silver K,Ferry H,Crockford T,Cornall RJ
MA1-80091 was used in flow cytometry to demonstrate that the effects of Toll-like receptors on autoimmunity occur in the established repertoire and not during B cell development
Thu Jun 01 00:00:00 EDT 2006
Bone marrow mesenchymal stem cells suppress lymphocyte proliferation in vitro but fail to prevent graft-versus-host disease in mice.
Journal of immunology (Baltimore, Md. : 1950)
Sudres M,Norol F,Trenado A,Grégoire S,Charlotte F,Levacher B,Lataillade JJ,Bourin P,Holy X,Vernant JP,Klatzmann D,Cohen JL
MA1-80091 was used in flow cytometry to test if mesenchymal stem cells can control graft-vs-host disease
Thu Jun 15 00:00:00 EDT 2006
Hydrolytic and nonenzymatic functions of acetylcholinesterase comodulate hemopoietic stress responses.
Journal of immunology (Baltimore, Md. : 1950)
Grisaru D,Pick M,Perry C,Sklan EH,Almog R,Goldberg I,Naparstek E,Lessing JB,Soreq H,Deutsch V
MA1-80091 was used in flow cytometry to demonstrate that AChE-R and ARP facilitate granulocytosis
Sun Jan 01 00:00:00 EST 2006
Lack of antigen-specific tissue remodeling in mice deficient in the macrophage galactose-type calcium-type lectin 1/CD301a.
Blood
Sato K,Imai Y,Higashi N,Kumamoto Y,Onami TM,Hedrick SM,Irimura T
MA1-80091 was used in flow cytometry to investigate the contribution of macrophage galactose-type C-type lectin1 to granulation tissue formation
Fri Jul 01 00:00:00 EDT 2005
Granulation tissue formation by nonspecific inflammatory agent occurs independently of macrophage galactose-type C-type lectin-1.
Clinical immunology (Orlando, Fla.)
Sato K,Komatsu N,Higashi N,Imai Y,Irimura T
MA1-80091 was used in immunohistochemistry - frozen section to determine the role of a macrophage galactose-type calcium-type lectin-1 in antigen-independent granulation tissue formation
Fri Apr 01 00:00:00 EST 2005
Tick saliva inhibits differentiation, maturation and function of murine bone-marrow-derived dendritic cells.
Immunology
Cavassani KA,Aliberti JC,Dias AR,Silva JS,Ferreira BR
MA1-80091 was used in flow cytometry to assess the effects of Rhipicephalus sanguineus tick saliva on dendritic cell differentiation and maturation
Tue Feb 01 00:00:00 EST 2005
Time-course expression of Toll-like receptors 2 and 4 in inflammatory bowel disease and homeostatic effect of VIP.
Journal of leukocyte biology
Gomariz RP,Arranz A,Abad C,Torroba M,Martinez C,Rosignoli F,Garcia-Gómez M,Leceta J,Juarranz Y
MA1-80091 was used in flow cytometry to study TLR2 and 4 expression in a trinitrobenzene sulfonic acid-induced colitis model of Crohn's disease
Mon Aug 01 00:00:00 EDT 2005
Redistributions of macrophages expressing the macrophage galactose-type C-type lectin (MGL) during antigen-induced chronic granulation tissue formation.
International immunology
Sato K,Imai Y,Higashi N,Kumamoto Y,Mukaida N,Irimura T
MA1-80091 was used in flow cytometry to examine the roles of macrophage galactose-type C-type lectin 1 and 2 expressing cells during inflammation
Sun May 01 00:00:00 EDT 2005
Redistributions of macrophages expressing the macrophage galactose-type C-type lectin (MGL) during antigen-induced chronic granulation tissue formation.
International immunology
Sato K,Imai Y,Higashi N,Kumamoto Y,Mukaida N,Irimura T
MA1-80091 was used in flow cytometry to examine the roles of macrophage galactose-type C-type lectin 1 and 2 expressing cells during inflammation
Sun May 01 00:00:00 EDT 2005
Toll-like receptor 3 mediates West Nile virus entry into the brain causing lethal encephalitis.
Nature medicine
Wang T,Town T,Alexopoulou L,Anderson JF,Fikrig E,Flavell RA
MA1-80091 was used in immunohistochemistry to determine the role of Toll-like receptor 3 during West Nile virus infection
Wed Dec 01 00:00:00 EST 2004
In vivo self-renewing divisions of haematopoietic stem cells are increased in the absence of the early G1-phase inhibitor, p18INK4C.
Nature cell biology
Yuan Y,Shen H,Franklin DS,Scadden DT,Cheng T
MA1-80091 was used in flow cytometry to demonstrate that deletion p18(INK4C) results in improved long-term engraftment
Sat May 01 00:00:00 EDT 2004
Modulation of interferon-induced genes by lipoxin analogue in anti-glomerular basement membrane nephritis.
Journal of the American Society of Nephrology : JASN
Ohse T,Ota T,Kieran N,Godson C,Yamada K,Tanaka T,Fujita T,Nangaku M
MA1-80091 was used in flow cytometry to assess the effect of aspirin-triggered 15-epi-lipoxin A4 in experimental anti-glomerular basement membrane antibody nephritis
Thu Apr 01 00:00:00 EST 2004
Control of Leishmania major in the absence of Tyk2 kinase.
European journal of immunology
Schleicher U,Mattner J,Blos M,Schindler H,Röllinghoff M,Karaghiosoff M,Müller M,Werner-Felmayer G,Bogdan C
MA1-80091 was used in flow cytometry to test if Tyk2 kinase is required for protection against Leishmania major.
Sun Feb 01 00:00:00 EST 2004
Macrophages and stromal cells phagocytose apoptotic bone marrow-derived B lineage cells.
Journal of immunology (Baltimore, Md. : 1950)
Dogusan Z,Montecino-Rodriguez E,Dorshkind K
MA1-80091 was used in flow cytometry to show that macrophages efficiently phagocytose apoptotic murine B lineage cells.
Thu Apr 15 00:00:00 EDT 2004
Reduced apoptosis and ameliorated listeriosis in TRAIL-null mice.
Journal of immunology (Baltimore, Md. : 1950)
Zheng SJ,Jiang J,Shen H,Chen YH
MA1-80091 was used in flow cytometry to identify a role for TRAIL in apoptosis during listeriosis
Mon Nov 01 00:00:00 EST 2004
Protection against progressive leishmaniasis by IFN-beta.
Journal of immunology (Baltimore, Md. : 1950)
Mattner J,Wandersee-Steinhäuser A,Pahl A,Röllinghoff M,Majeau GR,Hochman PS,Bogdan C
MA1-80091 was used in flow cytometry to determine the contribution of IFN during infection with Leishmania major.
Tue Jun 15 00:00:00 EDT 2004
Purification of interstitial cells of Cajal by fluorescence-activated cell sorting.
American journal of physiology. Cell physiology
Ordög T,Redelman D,Miller LJ,Horváth VJ,Zhong Q,Almeida-Porada G,Zanjani ED,Horowitz B,Sanders KM
MA1-80091 was used in flow cytometry to describe a protocol for large-scale purification of interstitial cells of Cajal.
Sun Feb 01 00:00:00 EST 2004
Quantitative analysis by flow cytometry of interstitial cells of Cajal, pacemakers, and mediators of neurotransmission in the gastrointestinal tract.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Ordög T,Redelman D,Horváth VJ,Miller LJ,Horowitz B,Sanders KM
MA1-80091 was used in flow cytometry to assess the use of flow cytometry for the enumeration of interstitial cells of Cajal
Wed Dec 01 00:00:00 EST 2004
Detection of antigen-specific T cells in experimental immune-mediated blepharoconjunctivitis in DO11.10 T cell receptor transgenic mice.
Microbiology and immunology
Ozaki A,Ishida W,Fukata K,Fukushima A,Ueno H
MA1-80091 was used in immunohistochemistry to develop a model of experimental immune-mediated blepharoconjunctivitis to study the trafficking of antigen-specific T cells
Fri Apr 16 00:00:00 EDT 2004
Organ-specific and stage-dependent control of Leishmania major infection by inducible nitric oxide synthase and phagocyte NADPH oxidase.
European journal of immunology
Blos M,Schleicher U,Soares Rocha FJ,Meissner U,Röllinghoff M,Bogdan C
MA1-80091 was used in flow cytometry to report that iNOS and phox are both required for the control of L. major in different tissues and at different stages.
Thu May 01 00:00:00 EDT 2003
Granulocyte-dendritic cell unbalance in the non-obese diabetic mice.
Cellular immunology
Morin J,Chimènes A,Boitard C,Berthier R,Boudaly S
MA1-80091 was used in flow cytometry to investigate DCs in non-obese diabetic mice.
Thu May 01 00:00:00 EDT 2003
Butyric acid derivative induces allospecific T cell anergy and prevents graft-versus-host disease.
Immunopharmacology and immunotoxicology
Gilbert KM,Boger S,Fifer EK
MA1-80091 was used in flow cytometry to test if n-butyrate 2-(4-morpholinyl) ethyl butyrate hydrochloride inactivates the alloantigen-specific T cells that mediate graft-versus-host-disease
Sat Feb 01 00:00:00 EST 2003
TNF regulates chemokine induction essential for cell recruitment, granuloma formation, and clearance of mycobacterial infection.
Journal of immunology (Baltimore, Md. : 1950)
Roach DR,Bean AG,Demangel C,France MP,Briscoe H,Britton WJ
MA1-80091 was used in flow cytometry to assess the role of TNF in the formation and maintenance of granulomas.
Wed May 01 00:00:00 EDT 2002
Inflammatory responses to amyloidosis in a transgenic mouse model of Alzheimer's disease.
The American journal of pathology
Matsuoka Y,Picciano M,Malester B,LaFrancois J,Zehr C,Daeschner JM,Olschowka JA,Fonseca MI,O'Banion MK,Tenner AJ,Lemere CA,Duff K
MA1-80091 was used in immunohistochemistry (frozen) to explore how Abeta deposition is associated with immune responses.
Sun Apr 01 00:00:00 EST 2001
Tumor-infiltrating macrophages induce apoptosis in activated CD8(+) T cells by a mechanism requiring cell contact and mediated by both the cell-associated form of TNF and nitric oxide.
Journal of immunology (Baltimore, Md. : 1950)
Saio M,Radoja S,Marino M,Frey AB
MA1-80091 was used in flow cytometry to examine the ability of different tumor cells to induce apoptosis of activated CD8(+) T cells in vitro.
Thu Nov 15 00:00:00 EST 2001
Induction of germinal centers by MMTV encoded superantigen on B cells.
Developmental immunology
Simmons WJ,Simms M,Chiarle R,Mackay F,Tsiagbe VK,Browning J,Inghirami G,Thorbecke GJ
MA1-80091 was used in flow cytometry to test if direct stimulation of Mtv7+ DBA/2 B cells by vSAg-responsive T cells results in germinal centers.
Fri Aug 09 00:00:00 EDT 2002
Reduced oncogenicity of p190 Bcr/Abl F-actin-binding domain mutants.
Blood
Heisterkamp N,Voncken JW,Senadheera D,Gonzalez-Gomez I,Reichert A,Haataja L,Reinikainen A,Pattengale PK,Groffen J
MA1-80091 was used in flow cytometry to examine the contribution of the C-terminal Abl actin-binding domain within Bcr/Abl p190 in the development of leukemia/lymphoma in vivo.
Fri Sep 15 00:00:00 EDT 2000
Antigen-specific T helper cell function: differential cytokine expression in primary and memory responses.
The Journal of experimental medicine
Panus JF,McHeyzer-Williams LJ,McHeyzer-Williams MG
MA1-80091 was used in flow cytometry to investigate the functional potential and specialized functions of T cells.
Mon Nov 06 00:00:00 EST 2000
Evolution of antigen-specific T cell receptors in vivo: preimmune and antigen-driven selection of preferred complementarity-determining region 3 (CDR3) motifs.
The Journal of experimental medicine
McHeyzer-Williams LJ,Panus JF,Mikszta JA,McHeyzer-Williams MG
MA1-80091 was used in flow cytometry to investigate antigen-driven selection of helper T cells in normal animals.
Mon Jun 07 00:00:00 EDT 1999
Developmental dissociation of thymic dendritic cell and thymocyte lineages revealed in growth factor receptor mutant mice.
Proceedings of the National Academy of Sciences of the United States of America
Rodewald HR,Brocker T,Haller C
MA1-80091 was used in flow cytometry to test if thymocytes and thymic dendritic cell develop from a common intrathymic progenitor.
Tue Dec 21 00:00:00 EST 1999
Depletion of CD8+ cells abolishes memory in acquired immunity against Chlamydia pneumoniae in BALB/c mice.
Immunology
Penttilä JM,Anttila M,Varkila K,Puolakkainen M,Sarvas M,Mäkelä PH,Rautonen N
MA1-80091 was used in flow cytometry to determine the importance of T cells in Chlamydia pneumoniae infection.
Thu Jul 01 00:00:00 EDT 1999
Comparison of erythropoietic response to androgen in young and old senescence accelerated mice.
Mechanisms of ageing and development
Saitoh T,Morimoto K,Kumagai T,Tsuboi I,Aikawa S,Horie T
MA1-80091 was used in flow cytometry to test if the functional capacity of hemopoietic progenitor cells and the micro-environment of aged and young mice are identical.
Mon Aug 30 00:00:00 EDT 1999
Disruption of CD154:CD40 blocks generation of allograft immunity without affecting APC activation.
Journal of immunology (Baltimore, Md. : 1950)
Shepherd DM,Kerkvliet NI
MA1-80091 was used in flow cytometry to assess the contribution of the CD154:CD40 pathway using a murine model of allograft rejection.
Wed Sep 01 00:00:00 EDT 1999
Antigen-driven selection of TCR In vivo: related TCR alpha-chains pair with diverse TCR beta-chains.
Journal of immunology (Baltimore, Md. : 1950)
Mikszta JA,McHeyzer-Williams LJ,McHeyzer-Williams MG
MA1-80091 was used in flow cytometry to examine the dynamics of interclonal T cell competition during the selection process in vivo.
Wed Dec 01 00:00:00 EST 1999
Abnormal myelocytic cell development in interleukin-2 (IL-2)-deficient mice: evidence for the involvement of IL-2 in myelopoiesis.
Blood
Reya T,Contractor NV,Couzens MS,Wasik MA,Emerson SG,Carding SR
MA1-80091 was used in flow cytometry to characterize IL-2 deficient mice.
Wed Apr 15 00:00:00 EDT 1998
Lymphoid hyperplasia, autoimmunity, and compromised intestinal intraepithelial lymphocyte development in colitis-free gnotobiotic IL-2-deficient mice.
Journal of immunology (Baltimore, Md. : 1950)
Contractor NV,Bassiri H,Reya T,Park AY,Baumgart DC,Wasik MA,Emerson SG,Carding SR
MA1-80091 was used in flow cytometry to determine if the microbiota contributes to autoimmunity in IL-2 deficient mice.
Thu Jan 01 00:00:00 EST 1998
Targets of p56(lck) activity in immature thymoblasts: stimulation of the Ras/Raf/MAPK pathway.
International immunology
Lin K,Abraham KM
MA1-80091 was used in flow cytometry to demonstrate that the Ras/Raf/MAPK pathway is constitutively activated in lck-transformed immature thymoblasts.
Sat Feb 01 00:00:00 EST 1997
Growth hormone stimulates the proliferation of activated mouse T lymphocytes.
Endocrinology
Postel-Vinay MC,de Mello Coelho V,Gagnerault MC,Dardenne M
MA1-80091 was used in flow cytometry to report that bGH stimulates the proliferation of activated murine T cells.
Thu May 01 00:00:00 EDT 1997
Isolation and functional properties of murine hematopoietic stem cells that are replicating in vivo.
The Journal of experimental medicine
Goodell MA,Brose K,Paradis G,Conner AS,Mulligan RC
MA1-80091 was used in immunoprecipitation to use Hoechst 33342 to characterize hematopoietic stem cells.
Mon Apr 01 00:00:00 EST 1996
BCR/ABL P210 and P190 cause distinct leukemia in transgenic mice.
Blood
Voncken JW,Kaartinen V,Pattengale PK,Germeraad WT,Groffen J,Heisterkamp N
MA1-80091 was used in flow cytometry to characterize BCR/ABL P210 expressing transgenic mice.
Fri Dec 15 00:00:00 EST 1995
Reactive oxygen intermediates in autoimmune islet cell destruction of the NOD mouse induced by peritoneal exudate cells (rich in macrophages) but not T cells.
Diabetologia
Horio F,Fukuda M,Katoh H,Petruzzelli M,Yano N,Rittershaus C,Bonner-Weir S,Hattori M
MA1-80091 was used in flow cytometry to examine the contribution of oxygen free radicals to the development of insulitis in NOD mice.
Sat Jan 01 00:00:00 EST 1994
Expression of prolactin receptors in murine lymphoid cells in normal and autoimmune situations.
Journal of immunology (Baltimore, Md. : 1950)
Gagnerault MC,Touraine P,Savino W,Kelly PA,Dardenne M
MA1-80091 was used in flow cytometry to investigate the expression of prolactin receptor in lymphocyte subsets
Tue Jun 15 00:00:00 EDT 1993
Retrovirus-induced immunodeficiency in mice exacerbates gastrointestinal candidiasis.
Infection and immunity
Cole GT,Saha K,Seshan KR,Lynn KT,Franco M,Wong PK
MA1-80091 was used in immunohistochemistry (paraffin) to develop a murine model of invasive gastrointestinal candidiasis that is associated with an AIDS-related murine immunodeficiency syndrome.
Thu Oct 01 00:00:00 EDT 1992
Enhanced resistance against Listeria monocytogenes achieved by pretreatment with granulocyte colony-stimulating factor.
Infection and immunity
Shinomiya N,Tsuru S,Katsura Y,Kayashima S,Nomoto K
MA1-80091 was used in flow cytometry to study the roles of G-CSF and polymorphonuclear leukocytes in Listeria monocytogenes-infected mice.
Sun Dec 01 00:00:00 EST 1991
Mac-1: a macrophage differentiation antigen identified by monoclonal antibody.
European journal of immunology
Springer T,Galfré G,Secher DS,Milstein C
MA1-80091 was used in flow cytometry and immunoprecipitation to discuss a macrophage differentiation antigen identified by monoclonal antibody
Sun Apr 01 00:00:00 EST 1979
Monoclonal xenogeneic antibodies to murine cell surface antigens: identification of novel leukocyte differentiation antigens.
European journal of immunology
Springer T,Galfrè G,Secher DS,Milstein C
MA1-80091 was used to characterize monoclonal antibodies that recognize mouse cell surface antigens
Tue Aug 01 00:00:00 EDT 1978