Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LCA5L gene ontology annotations related to this gene include intraciliary transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lca5l; Leber congenital amaurosis 5-like; Leber congenital amaurosis 5-like protein; Lebercilin-like protein
Gene Aliases: C21orf13; LCA5L
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support