Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA5-28145 targets LRP11 in IHC (P) and WB applications and shows reactivity with Human samples.
The PA5-28145 immunogen is recombinant fragment corresponding to a region within amino acids 263 and 500 of Human LRP11.
LRP11 is a protein coding gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: low density lipoprotein receptor-related protein 11; Low-density lipoprotein receptor-related protein 11; LRP-11
Gene Aliases: LRP11
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support