Product References
An induced pluripotent stem cell line (TRNDi001-D) from a Niemann-Pick disease type C1 (NPC1) patient carrying a homozygous p. I1061T (c. 3182T>C) mutation in the NPC1 gene.
Stem cell research
Li R,Pradhan M,Xu M,Roeder A,Beers J,Zou J,Liu C,Porter FD,Zheng W
Published figure using OCT4 monoclonal antibody (Product # A13998) in Immunohistochemistry
Wed Apr 01 00:00:00 EDT 2020
Generation of an induced pluripotent stem cell line (TRNDi004-I) from a Niemann-Pick disease type B patient carrying a heterozygous mutation of p.L43_A44delLA in the SMPD1 gene.
Stem cell research
Baskfield A,Li R,Beers J,Zou J,Liu C,Zheng W
Published figure using OCT4 monoclonal antibody (Product # A13998) in Immunocytochemistry
Wed May 01 00:00:00 EDT 2019
An induced pluripotent stem cell line (TRNDi006-A) from a MPS IIIB patient carrying homozygous mutation of p.Glu153Lys in the NAGLU gene.
Stem cell research
Huang W,Xu M,Li R,Baskfield A,Kouznetsova J,Beers J,Zou J,Liu C,Zheng W
Published figure using OCT4 monoclonal antibody (Product # A13998) in Immunocytochemistry
Wed May 01 00:00:00 EDT 2019
Generation of an induced pluripotent stem cell line (TRNDi008-A) from a Hunter syndrome patient carrying a hemizygous 208insC mutation in the IDS gene.
Stem cell research
Hong J,Xu M,Li R,Cheng YS,Kouznetsova J,Beers J,Liu C,Zou J,Zheng W
Published figure using OCT4 monoclonal antibody (Product # A13998) in Immunocytochemistry
Wed May 01 00:00:00 EDT 2019
β-Hydroxybutyrate Prevents Vascular Senescence through hnRNP A1-Mediated Upregulation of Oct4.
Molecular cell
Han YM,Bedarida T,Ding Y,Somba BK,Lu Q,Wang Q,Song P,Zou MH
Published figure using OCT4 monoclonal antibody (Product # A13998) in Immunofluorescence
Thu Sep 20 00:00:00 EDT 2018
β-Hydroxybutyrate Prevents Vascular Senescence through hnRNP A1-Mediated Upregulation of Oct4.
Molecular cell
Han YM,Bedarida T,Ding Y,Somba BK,Lu Q,Wang Q,Song P,Zou MH
Published figure using OCT4 monoclonal antibody (Product # A13998) in Immunofluorescence
Thu Sep 20 00:00:00 EDT 2018
Induced Pluripotent Stem Cells for Disease Modeling and Evaluation of Therapeutics for Niemann-Pick Disease Type A.
Stem cells translational medicine
Long Y,Xu M,Li R,Dai S,Beers J,Chen G,Soheilian F,Baxa U,Wang M,Marugan JJ,Muro S,Li Z,Brady R,Zheng W
A13998 was used in immunocytochemistry to generate a model for Niemann-Pick disease type A and screen for compounds that inhibit reduce lysosomal lipid accumulation
Thu Dec 01 00:00:00 EST 2016
Morphokinetics of cloned mouse embryos treated with epigenetic drugs and blastocyst prediction.
Reproduction (Cambridge, England)
Mallol A,Piqué L,Santaló J,Ibáñez E
A13998 was used in immunocytochemistry to analyze cloned mouse embryos mophokinetics after treatment with epigenetic drugs and blastocyst prediction
Tue Mar 01 00:00:00 EST 2016
Development and Dynamic Regulation of Mitochondrial Network in Human Midbrain Dopaminergic Neurons Differentiated from iPSCs.
Stem cell reports
Fang D,Qing Y,Yan S,Chen D,Yan SS
Published figure using OCT4 monoclonal antibody (Product # A13998) in Western Blot
Tue Oct 11 00:00:00 EDT 2016
Mfn2 is Required for Mitochondrial Development and Synapse Formation in Human Induced Pluripotent Stem Cells/hiPSC Derived Cortical Neurons.
Scientific reports
Fang D,Yan S,Yu Q,Chen D,Yan SS
A13998 was used in immunocytochemistry and western blot study mitochondrial development, neurogenesis, and synapse formation in human induced pluripotent stem cell-derived cortical neurons
Thu Aug 18 00:00:00 EDT 2016
Endosialin‑expressing bone sarcoma stem‑like cells are highly tumor‑initiating and invasive.
Molecular medicine reports
Sun DX,Liao GJ,Liu KG,Jian H
A13998 was used in immunocytochemistry and western blot to assess endosialin-expressing bone sarcoma stem-like cells and their invasive and highly-tumor initiating properties
Thu Oct 01 00:00:00 EDT 2015
Gene expression profiling for human iPS-derived motor neurons from sporadic ALS patients reveals a strong association between mitochondrial functions and neurodegeneration.
Frontiers in cellular neuroscience
Alves CJ,Dariolli R,Jorge FM,Monteiro MR,Maximino JR,Martins RS,Strauss BE,Krieger JE,Callegaro D,Chadi G
A13998 was used in immunocytochemistry to research human iPS-derived motor neurons from sporadic ALS patients for gene expression and a strong association between neurodegeneration and mitochondrial functions
Mon Aug 24 00:00:00 EDT 2015
Cellular characterization of human pluripotent stem cells.
Methods in molecular biology (Clifton, N.J.)
Quintanilla RH
Thu Sep 19 00:00:00 EDT 2013