Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with hamster, mouse and rat based on sequence homology.
PGAP2 is involved in the lipid remodeling steps of GPI-anchor maturation. Required for stable expression of GPI-anchored proteins at the cell surface.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell wall biogenesis 43 N-terminal homolog; FGF receptor activating protein 1; FGF receptor-activating protein 1; mental retardation, non-syndromic, autosomal recessive, 21; Post-GPI attachment to proteins factor 2
Gene Aliases: FRAG1; PGAP2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support