Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control for Western blot is 293 cells.
Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumor-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brca 2; brca-2; BRCA1/BRCA2-containing complex, subunit 2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; Breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; FANCB; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; mutant BRCA2; RP11-298P3.4
Gene Aliases: BRCA2; BRCC2; BROVCA2; FACD; FAD; FAD1; FANCD; FANCD1; GLM3; PNCA2; RAB163; XRCC11
UniProt ID: (Human) P51587, (Mouse) P97929
Entrez Gene ID: (Human) 675, (Mouse) 12190, (Rat) 360254
Molecular Function: damaged DNA-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support