Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein transport protein Sec24C; SEC24 family member C; SEC24 family, member C; SEC24 related gene family, member C; SEC24-related protein C
Gene Aliases: KIAA0079; SEC24C
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support