Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-91686 detects TGF alpha from human, mouse, rat, canine and hamster samples. MA1-91686 does not cross react with human or mouse EGF.
MA1-91686 has been successfully used in immunocytochemistry, immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue) applications. MA1-91686 is not suitable for use in Western blot applications.
The MA1-91686 immunogen is: Recombinant full length protein (Human).
TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EGF-like TGF; ETGF; Protransforming growth factor alpha; TGF a; TGF type 1; TGF α; TGF-alpha; TGFA; TGFalpha; TGFα; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; waved 1
Gene Aliases: TGFA
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support