Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Reconstitute with 100 µL of distilled water.
Centrifuge to remove any insoluble material.
Putative pheromone receptor implicated in the regulation of social and reproductive behavior. Expressed in a subset of sensory neurons located in the apical layer of the vomeronasal organ. Mice lacking V1RB2 show a disrupted pattern of axonal projections to the accessory olfactory bulb. Mice lacking all but one V1RA and V1RB gene (12% of the V1R repertoire) show a lack of chemosensory response to a subset of known pheromonal ligands and changes in maternal aggression as well as male reproductive behavior.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: VN1B2; VN1B4; vomernasal 1 receptor Vmn1r44; vomeronasal 1 receptor Vmn1r49; vomeronasal 1 receptor, B4; vomeronasal 1, receptor B2; vomeronasal organ family 1, receptor 5; vomeronasal organ family 1, receptor B2; vomeronasal receptor 1 A10; vomeronasal receptor V1RB11; Vomeronasal receptor VRi2; Vomeronasal type-1 receptor 44; Vomeronasal type-1 receptor 49; Vomeronasal type-1 receptor A10; Vomeronasal type-1 receptor B11; Vomeronasal type-1 receptor B2; Vomeronasal type-1 receptor B4
Gene Aliases: V1ra10; V1rb11; V1rb2; V1rb4; Vmn1r44; Vmn1r49
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support