Product References
BCL2, BCL6, IGH, TP53, and MYC protein expression and gene rearrangements as prognostic markers in diffuse large B-cell lymphoma: a study of 44 Turkish patients.
Cancer genetics
Akay OM,Aras BD,Isiksoy S,Toprak C,Mutlu FS,Artan S,Oner U,Gulbas Z
MA5-12077 was used in immunohistochemistry to study the prognostic value of immunohistochemical and FISH analysis of BCL2, BCL6, IGH, TP53, and MYC in Turkish patients with diffuse large B-cell lymphoma
Sat Mar 01 00:00:00 EST 2014
MYC expression and distribution in normal mature lymphoid cells.
The Journal of pathology
Cattoretti G
MA5-12077 was used in western blot to study which normal mature lymphoid cells express myc
Fri Feb 01 00:00:00 EST 2013
The dichloromethane extract of the ethnomedicinal plant Neurolaena lobata inhibits NPM/ALK expression which is causal for anaplastic large cell lymphomagenesis.
International journal of oncology
Unger C,Popescu R,Giessrigl B,Laimer D,Heider S,Seelinger M,Diaz R,Wallnöfer B,Egger G,Hassler M,Knöfler M,Saleh L,Sahin E,Grusch M,Fritzer-Szekeres M,Dolznig H,Frisch R,Kenner L,Kopp B,Krupitza G
MA5-12077 was used in western blot to study the role of reduced NPM/ALK expression in the potential therapeutic mechanism of an ethnomedical plant extract against anaplastic large cell lymphoma
Tue Jan 01 00:00:00 EST 2013
EGFR and MYC gene copy number aberrations are more common in squamous cell carcinoma than keratoacanthoma: a FISH study.
Journal of cutaneous pathology
Jacobs MS,Persons DL,Fraga GR
MA5-12077 was used in immunohistochemistry to use FISH to study the higher frequency of EGFR and MYC gene amplification in squamous cell carcinoma than in keratoacanthoma
Wed May 01 00:00:00 EDT 2013
Nonconserved lysine residues attenuate the biological function of the low-risk human papillomavirus E7 protein.
Journal of virology
Genovese NJ,Broker TR,Chow LT
MA5-12077 was used in western blot to study the effect of non-conserved lysine residues on the function of human papillomavirus E7 protein
Wed Jun 01 00:00:00 EDT 2011
Expression of p53, Ki-67 and c-Myc proteins is predictive of the surgical molecular margin in colorectal carcinoma.
Pathology oncology research : POR
Li N,Wang J,Shen S,Bu X,Tian X,Huang P
MA5-12077 was used in immunohistochemistry to study the value of p53, Ki-67 and c-myc expression in determining the surgical margin in colorectal carcinoma resection
Thu Sep 01 00:00:00 EDT 2011
Delaying Gal4-driven gene expression in the zebrafish with morpholinos and Gal80.
PloS one
Faucherre A,López-Schier H
MA5-12077 was used in immunohistochemistry and western blot to study the effect of morpholinos and Gal80 on gene expression driven by Gal40 in zebrafish
Wed Jan 26 00:00:00 EST 2011
Pro- and anticarcinogenic mechanisms of piceatannol are activated dose dependently in MCF-7 breast cancer cells.
Carcinogenesis
Vo NT,Madlener S,Bago-Horvath Z,Herbacek I,Stark N,Gridling M,Probst P,Giessrigl B,Bauer S,Vonach C,Saiko P,Grusch M,Szekeres T,Fritzer-Szekeres M,Jäger W,Krupitza G,Soleiman A
MA5-12077 was used in western blot to investigate the mechanism for the biphasic effect of piceatannol on carcinogenesis
Wed Dec 01 00:00:00 EST 2010
Detection of protein-RNA complexes in Xenopus oocytes.
Methods (San Diego, Calif.)
Huber PW,Zhao WM
MA5-12077 was used in immunoprecipitation to develop a method for detecting protein-RNA complexes in Xenopus oocytes
Sat May 01 00:00:00 EDT 2010
Proteomic analysis reveals Hrs ubiquitin-interacting motif-mediated ubiquitin signaling in multiple cellular processes.
The FEBS journal
Pridgeon JW,Webber EA,Sha D,Li L,Chin LS
MA5-12077 was used in western blot to study the role of ubiquitin signaling in multiple cellular processes using a proteomic analysis
Thu Jan 01 00:00:00 EST 2009
MYC insertions in diffuse-type gastric adenocarcinoma.
Anticancer research
Calcagno DQ,Guimarães AC,Leal MF,Seabra AD,Khayat AS,Pontes TB,Assumpção PP,De Arruda Cardoso Smith M,Burbano RR
MA5-12077 was used in immunohistochemistry to evaluate the genomic alterations of MYC in diffuse-type gastric adenocarcinoma
Wed Jul 01 00:00:00 EDT 2009
Prognostic significance of oncogenic markers in ductal carcinoma in situ of the breast: a clinicopathologic study.
The breast journal
Altintas S,Lambein K,Huizing MT,Braems G,Asjoe FT,Hellemans H,Van Marck E,Weyler J,Praet M,Van den Broecke R,Vermorken JB,Tjalma WA
MA5-12077 was used in immunohistochemistry to evaluate oncogenic markers with prognostic significance in breast ductal carcinoma in situ
Wed Jul 08 00:00:00 EDT 2009
Mitochondrial p32 is a critical mediator of ARF-induced apoptosis.
Cancer cell
Itahana K,Zhang Y
MA5-12077 was used in western blot to examine the role of mitochondrial p32 in ARF-induced apoptosis
Sun Jun 01 00:00:00 EDT 2008
Molecular risk stratification of medulloblastoma patients based on immunohistochemical analysis of MYC, LDHB, and CCNB1 expression.
Clinical cancer research : an official journal of the American Association for Cancer Research
de Haas T,Hasselt N,Troost D,Caron H,Popovic M,Zadravec-Zaletel L,Grajkowska W,Perek M,Osterheld MC,Ellison D,Baas F,Versteeg R,Kool M
MA5-12077 was used in immunohistochemistry to develop a molecular risk stratification model for medulloblastoma patients
Tue Jul 01 00:00:00 EDT 2008
Downregulation of EGF receptor signaling in pancreatic islets causes diabetes due to impaired postnatal beta-cell growth.
Diabetes
Miettinen PJ,Ustinov J,Ormio P,Gao R,Palgi J,Hakonen E,Juntti-Berggren L,Berggren PO,Otonkoski T
MA5-12077 was used in immunohistochemistry and western blot to study the role of EGF receptor signaling in postnatal beta-cell growth in pancreatic islets
Fri Dec 01 00:00:00 EST 2006
Expression and prognostic implications of apoptosis-related proteins in locally unresectable non-small cell lung cancers.
Lung cancer (Amsterdam, Netherlands)
Fokkema E,Timens W,de Vries EG,de Jong S,Fidler V,Meijer C,Groen HJ
MA5-12077 was used in immunohistochemistry to study the prognostic value of apoptosis-related protein expression in locally unresectable non-small cell lung cancers
Mon May 01 00:00:00 EDT 2006
Defined genetic events associated with the spontaneous in vitro transformation of ElA/Ras-expressing human IMR90 fibroblasts.
Carcinogenesis
Mason DX,Keppler D,Zhang J,Jackson TJ,Seger YR,Matsui S,Abreo F,Cowell JK,Hannon GJ,Lowe SW,Lin AW
MA5-12077 was used in western blot to study the genetic events associated with spontaneous in vitro transformation of EIA/Ras-expressing human fibroblasts
Wed Feb 01 00:00:00 EST 2006
The shed ectodomain of type XIII collagen associates with the fibrillar fibronectin matrix and may interfere with its assembly in vitro.
The Biochemical journal
Väisänen MR,Väisänen T,Tu H,Pirilä P,Sormunen R,Pihlajaniemi T
MA5-12077 was used in western blot to study the interplay between the shed soluble ectodomain of type XIII collagen and the fibrillar fibronectin matrix
Sun Jan 01 00:00:00 EST 2006
Metastasis-associated protein 1 interacts with NRIF3, an estrogen-inducible nuclear receptor coregulator.
Molecular and cellular biology
Talukder AH,Gururaj A,Mishra SK,Vadlamudi RK,Kumar R
MA5-12077 was used in immunoprecipitation to investigate the interaction between MTA1 and NRIF3
Sun Aug 01 00:00:00 EDT 2004
Upstream stimulatory factor but not c-Myc enhances transcription of the human polymeric immunoglobulin receptor gene.
Molecular immunology
Bruno ME,West RB,Schneeman TA,Bresnick EH,Kaetzel CS
MA5-12077 was used in western blot to study an upstream regulatory factor that enhances expression of the human polymeric immunoglobulin receptor
Thu Jan 01 00:00:00 EST 2004
Transmembrane peptides as inhibitors of ErbB receptor signaling.
Molecular biology of the cell
Bennasroune A,Fickova M,Gardin A,Dirrig-Grosch S,Aunis D,Crémel G,Hubert P
MA5-12077 was used in western blot to investigate the interaction between transmembrane domain of receptor tyrosine kinases and ErbB receptor
Thu Jul 01 00:00:00 EDT 2004
Sue1p is required for degradation of labile forms of altered cytochromes C in yeast mitochondria.
The Journal of biological chemistry
Wei J,Sherman F
MA5-12077 was used in western blot to study the role of the nuclear encoded mitochondrial protein Sue1p in the degradation of altered holo-iso-1-cytochrome c isoforms in yeast mitochondria
Fri Jul 16 00:00:00 EDT 2004
Construction of tumor-specific cells expressing a membrane-anchored single-chain Fv of anti-ErbB-2 antibody.
Biochimica et biophysica acta
Suzuki M,Shinkai M,Honda H,Kamihira M,Iijima S,Kobayashi T
MA5-12077 was used in flow cytometry to develop tumor-specific cells expressing a membrane-anchored single-chain Fv of anti-ErbB-2 antibody for drug targeting applications
Fri Feb 16 00:00:00 EST 2001
Mutations in human ARF exon 2 disrupt its nucleolar localization and impair its ability to block nuclear export of MDM2 and p53.
Molecular cell
Zhang Y,Xiong Y
MA5-12077 was used in immunocytochemistry to study the effect of mutations in exon 2 of human ARF on its nucleolar localization and ability to regulate nuclear export of MDM2 and p53
Sat May 01 00:00:00 EDT 1999
ARF promotes MDM2 degradation and stabilizes p53: ARF-INK4a locus deletion impairs both the Rb and p53 tumor suppression pathways.
Cell
Zhang Y,Xiong Y,Yarbrough WG
MA5-12077 was used in immunocytochemistry to investigate the effect of ARF on p53 regulation and its mechanism
Fri Mar 20 00:00:00 EST 1998