Through high-affinity antibodies attached to the surface of magnetic Dynabeads™—known as immunomagnetic separation (IMS) technology—a valuable means to isolate specific biological entities from complex matrices is provided for your work!  Their use in sample preparation greatly improves the speed, sensitivity, and reproducibility of microbial analysis.  Dynabeads™ IMS food products have been designed for both manual and automated separation of the specific target organism.
View Dynabeads™ Products
Product Applications
Dynabeads™MAX E. coli O157 kit
For the selective capture and concentration of E. coli O157:H7 directly from pre-enrichment, water, or environmental samples. Sample preparation designed for PCR analysis and plating culture or ELISA–based assays. Can be automated using the BeadRetriever™ System or performed manually.
Dynabeads™anti-E. coli O157
For the selective capture and concentration of E. coli O157:H7 directly from pre-enrichment, water, or environmental samples. Sample preparation designed for plating culture or ELISA–based assays. Can be automated using the BeadRetriever™ System or performed manually.
Dynabeads™anti-Listeria
For the selective capture and concentration of Listeria directly from pre-enrichment samples.  Sample preparation designed for plating culture, ELISA, or PCR–based assays. Can be automated using the BeadRetriever™ System or performed manually.
Dynabeads™anti-Salmonella
For the selective capture and concentration of Salmonella directly from pre-enrichment samples. Sample preparation designed for plating culture, ELISA, or PCR–based assays. Can be automated using the BeadRetriever™ System or performed manually.
Dynabeads™EPEC/VTEC O145 For the rapid selective concentration of E. coli O145.
Dynabeads™EPEC/VTEC O111 For the rapid selective concentration of E. coli O111.
Dynabeads™EPEC/VTEC O103 For the rapid selective concentration of E. coli O103.
Dynabeads™EPEC/VTEC O26 For the rapid selective concentration of E. coli O26.
BeadRetriever™ Bench-top instrument to perform automated Immunomagnetic Separation (IMS). Pre-programmed to run auto-IMS with Dynabeads™ . Protocols provided for the capture of Cryptosporidium, E. coli O157, EPEC/VTEC, Giardia, Legionella, Listeria and Salmonella. Processes 15 1ml test samples in 20 minutes. After auto-IMS, processed samples can be used in any detection method.
BeadRetriever™ tubes & tips BeadRetriever accessories
BeadRetriever™ tube rack BeadRetriever accessories
CT-Supplement CT Supplement is added to standard Sorbitol MacConkey media (SMAC) to form CT-SMAC. CT-SMAC selects for E. coli O157 after IMS.