Featured articles      |     New product releases      |     Resources and guides      |     News and views
Fluorescent immunohistochemistry using fresh frozen tissue samples
Fluorescent Immunohistochemistry (IHC) is a simpler alternative to the conventional chromogenic IHC method, and has a few advantages: stained slides have a longer shelf life as the signal is more stable and also do not require complicated advanced equipment to visualize the result. The fluorescent IHC technique however has an edge over the traditional method of chromogenic IHC in the following ways: 
  • It involves fewer steps; for example the addition of substrate as in the case of chromogenic methods is omitted
  • Images with higher resolution can be obtained
  • Both high and low abundant proteins can be detected
  • More than one protein can be detected through multiplexing, permitting co-localization studies
  • Subcellular particulate data (visualizing DNA or chromosomes) can be obtained more easily
IHC can be performed using paraffin-embedded tissues or fresh frozen tissue samples. A fresh frozen tissue sample is preferred for fluorescent IHC as it is a better starting material from which to analyze post-translation modifications. The following are some advantages the fresh frozen tissue samples have over the paraffin-embedded tissue samples: 
  • The antigenicity of the protein is better preserved
  • Working with fresh tissues is non-toxic or non-carcinogenic since formalin is not used
  • Processing time involved is considerably reduced with steps like epitope retrieval being eliminated
Tips and tricks
  • Since OCT (optimal cutting temperature) compound has a tendency to autofluoresce, wash slides before fixing
  • While washing the tissue section slides, extreme care should be taken to avoid peeling off of the sections
  • To avoid the peeling-off of tissue sections from the slides, tissue can be sectioned on coated/charged slides
  • To avoid cross contamination of samples on the same slide, use a PAP pen to create a hydrophobic barrier
  • Since PAP pen marking also has a tendency to autofluoresce, clean slides before mounting without disturbing the stained tissue sections
Some examples of IHC detection by immunofluorescence using fresh frozen tissue samples are shown below:
Immunofluorescence analysis of sarcomeric alpha actin in mouse heart tissue
Immunofluorescence analysis of sarcomeric alpha actin in mouse heart tissue using alpha actinin 2 Antibody (7H1L69), ABfinity Rabbit Monoclonal Antibody detected using goat anti-rabbit IgG (H+L) Secondary Antibody, Alexa Fluor 488 conjugate.
Immunofluorescence analysis of sarcomeric alpha actin in mouse heart tissue
Immunofluorescence analysis of corin in mouse heart tissue using alpha corin antibody, ABfinity Rabbit Oligoclonal Antibody, detected with goat anti-rabbit IgG (H+L) Superclonal Secondary Antibody, Alexa Fluor 488 conjugate.
Immunofluorescence analysis of cardiac troponin T in mouse heart tissue
Immunofluorescence analysis of cardiac troponin T in mouse heart tissue using cardiac troponin T monoclonal antibody, detected with goat anti-mouse IgG (H+L) Superclonal Secondary Antibody, Alexa Fluor 488 conjugate.
Search for the antibodies you need at thermofisher.com/antibodies
Learn more about IHC and how to get great images at thermofisher.com/ihc5steps
IHC workflow for advanced verification of antibodies
From sample preparation through image acquisition, Thermo Fisher Scientific offers a broad portfolio of superior reagents and instruments to help with every step of IHC workflow.
Overview of the IHC workflow
Overview of the IHC workflow.
One of the critical reagents for a successful IHC experiment is the detection antibody. Thermo Fisher Scientific is striving to redefine the criteria of antibody performance by testing the specificity of antibodies in accordance with the newly adopted advanced verification testing methods for antibody validation
IHC analysis of p53 expression
Proteins can be expressed in some cell or tissue types but not in others. Antibody target verification can be determined by analyzing the relative expression of target proteins in different cell or tissue types by IHC. For example, the tumor suppressor p53 protein is known to be strongly expressed in the nuclei of tumor cells within ovarian cancer tissue compared to low expression seen in normal ovary tissue. Additionally, p53 expression is not detectable in other tissues, such as normal human liver. The immunohistochemistry (IHC) analysis of p53 expression in positive ovarian cancer and negative ovary and liver tissues shown in the figure demonstrates the specificity of the p53 antibody used for detection. As expected, p53 is highly expressed (stained brown) in the nuclei of ovarian cancer, but low in normal ovary tissue and not expressed in normal liver tissue.
Detection of relative p53 expression by IHC
Detection of relative p53 expression by IHC. The specificity of anti-p53 monoclonal antibody was demonstrated by detecting relative p53 expression in ovarian cancer (positive, left panel), normal ovary (low expression, middle panel) and normal human liver (negative, right panel) tissues. Tissues were deparaffinized with xylene, followed by rehydration in sequential washes of 100% ethanol, 95% ethanol, 80% ethanol, and water. To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0) and heated for 20 min in Lab Vision PT Module. Following antigen retrieval, tissues were blocked in a 10% goat serum in wash buffer solution for 30 minutes at room temperature and endogenous peroxidase activity quenched with Peroxidase Suppressor. Tissue was then probed with a p53 mouse monoclonal antibody at a concentration of 2.5µg/ml in 10% goat serum in wash buffer for 1 hour at room temperature in a humidified chamber. Tissues were washed extensively with PBST, and detection was performed using a SuperBoost goat anti-mouse Poly HRP secondary antibody reagent followed by chromogenic detection using DAB Quanto. Tissues were then counterstained with hematoxylin, mounted and imaged on EVOS FL Auto Imaging Station.
IF analysis of p53 expression
The same antibody verification and IHC workflow described above can be performed using immunofluorescence (IF) in place of the chromogenic detection. For fluorescence detection, the primary or secondary antibody is conjugated to a fluorophore that is detected by fluorescent microscope. Recently introduced, Alexa Fluor Plus secondary antibodies offer high signal-to-noise ratios and superior brightness. This figure depicts an example of IHC detection by IF using a p53 monoclonal primary antibody and an Alexa Fluor Plus 488 conjugated secondary antibody to visualize the relative expression of p53.
IHC detection of p53 across tissues by immunofluorescence
IHC detection of p53 across tissues by immunofluorescence. The specificity of anti-p53 monoclonal antibody was demonstrated by detecting relative p53 expression in ovarian cancer (positive, left panel), normal ovary (low expression, middle panel) and normal human liver (negative, right panel) tissues. Tissue sections were prepared, blocked and incubated with a p53 mouse monoclonal antibody at a concentration of 2.5µg/ml in 10% goat serum in wash buffer for 1 hour at room temperature in a humidified chamber. Tissues were washed extensively with PBST, and detection was performed using a goat anti-mouse IgG (H+L) Secondary Antibody, Alexa Fluor Plus 488 conjugate at a dilution of 1:100 for at least 30 minutes at room temperature in the dark (green). Nuclei (blue) were stained with Hoechst 33342. Images were taken on EVOS FL Auto Imaging Station.
Learn more about antibody verification
Sharper and brighter images
Get sharper and brighter images when using Invitrogen Alexa Fluor Plus secondary antibodies with new Invitrogen ProLong Glass Antifade Mountant. Alexa Fluor Plus secondary antibodies are highly cross-adsorbed and generate up to four times brighter signal than Invitrogen Alexa Fluor secondary antibodies when used in IF/ICC. ProLong Glass Antifade Mountant preserves the brighter signal from Alexa Fluor Plus secondary antibodies, minimizing signal loss due to photobleaching. When cells were exposed to constant illumination over 90 seconds, ProLong Glass reagent preserved over 90% of the signal from Alexa Fluor Plus secondary antibodies, illustrating its superior photobleaching protection. Meanwhile, ProLong Glass reagemt features low background in all fluorescence channels. Brighter signals combined with low background and better photobleaching protection yield higher signal-to-noise ratios, resulting in enhanced sensitivity required for detection of low-abundance targets.
Improved resolution with same-as-glass RI
ProLong Glass Antifade Mountant, which features improved refractive index of 1.52, increases axial resolution up to 75% and imageable focal depth by 3-4 times compared to mountants with refractive index of 1.47. ProLong Glass Antifade Mountant’s refractive index is same as slide glass, immersion oil and microscope objective, which minimizes optical refraction path and allows more light in microscope objective. Now, for the first time, researchers can take bright, sharp images of tissues and samples up to 150 µm thick, preserved with a hard-set mountant.
Alexa Fluor Plus secondary antibodies with Invitrogen ProLong Glass Antifade Mountant
Get the most from your samples and microscope by using Alexa Fluor Plus secondary antibodies with ProLong Glass Antifade Mountant. Alexa Fluor Plus secondary antibodies exhibit bright fluorophore technology, with the least background. ProLong Glass reagent preserves the bright signal and low background with antifade technology, and its near-perfect refractive index enhances focal depth and resolution of images. Bright and sharp images are possible when these two innovative technologies are used together.
Differentiated H9 NSC-derived neurons mounted with Prolong Glass antifade mountant, Alexa Fluor Plus secondary antibodies and imaged with EVOS FL Auto 2
Differentiated H9 NSC-derived neurons mounted with Prolong Glass Antifade Mountant, Alexa Fluor Plus secondary antibodies and imaged with EVOS FL Auto 2. H9 NSC-derived neurons after 3 weeks of differentiation in the Gibco B-27 Plus Neuronal Culture System, containing B-27 Plus Supplement and Neurobasal Plus Medium, and CultureOne Supplement. Cells were fixed and labeled with NucBlue Fixed Cell ReadyProbes Reagent, mouse anti-beta-3 Tubulin Antibody detected with Invitrogen Alexa Fluor Plus 488 Goat Anti-Mouse Secondary Antibody, ActinRed ReadyProbes Reagent, and rabbit anti-Synaptophysin Antibody (Cat. No. 18-0130) detected with Invitrogen Alexa Fluor Plus 647 Goat Anti-Rabbit Secondary Antibody (Cat. No. 32733). Cells were mounted in ProLong Glass Antifade Mounting Medium. Images acquired on EVOS FL Auto 2 using a 10x Olympus air objective.
Learn more
NEW PRODUCT RELEASES
Introducing the EVOS FL Auto 2 Imaging System
Invitrogen EVOS FL Auto 2 Imaging System
Bring high performance and fast automated imaging right to your lab bench with the new Invitrogen EVOS FL Auto 2 Imaging System. This system has been designed with advanced capabilities to simplify demanding cell-based imaging applications such as live-cell imaging, image tiling, and Z-stacking, so researchers can focus on their data rather than instrument operation.
Features
  • Speed—scan a 96-well plate in 3 fluorescent channels in less than 5 minutes
  • Flexibility—customize the system with more than 20 user-changeable LED light cubes, dual cameras (monochrome and color), a variety of objectives ranging from 1.25x to 100x, and multiple vessel holders
  • Time-lapse live-cell imaging—option for precise control of temperature, humidity, and gases for normoxic or hypoxic conditions allows a wide range of biological studies under physiological conditions with an onstage incubator
  • Automation—time-saving features such as autofocus, rapid stage movement, and automated routines help reduce time to complete experiments, allowing high throughput, high data quality, and improved experimental reproducibility
  • Data analysis—optional Celleste advanced software package for quantitative and statistical analysis
FFPE section of rat intestine tissue labeled with the Invitrogen Click-iT EdU Colorimetric IHC Detection Kit  combined with Movat’s pentachrome stain to show proliferating cells
FFPE section of rat intestine tissue labeled with the Invitrogen Click-iT EdU Colorimetric IHC Detection Kit combined with Movat’s pentachrome stain to show proliferating cells. Image was collected on the EVOS FL Auto 2 Imaging System using the color camera and 20x objective.
Learn more about our EVOS imaging solutions at thermofisher.com/microscopes
Exciting news for high-content analysis: Introducing the CellInsight CX7 LZR
CellInsight CX7 LZR
The new Thermo Scientific CellInsight CX7 LZR platform harnesses advanced laser-based illumination and enables customers to collect data faster and achieve superior imaging and analysis, especially with thicker samples. Designed to provide customers with the sensitivity and speed they need for emerging assays by allowing better penetration to sample, the CellInsight CX7 LZR accelerates confocal imaging by speeds up to 3x for broad assay development to enable faster sample to data times at a superior value.
At a glance
The CellInsight CX7 LZR instrument offers:
  • Faster scan times—take only seconds for confocal imaging of one picture
  • Superior image quality—enhanced light performance illuminates the field evenly enabling image-quality quantitation
  • Higher sensitivity at more wavelengths—expand your research with the capability to use a diverse array of dyes and multiplexing with no bleed through
Learn more about the CellInsight CX7 LZR
CellInsight CX7 LZR brochure
Download the CellInsight CX7 LZR brochure
Live-cell imaging for our high-content imagers
Invitrogen HCA Onstage Incubator
The all new Invitrogen HCA Onstage Incubator
Live-cell imaging enables researchers to observe biological processes as they happen. On-stage incubation creates an environment for longer-term imaging for quantitative analysis. This enables researchers to understand the context of cell biology over time for identification of minority populations and the ability to perform experimental manipulations on cell samples during observation. In addition, customers can access increased statistical power through our powerful HCS Studio Software. 
The On-Stage Incubator for the Thermo Scientific CellInsight platforms was designed for:
  • Stable and consistent live-cell imaging for at least 24 hours
  • Movement/motility studies in cells
  • Kinetics of cell-level measurements
  • Dual purpose screening and environmental control
  • Upgrades to current and future platforms
Features:
  • Enables short- and longer-term high-content experiments
  • On-stage incubator with precise control of temperature, humidity, and CO2
  • Includes all scheduling software and kinetic and motility measurements
  • The ability to generate multicolor movies
  • Robot-compatible for fixed-endpoint screens
Learn more about the Invitrogen HCA On-Stage Incubator
GFP compensation for flow cytometry
GFP BrightComp eBeads Compensation Beads were designed for setting flow cytometry compensation when using green fluorescent protein (GFP).
  • Consistent
  • Accurate
  • Simple-to-use reagent
With a near-identical spectral match to GFP at 3 levels of intensity, dispense the eBeads as a single drop for compensation made easy (see figures).
Multiple intensities of GFP BrightComp eBeads
Multiple intensities of GFP BrightComp eBeads. Fluorescent proteins can be expressed at varying levels, resulting in the detection of a range of fluorescent intensities. When setting compensation, selection of the bead peak with a higher intensity than the experiment sample is recommended. Data were acquired using a 488-nm laser and emission was collected using 525/50 nm bandpass filter for GFP.
GFP BrightComp eBeads are used to compensate GFP
GFP BrightComp eBeads are used to compensate GFP. (A–C) HeLa cells, a human cervical cancer cell line, were transduced with CellLight Histone 2B-GFP, BacMam 2.0, resulting in GFP expression. Cells were subsequently harvested and stained with rat anti-human/mouse CD44-PE antibody, then analyzed using a 488-nm laser. Samples compensated with GFP BrightComp eBeads (C) show the same degree of compensation as the samples compensated using the GFP-expressing HeLa cells (B).
Learn more
Ready-to-use flow cytometry reagents
Ready-to-use flow cytometry reagents
Invitrogen Ready Flow reagents are ready-to-use, room temperature- stable solutions designed to allow you to stain your cells for analysis by flow cytometry with:
  • No calculations
  • No dilutions
  • No pipetting
Reagents are available for most common needs—dead cell identification, cell cycle, and apoptosis.
View the selection guide at thermofisher.com/readyflow
eBioscience Super Bright 780 antibody conjugates for flow cytometry
Similar to Brilliant Violet 785 and Brilliant Violet 786, the new eBioscience Super Bright 780 dye expands your options for available violet-laser excitable antibody conjugates and joins the growing family of over 700 Super Bright antibody conjugates. Now available in five fluorophore formats (Super Bright 436, 600, 645, 702, and 780), view the Super Bright selection guide, see comparative data and select the Super Bright antibody conjugate for your flow cytometry panel.
Find out more at thermofisher.com/superbright
Fluorescence intensity and dot plot comparison of Super Bright 780 conjugates and Brilliant Violet 786 conjugates
Fluorescence intensity and dot plot comparison of Super Bright 780 conjugates and Brilliant Violet 786 conjugates. (A) Mouse splenocytes stained with anti-CD4 conjugated to Super Bright 780 (red) or Brilliant Violet 786 conjugate (gray), at the same concentration of antibody. (B) Human peripheral blood cells stained with anti-CD19 conjugated to (B) Brilliant Violet 786 or (C) Super Bright 780 at the same concentration of antibody. 
RESOURCES AND GUIDES
Tool box—Five steps for publication-quality fixed-cell images
fixed cell imaging brochure
You can access a toolbox of resources to help you obtain publication-quality fixed-cell images utilizing the proven five steps that were developed using the expertise of our R&D scientists. You can download an easy-to-follow guide, watch a webinar, and visit our web page to get the best practices and solutions for fixed cell imaging. 
Coating chamber slides with extracellular matrix (ECM) for finicky cells
Coating chamber slides
Have you ever experienced these problems with your cells on chamber slides?
  • Initial cell death after seeding
  • Detachment of cells during washing steps
  • Spontaneous differentiation
  • Unusual morphology and behavior of the cells
If you answer yes to any of the above, you may want to consider coating the chamber slides with specific ECM to promote cell adherence and cellular function.
Download the protocol
BioProbes Journal Issue 76
BioProbes Journal Issue 76
Browse the 76th issue of the BioProbes Journal, featuring articles around high content imaging and cell analysis, antibodies as research tools, and tools for protein analysis.
Read BioProbes 76
The Thermo Fisher Scientific education resource
Thermo Fisher Scientific education resource
With this free virtual learning platform, you can access webinars, posters, and papers about applications and techniques related to protein gel chemistries, western detection, mass spectrometry, flow cytometry, fluorescence imaging, high-content imaging, antibodies, immunoassays, and other topics.
Learn more, visit thermofisher.com/proteincelledu
View these informative on-demand webinars
Representative titles include:
  • Addressing the antibody reproducibility crisis: A panel discussion with key scientific leaders
  • Understanding cell reprogramming in treatment-resistant prostate cancer using organoids
  • Innovative quantitative imaging techniques and their utility in advancing neurobiology research
  • Five steps for publication-quality fixed-cell imaging the first time
  • What are the current advances in flow cytometry?
To access this and other webinars, visit themofisher.com/pcawebinars
Take the Antibody Validation* eLearning course today
Antibody Validation eLearning
Researchers need antibodies that bind to the right target and work predictably in their applications. Underperforming antibodies can lead to inconsistent results, a lack of reproducibility, and a waste of time and resources. To help ensure superior antibody results, Thermo Fisher Scientific expanded specificity and validation testing methodologies. We implemented a 2-part approach that involves target specificity verification and functional application validation*. 
In our four-module eLearning course, you’ll learn more about our antibody specificity testing approach, including detailed data, protocols and reporting information regarding antibody clone, isotype identity and functional characteristics to help facilitate the replication of published results.
Register to access the free course
*The use or any variation of the word “validation” refers only to research use antibodies that were subject to functional testing to confirm that the antibody can be used with the research techniques indicated. It does not ensure that the product, or products were validated for clinical or diagnostic uses.
NEWS AND VIEWS
Show us your Ab-moji with the antibody character digital stickers!
Abmoji
Now available in iMessage on iPhone and iPad
The wildly popular antibody character stickers are now available in digital format! Make your iMessages more fun when you “virtually peel and stick” our characters into your messages. Download the App for free via the App Store. 
Follow these 3 easy steps and start sharing today:
Abmoji step 1
Step 1
Search for and download the “Scientific Stickers” App from Thermo Fisher Scientific.
Abmoji step 2
Step 2
The App will then automatically appear in your App Tray. To access the “stickers” and place them in your iMessage, tap the “App Store” icon.
Abmoji troubleshoot
NOTE: If you don’t see it, ensure the App is enabled via Keyboards, which can be found within General Settings.
Abmoji
Step 3
Tap on the character you want in your message and send. Have fun!
If you need any assistance, contact us.
Abmoji step 1
Step 1
Search for and download the “Scientific Stickers” App from Thermo Fisher Scientific.
Abmoji step 2
Step 2
The App will then automatically appear in your App Tray. To access the “stickers” and place them in your iMessage, tap the “App Store” icon.
Abmoji troubleshoot
NOTE: If you don’t see it, ensure the App is enabled via Keyboards, which can be found within General Settings.
Abmoji
Step 3
Tap on the character you want in your message and send. Have fun!
If you need any assistance, contact us.