Lamin AC Antibodies, MA3-1000
Lamin A and C are A-type lamins that arise from alternative splicing of the LMNA gene. As part of the lamin class of nuclear intermediate filament proteins, lamin A and C are major structural components of the matrix on the inner surface of the nuclear envelope.
During mitosis, lamin proteins are phosphorylated, resulting in the reversible disassembly of the lamina matrix. Lamins are also involved in chromatin structure, gene expression, nuclear stability, and apoptosis. Because they are ubiquitously expressed nuclear proteins, lamin A and C are useful as loading controls.
Mutations in lamin A and C have been linked to several human diseases including Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy, limb-girdle muscular dystrophy, dilated cardiomyopathy, Charcot-Marie-Tooth disease, and Hutchinson-Gilford progeria syndrome.
A variety of quality Invitrogen lamin A/C antibodies are available for your research, and most have been extensively validated for western blotting and immunostaining controls.
See all lamin antibodies
Featured product data
Immunofluorescent analysis of Lamin A/C shows staining in U251 Cells.
Immunofluorescent analysis of Lamin A/C shows staining in U251 Cells. Lamin A/C (green), F-actin staining with phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with an antibody recognizing Lamin A/C (Cat. No. MA3-1000) at a dilution of 1:200 overnight at 4oC, washed with PBS and incubated with a DyLight 488-conjugated secondary antibody (Cat. No. 35552 for GAR; Cat. No. 35503 for GAM). Images were taken at 60x magnification.
Immunohistochemistry analysis of Lamin C on formalin-fixed paraffin embedded skin tissue.
Immunohistochemistry analysis of Lamin C on formalin-fixed paraffin embedded skin tissue.
Specific detection was accomplished using lamin C polyclonal antibody (Cat. No. PA1-5827) at a dilution of 1:100.
Annotated product references
Cat. No. MA3-1000 was used in immunocytochemistry to study the role of the HIV regulatory protein Vpr in G2 arrest. Virology (Oct 2012; 432: 444) "Perinuclear localization of the HIV-1 regulatory protein Vpr is important for induction of G2-arrest." Sörgel S,Fraedrich K,Votteler J,Thomas M,Stamminger T,Schubert U
Cat. No. MA3-1000 was used in western blot to investigate the effect of RANBP 16 and 17 on bHLH transcription factor E12 function. Journal of cellular biochemistry (Sep 2010; 111: 195) "Identification of RANBP16 and RANBP17 as novel interaction partners for the bHLH transcription factor E12." Lee JH,Zhou S,Smas CM
Need antibodies in bulk quantities or custom packaging?
Inquire now. Complete Request Form
Want us to produce a custom antibody for you?
Learn more. Custom Antibody Services