Peroxisome Marker Antibodies, PA5-18531
Peroxisomes are bound by a single membrane and can be found in most eukaryotic cells. They bud off of the endoplasmic reticulum and function primarily to enzymatically oxidize fatty acids and then catalyze the breakdown of hydrogen peroxide, a result of fatty acid degradation. In plant cells, peroxisomes assist chloroplasts in photorespiration and convert fatty acids to sugar. In mammals, they are critical in preserving the normal function of brain and lungs and play a role in energy metabolism.
Peroxisome marker antibodies detect proteins specific to peroxisomes and can aid in the study of the structure and dynamics of peroxisomes. Peroxisome marker antibodies can also help elucidate the role or roles a protein may play in the tasks that are performed by peroxisomes. Peroxisome marker antibodies are particularly useful in targeting peroxisomes in fixed cells. Quality Invitrogen peroxisome marker antibodies are available for a variety of research needs.
Peroxisome marker antibody targets
  • ABCD3
  • ACOX1
  • ACOT8
  • ATG7
  • ATG12
  • BAAT
  • CAT
  • DNM1L
  • FOXP2
  • HMGCL
  • MAVS
  • MLYCD
  • MUL1
  • PEX3
  • PEX5
  • PEX19
  • PEX26
  • PMP70
  • SLC25A17
  • SLC27A2
Featured product data
Immunofluorescent analysis of PMP70 using anti-PMP70 polyclonal antibody
Immunofluorescent analysis of PMP70 using anti-PMP70 polyclonal antibody (Cat. No. PA1-650) shows staining in NS-1 Cells.
Immunohistochemical analysis of Catalase in paraffin-embedded A549 xenograft using a Catalase polyclonal antibody 
Immunohistochemical analysis of Catalase in paraffin-embedded A549 xenograft using a Catalase polyclonal antibody (Cat. No. PA5-29183) at a dilution of 1:500.
Annotated product references
Cat. No. PA1-650 was used in western blot to study the sub-cellular localisation of beta-lipotropin and beta-endorphin. Endocrinology (Oct 2010; 151: 4801) "Peroxisomal localization of the proopiomelanocortin-derived peptides beta-lipotropin and beta-endorphin." Höftberger R,Kunze M,Voigtländer T,Unterberger U,Regelsberger G,Bauer J,Aboul-Enein F,Garzuly F,Forss-Petter S,Bernheimer H,Berger J,Budka H
Cat. No. PA1-650 was used in flow cytometry, immunohistochemistry and western blot to investigate peroxisome proliferation in frozen rat and primate livers by means of flow cytometry. International journal of toxicology (Mar 2006; 25: 41) "Flow cytometric assessment of peroxisome proliferation from frozen liver of fibrate-treated monkeys." Kwanyuen P,Witherspoon SM,Creech DR,Colton HM,Falls JG,Cariello NF