HCS image with synaptogenesis features shown as an overlay
Optimized, automated assays for HCS
This automated assay uses the co-localization of presynaptic and postsynaptic markers to identify synapses and correlate them with neuronal and neurite morphology. Neuronal morphology is captured using a nuclear stain to locate the cell and a choice of stains or immunofluorescent labels to detect the cell body and neurites. Synapses are located using two channels for co-localization and immunofluorescent detection with antibodies for presynaptic (e.g., synaptophysin) and postsynaptic (e.g., PSD95) markers.
Request a demo  Access HCA assay builder  Request a poster
Typical synaptogenesis assay
Fixed-cell immunofluorescent assay protocol.
Imaging mode
  • Widefield or confocal
  • 20x magnification
Automatically measured properties
  • Neurite count
  • Neurite length
  • Neurite branching
  • Presynaptic vesicles
  • Postsynaptic spots
  • Synapse number
Since analysis is performed during image acquisition, it is possible to acquire an optimized data set to ensure the number of selected neurons to satisfy your statistical needs.
Synaptogenesis assay images
Three-panel HCS synaptogenesis assay image showing stained cells and software overlays
Syantogenesis assay. (Left) A hippocampal primary neuron preparation was labeled using Invitrogen Hoechst 33342 stain (blue) and an Invitrogen Alexa Fluor 488 MAP2 antibody conjugate. (Center and Right) Synaptogenesis features were automatically identified using Thermo Scientific HCS Studio Software. Cell bodies are identified in blue, and spots are identified in pink. Neurites are identified in green, and overlaps between spots and neurites are labeled in yellow.
Sample experimental data
Four-panel graph of different synaptogenesis assay parameters
Dose-response plots showing the effect of glutamate and kainite concentration on rat hippocampal neurons using automated measurements of presynaptic intensity, postsynaptic intensity, vesicle counts, and synapse counts.
Resources
Instrument systems
HCS application notes
See citations
HCS reagents optimized for use with your HCS platform