Pacific Orange dye
Pacific Orange dye is a bright orange-fluorescent dye optimally excited by the 405 nm line of the violet laser and with excitation/emission at 400/551 nm. Conjugates of this dye are strongly fluorescent even at neutral pH.
Pacific Blue, Pacific Green and Pacific Orange dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm, 500 nm and 551 nm, respectively, facilitating three-color analysis.
We offer Pacific Orange dye conjugated to a variety of primary antibodies, secondary antibodies and streptavidin. In addition, reactive and protein labeling kits are available so that you can produce your own antibody conjugates or probes.
Pacific Orange dye dashboard
Initial brightness
Pacific Orange dye is an orange fluorophore used primarily for flow cytometry, optimally excited by the 405 nm violet laser line.  
Photostability in buffer
Pacific Orange dye products
We offer a broad range of Pacific Orange dye products including labeling kits, labeled antibodies, bioconjugates and more:
Pacific Orange™ dye
Fluorescence excitation and emission spectra of Pacific Orange dye.
Pacific Orange reactive dyes
Select dye-labeling chemistries for conjugation with the following reactive groups: 
Amine
Pacific Orange dye protein/antibody labeling kits
Select an optimized labeling kit for your target antibody or protein:
Target protein Kit Cat. No.
Site-specific antibody labeling
<1–20 µg mouse IgG1 Zenon Pacific Orange Mouse IgG1 Labeling Kit Z25256
Conventional antibody/protein labeling
100 µg antibody/protein Pacific Orange Antibody/Protein Labeling Kit P30014
1 mg antibody/protein Pacific Orange Antibody/Protein Labeling Kit P30016
Violet laser–compatible antibody conjugates
We offer Pacific Blue, Pacific Green and Pacific Orange dyes as well as Qdot probes for use with the violet laser. In addition to primary and secondary antibody conjugates of these dyes, we’ve developed kits for cell proliferation, immunophenotyping, dead cell discrimination, apoptosis, and cell cycle analysis—all aimed at helping you maximize the data you can get from your violet laser–equipped flow cytometer.
Learn more about Cell Health and Viability Assays for Flow Cytometry
probesonline-may-2013.par.96977.image.320.285.1.pol-may-2013-pacific-green-jpg
Excitation spectra (dotted lines) and emission spectra (solid lines) for Molecular Probes violet laser–excited Pacific Blue dye (blue), Pacific Green dye (green), and Pacific Orange dye (orange). The shaded area represents the emission bandpass filter (522/31) used for detection of the Pacific Green dye.
Resources
Molecular Probes Handbook
Cell stain tool
The significantly revised 11th Edition is the most complete fluorescence labeling and detection reference available. The Molecular Probes Handbook can help you achieve the best experimental results for your fluorescence labeling and detection experiments. View the handbook online or order your own copy.
Go to the Handbook online
Order your copy of the Handbook
Flow cytometry instrument controls and standards
Cell stain tool
Keep your flow cytometry running at peak efficiency with instrument set-up, calibration, and counting beads including alignment and linearity beads, compensation tools, and cell-counting beads.
Go to flow cytometry instrument controls
Cell stain tool
Cell stain tool
Stain your own cell by using our simulation tool to develop reproducible results with many of our signature fluorescent dyes. Develop your multiplexing strategy for antibody probes, reactive dyes, and counterstains to create your perfectly labeled fluorescent cell.
Go to the cell stain tool
Fluorescence SpectraViewer
thumbnail50x50.par.42353.image.50.50.1.thu-spectraviewer-jpg
Plot and compare spectra and check the spectral compatibility for common fluorophores including all of the fluorescent probes from Thermo Fisher Scientific. The Fluorescence SpectraViewer tool is formatted for both microscope imaging and flow cytometry applications.
Go to the Fluorescence SpectraViewer