geneart_tal_effector_search_and_design_tool
Optimal TALEN design at the touch of a button
Design custom TAL effectors with ease and precision: introducing our new Invitrogen™ GeneArt™ TAL Effector Search and Design Tool. Quickly search our database of >600,000 predesigned TALENs targeting human and mouse genes. Use our TALEN in silico tool to find the optimal TALEN sequence for your genome editing goals.   
  • Once you’ve selected the optimal TAL Effector designs, you can purchase your designer TALENs and other recommended products for genome editing directly from the web tool.
  • Use this interface to search our database of >600,000 predesigned TALENs.
  • GeneArt™ TALs are optimized for gene knockout and typically target the first three transcribed exons of each gene.
  • Search results will include the top six TALEN sequences, potential binding sites, exon maps with TALEN locations, and recommendations based on potential off-target effects for each sequence.
  • New The tool will also design primers to be used with our GeneArt Genomic Cleavage Detection Kit, which is used to assess cleavage efficiency and confirm edited loci.
Start designing ›
Before starting, ensure the following:
Is your browser supported? Thermo Fisher Cloud includes the latest data security and display features, requiring the use of one of the following browsers/versions:
Click on one of the links above if you need to download and install a compatible browser, and thank you for using Thermo Fisher Cloud.
Once you are in a supported browser, you will need to log in to Thermo Fisher Cloud using your existing username and password.  
  • You will be asked to verify your email address the first time you sign in. 
  • A verification request email will be sent to the email address on file for your online account.  
  • Check your spam folder if you don’t immediately receive this verification request.  
  • Future visits to the GeneArt TAL Effector Search and Design Tool will not require Thermo Fisher Cloud sign-in. 
  • Bookmark the page for future reference.
Thank you for using Thermo Fisher Cloud.
New? Get started here!
Press release
Need more help? Contact us.