flammable bottles storage
Thermo Scientific flammable material and explosion-proof refrigerators are designed to meet your safety requirements for cold storage of volatile material or materials in hazardous environments. Our units will meet your temperature, reliability, performance, and compliance needs, and more. All flammable material refrigerators and explosion-proof refrigerators meet NFPA and OSHA standards. Availability of some units may be limited in certain geographic regions.
Quick links
Value flammable material storage (FMS) refrigerators
These refrigerators are designed to be used for the storage of volatiles, compounds, and solvents where a spark-free internal cabinet environment is required. Household/commercial refrigerators and freezers are not suitable; they have components in their electrical and refrigeration systems that can trigger explosions of flammable air-vapor mixtures inside the unit. Thermo Scientific flammable storage laboratory refrigerators are available from under-counter models to full-height, freestanding options.
  5 cu. ft. FMS Refrigerator 20 cu. ft. FMS Refrigerator 20 cu. ft. FMS Corrosion-Resistant Refrigerator 50 cu. ft. FMS Refrigerator
  TS 5cu. ft. flammable material refrigerator TS 20cu. ft. flammable material refrigerator TS corrosion-resistant flammable material refrigerator TS 50cu. ft. flammable material refrigerator
Request a quote 05FREETSA;
115V/60Hz
20FREETSA;
115V/60Hz
20FRCETSA;
115V/60Hz
50FREETSA;
115V/60Hz
Availability North America only North America only North America only North America only
Compliance UL, NFPA1, OSHA1 cCSAus, NFPA1, OSHA1 cCSAus, NFPA1, OSHA1 UL, NFPA1, OSHA1
Temperature range 1° to 12°C 1° to 12°C 1° to 12°C 1° to 12°C
Capacity 5.5 cu. ft. (155.7L) 20.9 cu. ft. (591.8L) 20.9 cu. ft. (591.8L) 50 cu. ft. (1415.8L)
Plug type NEMA 5-15P NEMA 5-15P NEMA 5-15P NEMA 5-15P
User interface Exterior thermostat Exterior thermostat Exterior thermostat Exterior thermostat
Defrost Manual Manual Manual Manual
Security Built-in key lock Built-in key lock Built-in key lock NA
Interior cabinet
ABS plastic Epoxy coated steel Epoxy coated steel; corrosion resistant stainless steel interior door and shelves
Stainless steel
Interior (DxWxH) 18 x 20 x 28 in.
(45 x 51 x 71cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
27 x 46 x 59 in.
(67 x 116 x 150cm)
Exterior (LxWxH) 28 x 24 x 34 in.
(71 x 60 x 85cm)
36 x 34 x 73 in.
(91 x 86 x 186cm)
36 x 34 x 73 in.
(91 x 86 x 186cm)
34 x 52 x 77 in.
(87 x 132 x 196cm)
Shipping weight 120 lb. (54.5kg) 230 lb. (104.3kg) 230 lb. (104.3kg) 420 lb. (190.5kg)
  151L ES Series FMS Refrigerator 288L ES Series FMS Refrigerator 592L FMS Refrigerator 592L FMS Corrosion Resistant Refrigerator
  TS ES Flammable material refrigerator TS ES Flammable material refrigerator TS ES Flammable material refrigerator TS corrosion-resistant flammable material refrigerator
Request a quote 151R-AXV-TS;
230V/50Hz
288R-AXV-TS;
230V/50Hz (EU)
288R-AXW-TS;
230/50Hz (UK)
20FREETSV;
230V/50Hz
20FRCETSV;
230V/50Hz
Availability Not available in North America nor other 60Hz regions
Not available in North America nor other 60Hz regions Not available in North America nor other 60Hz regions Not available in North America nor other 60Hz regions
Compliance CE, ATEX2 CE, ATEX2    
Temperature range 1° to 11°C 1° to 11°C 1° to 12°C 1° to 12°C
Capacity 151 L 232 L 591.8 L 591.8 L
Plug type Continental EU Continental EU NEMA 6-15P NEMA 6-15P
User interface Integrated digital controller
Integrated digital controller Exterior thermostat Exterior thermostat
Defrost Manual Manual Manual Manual
Security Standard door locks, door ajar alarm
Standard door locks, door ajar alarm Built-in key lock Built-in key lock
Interior cabinet
HIPS/ABS mix
(High-Impact Polystyrene and Acrylonitrile—Butadiene—Styrene)
HIPS/ABS mix
(High-Impact Polystyrene and Acrylonitrile—Butadiene—Styrene)
Epoxy-coated steel Epoxy-coated steel; corrosion resistant stainless steel interior door and shelves
Interior (DxWxH) 45 x 50 x 71 cm 48 x 44 x 130 cm 60 x 71 x 147 cm 60 x 71 x 147 cm
Exterior (LxWxH) 60 x 60 x 85 cm 64 x 60 x 157 cm 91 x 86 x 186 cm 91 x 86 x 186 cm
Shipping weight 79 lbs. (36 kg) 128 lbs. (58 kg) 230 lb. (1043 kg) 230 lb. (1043 kg)
2 Certified spark-free units are designed, evaluated, and comply with the ATEX Directive 94/9/EC Annex VIII. These units have an intrinsically safe (IS) barrier installed in the temperature probe circuit with no other internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. The internal wall construction has been reviewed and is not a source for electrostatic buildup due to the materials used in construction. ATEX-certified spark-free units are not designed for use in Class 1, Zone 0, or Zone 1 applications which require an explosion-proof unit.
  5 cu. ft. FMS Refrigerator 20 cu. ft. FMS Refrigerator 20 cu. ft. FMS Corrosion-Resistant Refrigerator 50 cu. ft. FMS Refrigerator
  TS 5cu. ft. flammable material refrigerator TS 20cu. ft. flammable material refrigerator TS corrosion-resistant flammable material refrigerator TS 50cu. ft. flammable material refrigerator
Request a quote 05FREETSA;
115V/60Hz
20FREETSA;
115V/60Hz
20FRCETSA;
115V/60Hz
50FREETSA;
115V/60Hz
Availability North America only North America only North America only North America only
Compliance UL, NFPA1, OSHA1 cCSAus, NFPA1, OSHA1 cCSAus, NFPA1, OSHA1 UL, NFPA1, OSHA1
Temperature range 1° to 12°C 1° to 12°C 1° to 12°C 1° to 12°C
Capacity 5.5 cu. ft. (155.7L) 20.9 cu. ft. (591.8L) 20.9 cu. ft. (591.8L) 50 cu. ft. (1415.8L)
Plug type NEMA 5-15P NEMA 5-15P NEMA 5-15P NEMA 5-15P
User interface Exterior thermostat Exterior thermostat Exterior thermostat Exterior thermostat
Defrost Manual Manual Manual Manual
Security Built-in key lock Built-in key lock Built-in key lock NA
Interior cabinet
ABS plastic Epoxy coated steel Epoxy coated steel; corrosion resistant stainless steel interior door and shelves
Stainless steel
Interior (DxWxH) 18 x 20 x 28 in.
(45 x 51 x 71cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
27 x 46 x 59 in.
(67 x 116 x 150cm)
Exterior (LxWxH) 28 x 24 x 34 in.
(71 x 60 x 85cm)
36 x 34 x 73 in.
(91 x 86 x 186cm)
36 x 34 x 73 in.
(91 x 86 x 186cm)
34 x 52 x 77 in.
(87 x 132 x 196cm)
Shipping weight 120 lb. (54.5kg) 230 lb. (104.3kg) 230 lb. (104.3kg) 420 lb. (190.5kg)
  151L ES Series FMS Refrigerator 288L ES Series FMS Refrigerator 592L FMS Refrigerator 592L FMS Corrosion Resistant Refrigerator
  TS ES Flammable material refrigerator TS ES Flammable material refrigerator TS ES Flammable material refrigerator TS corrosion-resistant flammable material refrigerator
Request a quote 151R-AXV-TS;
230V/50Hz
288R-AXV-TS;
230V/50Hz (EU)
288R-AXW-TS;
230/50Hz (UK)
20FREETSV;
230V/50Hz
20FRCETSV;
230V/50Hz
Availability Not available in North America nor other 60Hz regions
Not available in North America nor other 60Hz regions Not available in North America nor other 60Hz regions Not available in North America nor other 60Hz regions
Compliance CE, ATEX2 CE, ATEX2    
Temperature range 1° to 11°C 1° to 11°C 1° to 12°C 1° to 12°C
Capacity 151 L 232 L 591.8 L 591.8 L
Plug type Continental EU Continental EU NEMA 6-15P NEMA 6-15P
User interface Integrated digital controller
Integrated digital controller Exterior thermostat Exterior thermostat
Defrost Manual Manual Manual Manual
Security Standard door locks, door ajar alarm
Standard door locks, door ajar alarm Built-in key lock Built-in key lock
Interior cabinet
HIPS/ABS mix
(High-Impact Polystyrene and Acrylonitrile—Butadiene—Styrene)
HIPS/ABS mix
(High-Impact Polystyrene and Acrylonitrile—Butadiene—Styrene)
Epoxy-coated steel Epoxy-coated steel; corrosion resistant stainless steel interior door and shelves
Interior (DxWxH) 45 x 50 x 71 cm 48 x 44 x 130 cm 60 x 71 x 147 cm 60 x 71 x 147 cm
Exterior (LxWxH) 60 x 60 x 85 cm 64 x 60 x 157 cm 91 x 86 x 186 cm 91 x 86 x 186 cm
Shipping weight 79 lbs. (36 kg) 128 lbs. (58 kg) 230 lb. (1043 kg) 230 lb. (1043 kg)
2 Certified spark-free units are designed, evaluated, and comply with the ATEX Directive 94/9/EC Annex VIII. These units have an intrinsically safe (IS) barrier installed in the temperature probe circuit with no other internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. The internal wall construction has been reviewed and is not a source for electrostatic buildup due to the materials used in construction. ATEX-certified spark-free units are not designed for use in Class 1, Zone 0, or Zone 1 applications which require an explosion-proof unit.
Top
High-performance flammable storage refrigerators
Our high-performance manual defrost FMS refrigerators are designed to meet stringent US Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids.1 Each model is listed by UL as a special purpose refrigerator or freezer intended for flammable material storage in accordance with NFPA 45 and 99.1
  Revco Forma Thermo Scientific
  Revco FMS HP refrigerator Forma FMS HP refrigerator TSHP FMS HP refrigerator
Request a quote
RFMS2305A;
115V/60Hz
FFMS2305A;
115V/60Hz
TSFMS2305A;
115V/60Hz
Availability North America only North America only North America only
Compliance UL, NFPA1, OSHA1 UL, NFPA1, OSHA1 UL, NFPA1, OSHA1
Temperature range 1°C to 8°C 1°C to 8°C 1°C to 8°C
Capacity 23 cu. ft. (651L) 23 cu. ft. (651L) 23 cu. ft. (651L)
User interface Microprocessor controller Microprocessor controller Microprocessor controller
Access port & casters
1-inch port, casters standard 1-inch port, casters standard 1-inch port, casters standard
Chart recorder
Factory installed option (North America only) Factory installed option
(North America only)
Factory installed option (North America only)  
Defrost Manual Manual Manual
Security Lockable doors, audio/visual alarms Lockable doors, audio/visual alarms Lockable doors, audio/visual alarms
Interior cabinet
Ignition-free, scratch-resistent painted steel Ignition-free, scratch-resistent painted steel Ignition-free, scratch-resistent painted steel
Interior (DxWxH) 29 x 24 x 58 in.
(73.7 x 61 x 147.3cm)
29 x 24 x 58 in.
(73.7 x 61 x 147.3cm)
29 x 24 x 58 in.
(73.7 x 61 x 147.3cm)
Exterior (LxWxH) 37 x 28 x 78.6 in.
(94 x 72 x 200cm)
37 x 28 x 78.6 in.
(94 x 72 x 200cm)
37 x 28 x 78.6 in.
(94 x 72 x 200cm)
Shipping weight 425 lbs. (193kg) 425 lbs. (193kg) 425 lbs. (193kg)
1Products complying with the applicable requirements of NFPA 70, "National Electric Code", NPFA 45, "Fire Protection for Laboratories Using Chemicals," and NFPA 99 "Health Care Facilities" are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.
Top
Explosion-proof refrigerators
Spark-free interiors and exteriors reduce the risk of explosion. These lab refrigerators are designed to be used in Class 1, Groups C and D, and Class 1, Zone 1, Group IIB areas where ignitable materials are normally present. Thermo Scientific explosion-proof laboratory refrigerators are particularly suited for safe storage in hazardous environments. Models are available from under-counter to full height, freestanding.
  5.5 cu. ft. Explosion-Proof Refrigerator 20.9 cu. ft. Explosion-Proof Refrigerator 20.9 cu. ft. Explosion-Proof Corrosion Resistant Refrigerator
  TS small explosion proof refrigerator TS explosion proof refrigerator TS corrosion-resistant explosion proof refrigerator
Request a quote 05EREETSA;
115V/60Hz
20EREETSA;
115V/60Hz
20ERCETSA;
115V/60Hz
Availability North America only North America only North America only
Compliance UL, NFPA1 cCSAus cCSAus
Temperature range 1°C to 12°C 1°C to 12°C 1°C to 12°C
Capacity 5.5 cu. ft. (155.7L) 20.9 cu. ft. (592L) 20.9 cu. ft. (592L)
Plug type Hardwired to prevent sparking and accidental disconnection Hardwired to prevent sparking and accidental disconnection Hardwired to prevent sparking and accidental disconnection
User interface Thermostat and compressor components in vapor-proof enclosure Thermostat and compressor components in vapor-proof enclosure Thermostat and compressor components in vapor-proof enclosure
Defrost Manual Manual Manual
Security Built-in key lock Built-in key lock Built-in key lock
Interior cabinet
ABS plastic ABS plastic ABS plastic
Interior (DxWxH) 18 x 20 x 28 in.
(45 x 51 x 71cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
Exterior (LxWxH) 28 x 24 x 34 in.
(71 x 60 x 85cm)
34 x 34 x 78 in.
(82 x 86 x 186cm)
34 x 34 x 78 in.
(82 x 86 x 186cm)
Shipping weight 120 lbs. (54.5kg) 283 lbs. (128kg) 283 lbs. (128kg)
  20.9 cu. ft. Explosion-Proof Refrigerator 20.9 cu. ft. Explosion-Proof Corrosion Resistant Refrigerator
  TS explosion proof refrigerator TS corrosion-resistant explosion proof refrigerator
Request a Quote 20EREETSV;
230V/50Hz
20ERCETSV;
230V/50Hz
Availability Not available in North America nor other 60Hz regions Not available in North America nor other 60Hz regions
Compliance cCSAus  
Temperature range 1°C to 12°C 1°C to 12°C
Capacity 20.9 cu. ft. (592 L) 20.9 cu. ft. (592 L)
Plug type Hardwired to prevent sparking and accidental disconnection Hardwired to prevent sparking and accidental disconnection
User interface Thermostat and compressor components in vapor-proof enclosure Thermostat and compressor components in vapor-proof enclosure
Defrost Manual Manual
Security Built-in key lock Built-in key lock
Interior cabinet
ABS plastic ABS plastic
Interior (DxWxH) 24 x 28 x 58 in.
(60 x 71 x 147cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
Exterior (LxWxH) 35 x 34 x 73 in.
(88 x 86 x 186 cm)
35 x 34 x 73 in.
(88 x 86 x 186cm)
Shipping Weight 283 lbs. (128kg) 283 lbs. (128kg)
  5.5 cu. ft. Explosion-Proof Refrigerator 20.9 cu. ft. Explosion-Proof Refrigerator 20.9 cu. ft. Explosion-Proof Corrosion Resistant Refrigerator
  TS small explosion proof refrigerator TS explosion proof refrigerator TS corrosion-resistant explosion proof refrigerator
Request a quote 05EREETSA;
115V/60Hz
20EREETSA;
115V/60Hz
20ERCETSA;
115V/60Hz
Availability North America only North America only North America only
Compliance UL, NFPA1 cCSAus cCSAus
Temperature range 1°C to 12°C 1°C to 12°C 1°C to 12°C
Capacity 5.5 cu. ft. (155.7L) 20.9 cu. ft. (592L) 20.9 cu. ft. (592L)
Plug type Hardwired to prevent sparking and accidental disconnection Hardwired to prevent sparking and accidental disconnection Hardwired to prevent sparking and accidental disconnection
User interface Thermostat and compressor components in vapor-proof enclosure Thermostat and compressor components in vapor-proof enclosure Thermostat and compressor components in vapor-proof enclosure
Defrost Manual Manual Manual
Security Built-in key lock Built-in key lock Built-in key lock
Interior cabinet
ABS plastic ABS plastic ABS plastic
Interior (DxWxH) 18 x 20 x 28 in.
(45 x 51 x 71cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
Exterior (LxWxH) 28 x 24 x 34 in.
(71 x 60 x 85cm)
34 x 34 x 78 in.
(82 x 86 x 186cm)
34 x 34 x 78 in.
(82 x 86 x 186cm)
Shipping weight 120 lbs. (54.5kg) 283 lbs. (128kg) 283 lbs. (128kg)
  20.9 cu. ft. Explosion-Proof Refrigerator 20.9 cu. ft. Explosion-Proof Corrosion Resistant Refrigerator
  TS explosion proof refrigerator TS corrosion-resistant explosion proof refrigerator
Request a Quote 20EREETSV;
230V/50Hz
20ERCETSV;
230V/50Hz
Availability Not available in North America nor other 60Hz regions Not available in North America nor other 60Hz regions
Compliance cCSAus  
Temperature range 1°C to 12°C 1°C to 12°C
Capacity 20.9 cu. ft. (592 L) 20.9 cu. ft. (592 L)
Plug type Hardwired to prevent sparking and accidental disconnection Hardwired to prevent sparking and accidental disconnection
User interface Thermostat and compressor components in vapor-proof enclosure Thermostat and compressor components in vapor-proof enclosure
Defrost Manual Manual
Security Built-in key lock Built-in key lock
Interior cabinet
ABS plastic ABS plastic
Interior (DxWxH) 24 x 28 x 58 in.
(60 x 71 x 147cm)
24 x 28 x 58 in.
(60 x 71 x 147cm)
Exterior (LxWxH) 35 x 34 x 73 in.
(88 x 86 x 186 cm)
35 x 34 x 73 in.
(88 x 86 x 186cm)
Shipping Weight 283 lbs. (128kg) 283 lbs. (128kg)
Top
Is there a safer way to store potentially flammable, pharmaceutical-grade materials that require refrigerated or frozen temperatures?
Thermo Scientific FMS high-performance refrigerators and freezers meet industry protocol for the storage of critical, potentially flammable, pharmaceutical-grade materials or samples containing trace amounts of flammables that could cause a spark.
Learn more
Resources
Flammable material storage
Explosion proof storage
Support