Benchtop Centrifuges
Choose from a growing family of benchtop centrifuges designed to meet the needs of your specific applications. With careful attention to your space, process and protocol requirements, our line of centrifuges stand out for their performance, reliability, consistency and safety. Whether performing clinical protocols, cell culture procedures, microplate processing, or anything in between, Thermo Scientific centrifuges fulfill a variety of separation needs.
Browse products below or let one of our experts help you make a selection.
Ask an expert
Quick links
Which type of benchtop centrifuge fits your application best?
Compact 
Routine
High performance
  compact benchtop centrifuge routine benchtop centrifuge high-performance benchtop centrifuge
Our small benchtop centrifuges offer maximized capacity in a compact footprint, accommodating multi-laboratory settings with the flexibility to adapt to your evolving needs.
High safety, solid reliability and consistent results.
Our routine benchtop centrifuges are designed for mid-performance needs of basic research and clinical applications, often requiring repetitive and consistent protocols. With a focus on quality, throughput and reliability, they provide a robust solution to less technical applications.
Safe, simple comfortable operation with optimized performance.
Our high performance benchtop centrifuges are ideal for applications requiring more flexibility, speed and programming. A wider array of rotor choices, higher speeds and up to 99 stored programs ensure compatibility with a wide array of protocols, even across multiple applications.
Versatility and unparalleled processing performance with the same safety and durability.
Max capacity 4 x 145mL
4 x 400mL (1.6L)
4 x 1000mL (4L)
4 x 400mL (1.6L)
4 x 1000mL (4L)
Max speed 17,850rpm 15,200rpm 15,200rpm
Max RCF 30,279 x g 25,830 x g 25,830 x g
Number of rotors 14 optional rotors 10 optional rotors (1.6L)
12 optional rotors (4L)
14 optional rotors (1.6L)
17 optional rotors (4L)
Dimensions (H x W x D) Ventilated:
12.2 x 14.6 x 18.9 in.
(31 x 37 x 48cm)
Refrigerated:
12.6 x 18.1 x 26.4 in.
(32 x 46 x 67cm)
1.6L:
Ventilated:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Refrigerated:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
4L:
Ventilated:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Refrigerated:
14.2 x 29.3 x 26.4 in.
(36 x 74.5 x 67cm)
X1 (1.6L):
Ventilated:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Refrigerated:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
XT/XF/X3 (4L):
Ventilated:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Refrigerated:
14.2 x 29.3 x 26.4 in.
(36 x 74.5 x 67cm)
Floor standing:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67cm)
Display type LCD LCD LCD
  Learn more Learn more 
Learn more
Looking for a benchtop ultracentrifuge?
We also have the high performance Sorvall MTX 150 Bench Micro-Ultracentrifuge for rapid small-volume processing applications with acceleration rates that reach max speed in just 80 seconds.
  • Max capacity 6 x 30mL or 4 x 7mL
  • Speeds up to 150,000rpm
  • Max RCF 1,048,680 x g
Top
Benchtop centrifuges videos
Top
How does Auto-Lock rotor exchange facilitate rotor placement and improve safety, compared with traditional rotor tie-down systems?
With Auto-Lock rotor exchange, you can have the confidence that your rotor is safely and securely locked in place, and have the flexibility to change rotors and applications with one push of a button. 
  • No tools
  • No complicated rotor changing procedures
  • Swap between protocols instantly
  • Quick and efficient cleaning
Open Smart Note
Top
Related centrifuge categories
Microcentrifuges
Floor models
Centrifuge rotors
Centrifuge ware
Resources
Support
  • Unity Lab Services
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.