routine benchtop centrifuges
Designed to improve performance and endurance, our routine benchtop centrifuges come in medium (1.6L) and large (4L) capacities and offer a mid-performance drive. They are ideal for applications that need solid reliability and high throughput, such as:
  • Clinical chemistry
  • Clinical microbiology
  • Hematology
  • Immunology
  • Clinical studies
  • Cellular biology
  • Microbiology
  • Genomics/Molecular biology
  • Proteomics
  • Biochemistry
  • Pharmaceutical studies
Browse products below or let one of our experts help you make a selection.
Ask an expert
Innovative Thermo Scientific rotor technologies
clickseal biocontainment lids
One-handed sample protection, biocontainment certified*
Glove-friendly, one-handed open/close capability of our ClickSeal biocontainment lids provide simple operation for all laboratory users, eliminating multi-turn screw caps and complicated high pressure clips.
auto-lock rotor exchange
Secure, push-button rotor exchange in as little as 3 seconds
Trouble-free rotor installation and removal with Auto-Lock rotor exchange provides full chamber access for cleaning convenience, quick switch between applications, and the flexibility to evolve with the needs of your lab. No tools required. No complicated rotor changing procedures.
fiberlite carbon fiber rotors
Outstanding durability, ergonomics and performance
Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal. Plus, their corrosion- and fatigue-resistance secures the rotor’s structural integrity for outstanding durability, and it's backed by a 15-year limited warranty.**
Quick links
Which type of routine benchtop centrifuge fits your needs best?
routine benchtop centrifuge rotor
Choose 1.6 liter capacity models ›
Easy to use with intuitive controls and a design for easy sample loading, unloading, and cleaning.
  • Max capacity 4 x 400mL
  • Max speed 15,200rpm
  • Max RCF 25,830 x g
  • 12 optional rotors
routine benchtop centrifuge
Choose 4 liter capacity models › 
Spin 4 liters in a traditional 3-liter footprint when you combine our 4L benchtop centrifuges with the TX-1000 rotor.
  • Max capacity 4 x 1000mL
  • Max speed 15,200rpm
  • Max RCF 25,830 x g
  • 10 optional rotors
Medifuge-small-benchtop-centrifuge-230x100
Order online
Perform both routine and clinical life science separations with a quiet, compact design that stands out with the unique, "2-in-1" hybrid design of the Thermo Scientific DualSpin rotor. Allows 8 interchangeable fixed angle and swinging buckets.
  • Max capacity 8 x 15mL
  • Max speed 4,900rpm
  • Max RCF 3,490 x g
  • Comes as a package with 8 each fixed angle and swinging buckets and spacers
1.6L (4 x 400mL) benchtop centrifuges
Ventilated
Refrigerated
  routine benchtop centrifuge routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  routine benchtop centrifuge routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  routine benchtop centrifuge routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  routine benchtop centrifuge routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  routine benchtop centrifuge routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  routine benchtop centrifuge routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Top
4L (4 x 1000mL) benchtop centrifuges
Ventilated
Refrigerated
  ventilated routine benchtop centrifuge refrigerated routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
5,300rpm (swinging bucket)
15,200rpm (fixed angle)
5,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
5,590 x g (swinging bucket)
25,830 x g (fixed angle)
5,590 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated routine benchtop centrifuge refrigerated routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
5,300rpm (swinging bucket)
15,200rpm (fixed angle)
5,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
5,590 x g (swinging bucket)
25,830 x g (fixed angle)
5,590 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated routine benchtop centrifuge refrigerated routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
5,300rpm (swinging bucket)
15,200rpm (fixed angle)
5,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
5,590 x g (swinging bucket)
25,830 x g (fixed angle)
5,590 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated routine benchtop centrifuge refrigerated routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
5,300rpm (swinging bucket)
15,200rpm (fixed angle)
5,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
5,590 x g (swinging bucket)
25,830 x g (fixed angle)
5,590 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated routine benchtop centrifuge refrigerated routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
5,300rpm (swinging bucket)
15,200rpm (fixed angle)
5,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
5,590 x g (swinging bucket)
25,830 x g (fixed angle)
5,590 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated routine benchtop centrifuge refrigerated routine benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
5,300rpm (swinging bucket)
15,200rpm (fixed angle)
5,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
5,590 x g (swinging bucket)
25,830 x g (fixed angle)
5,590 x g (swinging bucket)
Programs 6 direct program buttons 6 direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
Display type LCD LCD
*Biocontainment certification by Public Health England Porton Down, UK.
**Subject to Thermo Fisher Scientific’s standard limited warranty and country and dealer participation.
Top
Rotors for routine benchtop centrifuges
Our extensive rotor portfolio provides application flexibility to support multiple lab settings, multiple users, and versatility as the needs of your laboratory evolve.
Swinging bucket rotors
routine benchtop centrifuge swinging bucket rotors
Rotors for 1.6L capacity centrifuges
Rotors for 4L capacity centrifuges
Rotors for 1.6L and 4L capacity centrifuges
Fixed angle rotors
routine benchtop centrifuge fixed angle rotors
Rotors for 1.6L capacity centrifuges
Rotors for 4L capacity centrifuges
Rotors for 1.6L and 4L capacity centrifuges
Top
Fiberlite carbon fiber rotors brochure
Thermo Scientific Fiberlite rotors accelerate productivity while offering unequaled durability. They will maximize your centrifuge performance with versatility, speed and a robust corrosion-free design. Backed by a 15-year warranty, the most comprehensive warranty coverage available.*  
  • Improved ergonomics for added safety
  • Lightweight design for faster acceleration and deceleration
  • Corrosion resistance for easier care and maintenance
  • Fatigue-resistant design for longer life
Open brochure
* Subject to Thermo Fisher Scientific’s standard limited warranty. Ask your sales representative for details or review our general terms and conditions of sale on our Terms of Use page.
Top
Take a virtual tour of our benchtop centrifuges
Take a self-guided tour and learn how our centrifuges and rotors are designed around you and your applications.
They fit in to: They stand out for:
  • your space
  • your processes and protocols
  • the evolving needs of your
    future in the lab
  • innovative design and technologies
  • solid reliability
  • consistent results
While in the tour, take a look at our convenient packages to find just the right centrifuge, rotor, and adapter combination you need to get started.
Take a tour
Top
Benchtop centrifuges videos
Top
Related products for routine benchtop centrifuges
Microcentrifuges
Floor models
Centrifuge rotors
Centrifuge ware
Resources
Support
  • Unity Lab Services
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.