Ultracentrifuges
Fast, high productivity separations with built-in safety and efficiency
Combining outstanding speed, safety and ergonomics in a compact design, our ultracentrifuges and micro-ultracentrifuges are designed to deliver exceptional performance and versatility for a variety of applications.
Browse products below or let one of our experts help you make a selection.
Ask an expert
ultracentrifuges
Designed for simplicity and ease-of-use, using carbon fiber rotors; plus our WX+ accepts most ultra rotors from other manufacturers.
micro-ultracentrifuges
Quick, small, simple, and powerful, on the bench and on the floor. Innovative software gives you more options for greater flexibility.
Quick links
Which fits your needs best?
Ultracentrifuge
Micro-ultracentrifuge
Especially ideal for labs where space is limited. Open rotor system allows flexibility to use most ultra rotors from other major manufacturers.
  • 37 rotors support labware from 1.5mL microtubes to 250mL bottles
  • RCF up to 802,000 x g
  • Available in 80,000, 90,000, and 100,000rpm models
Advanced functionality in a compact footprint, readily accessible with flexibility for a full range of small volume processing applications, and fast accel/decel rates.
  • 17 rotors with capacity from 4 x 2.2mL up to 6 x 30mL; supports tubes from 0.2mL to 20mL
  • RCF up to 1,048,000 x g
  • Available in 120,000 and 150,000rpm models
View all View all
Sorvall WX+ Ultracentrifuge Series
Maximize space in your lab with the small-footprint Thermo Scientific Sorvall WX+ ultracentrifuge series while maintaining high-performance speeds of up to 100,000rpm.
These are highly flexible centrifuge solutions for nucleic acid or proteomic applications, including exosome research, available in three top performance ranges, and includes carbon-fiber rotor advantages.
  Sorvall WX 80+ Ultracentrifuge Sorvall WX 90+ Ultracentrifuge Sorvall WX 100+ Ultracentrifuge
Request a quote
208 to 240V, 50/60Hz
Single phase
208 to 240V, 50/60Hz
Single phase
200 to 240V, 50/60Hz
Single phase
Max capacity 24 x 1.5mL microtubes to 6 x 250mL bottles 24 x 1.5mL microtubes to 6 x 250mL bottles 24 x 1.5mL microtubes to 6 x 250mL bottles
Max RCF 802,000 x g
(T-8100 rotor)
692,149 x g
(T-890 rotor)
546,883 x g
(T-890 rotor)
Max speed 80,000rpm 90,000rpm 100,000rpm
Rotors 38 optional rotors 38 optional rotors 38 optional rotors
Dimensions
(H x W x D)
Lid closed:
34.6 x 31.1 x 27.2 in.
(88 x 79 x 69cm)
Lid closed:
34.6 x 31.1 x 27.2 in.
(88 x 79 x 69cm)
Lid closed:
34.6 x 31.1 x 27.2 in.
(88 x 79 x 69cm)
Display type Touch screen color LCD panel Touch screen color LCD panel Touch screen color LCD panel
Performance range Speed 1,000-80,0000rpm
RCF 66,993-802,000 x g
Speed 1,000-90,000rpm
RCF 66,993-692,149 x g
Speed 1,000-100,000rpm
RCF 66,993-546,883 x g 
Weight 859.8 lb. (390kg) 859.8 lb. (390kg) 859.8 lb. (390kg)
Top
Sorvall MTX and MX+ Micro-Ultracentrifuge Series
Excellent performance with RCF up to 1,048,000 x g and versatility for rapid small-volume processing applications, our Thermo Scientific Sorvall MTX and MX+ microultracentrifuges feature advanced functionality in a compact footprint.
  Sorvall MTX 150 Bench Micro-Ultracentrifuge Sorvall MX 150 Plus Floor Micro-Ultracentrifuge Sorvall MX 120 Plus Floor Micro-Ultracentrifuge
  Ideal for labs with limited space and the need to run high performance ultra applications. Maximize productivity with impressive acceleration rates that reach max speed in just 80 seconds. Ideal for labs looking for a smaller solution but need to maximize bench space. All the capability of the benchtop MTX 150 in a floor-standing model with a very small footprint.   More economical floor-standing micro-ultracentrifuge solution with the same large, versatile rotor selection, but with a lower top speed.
Request a quote
Max capacity 6 x 30mL (S50A rotor) or
4 x 7mL (S50-ST rotor)
6 x 30mL (S50A rotor) or
4 x 7mL (S50-ST rotor)
6 x 30mL (S50A rotor) or
4 x 7mL (S50-ST rotor)
Max RCF 1,048,680 x g
(S140-AT rotor)
1,048,680 x g
(S140-AT rotor)
770,460 x g
(S140-AT rotor)
Max speed 150,000rpm 150,000rpm 120,000rpm
Rotors 17 optional rotors 17 optional rotors 17 optional rotors
Dimensions 
(H x W x D)
Lid closed:
16.1 x 23.2 x 22.9 in.
(40.8 x 59 x 58.2cm)
Lid closed:
33.5 x 17.3 x 19.3 in.
(85 x 44 x 49cm)
Lid closed:
33.5 x 17.3 x 19.3 in.
(85 x 44 x 49cm)
Display type Touch sensitive LCD panel Touch sensitive LCD panel Touch sensitive LCD panel
Performance range 1,000 to 150,0000rpm
RCF 201,046 to 1,048,680 x g
1,000 to 150,0000rpm
RCF 201,046 to 1,048,680 x g
1,000 to 120,0000rpm
RCF 201,046 to 770,460 x g
Weight 213 lb. (97kg) 231 lb. (105kg) 231 lb. (105kg)
Top
Rotors for ultracentrifuges
fiberlite-carbon-fiber-rotors-100x100
Fiberlite carbon fiber rotors
Fatigue-resistant, corrosion resistant, and lightweight for a safer work environment.
Self-locking-rotor-100x100
Self-locking rotor system
Automatically secures rotor to the drive shaft without manual intervention, facilitating installation and added safety.
extensive-rotor-sample-types-100x100
Versatility across sample types
Obtain application flexibility with extensive range of rotor formats that accommodate multiple tube volumes.
open-rotor-system-100x100
Open-rotor system
Optimize your inventory with flexibility of accepting ultra rotors from other major manufacturers. (Sorvall WX+ Ultracentrifuge)
Fixed angle rotors
Neo Angle Rotors
Continuous Flow Rotors
Swinging bucket rotors
Vertical rotors
Zonal Rotors
Fixed Angle Rotors
Swinging Bucket Rotors
Vertical Rotors
Top
Sorvall WX+ Ultracentrifuge Brochure
Ultracentrifugation simplified at every turn. Maximize space in your lab with the small-footprint Thermo Scientific Sorvall WX+ ultracentrifuge series while maintaining high-performance speeds of up to 100,000 rpm.
  • Lightweight, fatigue-resistant carbon fiber rotors
  • Intuitive, easily accessible touch screen
  • Automatic tube balancing
  • Quiet performance (<51 dBA)
  • Access controls for multi-user environments
  • LED Stat Light for across-the-lab visibility of status
Learn more
Technical note: Separation of influenza virus with our S50-A fixed angle rotor and a Sorvall micro-ultracentrifuge
influenza-virus-278x195
In this technical note, we describe a protocol for the separation of influenza virus using the S50-A fixed angle rotor and the Sorvall MTX 150 Micro-Ultracentrifuge or the Sorvall MX Micro-Ultracentrifuge. The Thermo Scientific S50-A fixed angle rotor provides the largest capacity in the Thermo Scientific micro-ultracentrifuge family and allows sample processing up to 180 mL.
Open tech note
Top
Related products for ultracentrifuges
Benchtop centrifuges
Microcentrifuges
High speed centrifuges
Centrifuge ware
Top
Resources
Support
  • Unity Lab Services:
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.