Accessories for Orion benchtop and portable meters
Enhance your productivity with accessories for benchtop and portable meters
Enhance your productivity while protecting your investment. We offer an assortment of meter accessories for our benchtop and portable meters.
Meter accessory quick links:
Featured benchtop meter accessories
Complement your benchtop meter with key accessories that make you more productive.
Stirrer Probes 
Uniformly mix solutions using five selectable stirring speeds with the meter-controlled Orion Star stirrer probe. Designed to fit into benchtop electrode stands, this stirrer probe can be easily positioned and quickly rinsed between samples. Our premium Versa Star Pro benchtop meters can operate two stirrer probes simultaneously, so you can measure your samples faster.
Meter-Attached Electrode Stand
Included with every Thermo Scientific Orion Star A and Versa Star Pro benchtop meter, the electrode stand holds a variety of sensors for easy placement in and out of samples, and allows for consolidated rinsing between samples. 
Featured portable meter accessories
Protection and organization for your portable meter.
Portable Meter Armor
Because anything can happen in the field, add extra protection with durable portable meter armor. Included with all Orion Star A portable meter kits, the armor features dedicated holders for pH, conductivity, and dissolved oxygen sensors. An adjustable wrist/belt strap can secure the meter to fixtures such as pipes. The strap also converts into a built-in meter stand.
Hard-sided Field Case
Our field case is specially designed to efficiently transport everything you need for field measurements. Sensors can stay connected with the meter while in the case. A removable door makes it easy to use the cable management system. The field case, which is included with all Orion Star A portable meter kits, helps you stay organized while protecting your instrumentation.
Increase your productivity by adding accessories to your benchtop or portable meter
Accessory category Cat No. Description Meter type Compatibility
Electrode holders & stands
090043 Orion Swing Arm and Electrode Holder Orion Dual Star, Star A111, Star A211, Star A214, Star A215, Star A216 and Versa Star Pro Meters
090070 Orion Vertical Electrode Holder and Stand Orion Dual Star, Star A111, Star A211, Star A214, Star A215, Star A216 and Versa Star Pro Meters
STARA-BEA Orion Star A Series Benchtop Meter-Attached Stand Orion Dual Star, Star A111, Star A112, Star A113, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216 and Versa Star Pro Meters
STARA-ELHD Orion Star A Series Electrode Holder for Stands Orion 090043 and STARA-BEA Stands
STARA-HB Orion Star A Series Weighted Base Orion 090043 and STARA-BEA Stands
Field carrying cases and armor 1210005 Orion Portable Soft-Sided Field Case Orion Star A121, Star A122, Star A123, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters
STARA-AR Orion Star A Series Portable Meter Protective Armor Orion Star A121, Star A122, Star A123, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters
STARA-CS Orion Star A Series Portable Meter Hard Field Case Orion Star A121, Star A122, Star A123, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters
STARA-ESCD Orion Star A Series Portable Holder for Conductivity & DO Probes Orion STARA-AR Meter Armor
STARA-ESPH Orion Star A Series Portable Holder for pH Electrode Orion STARA-AR Meter Armor
Interface cables and computer software 1010004 Orion RS232 Computer and Printer Cables for Star A Series Meters Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters
1010005 Orion RS232-to-USB Cable and Adapter for Star A Series Meters Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters
1010053 Orion RS232 Computer Cable for Star A Series Meters Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters
VSTAR-NPCD Orion Navigator Pro Software on CD for Versa Star Pro Meters Orion Versa Star Pro Meters
VSTAR-NPUSB Navigator Pro Software on USB for Versa Star Pro Meters Orion Versa Star Pro Meters
Power adapters and replacement parts 1010003 Orion Universal Power Adapter for Star A Meters Orion Dual Star, Star A111, Star A112, Star A113, Star A121, Star A122, Star A123, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters
STARA-PWR Orion Universal Power Adapter for Versa Star Pro Meters Orion Versa Star Pro Meters
090045 Orion BNC Shorting Plug for pH Meters Orion Dual Star, Star A111, Star A121, Star A211, Star A214, Star A215, Star A216, Star A221, Star A321, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro pH Meters
Printers and accessories STARA-106 Orion Star A Series Ink Ribbon Printer 110–240V Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters
STARA-108 Orion Ink Ribbons for Star A Series Printer, 6 Pack Orion STARA-106 Printer
STARA-109 Orion Paper Rolls for Star A Series Printer, 5 Pack Orion STARA-106 Printer
1010008 Orion Ink Cartridge for 1010006 Inkjet Printer Orion 1010006 Printer
1010009 Orion Paper Roll for 1010006 Inkjet Printer Orion 1010006 Printer
PRT300 Orion PRT300 Printer with Cable, 110V Legacy Orion Aplus and PerpHecT Meters
PRT301 Orion PRT300 Printer with Cable, 220V Legacy Orion Aplus and PerpHecT Meters
PRT302 Orion Printer Ribbon for PRT300 Printers Orion PRT 300 Printer
  Find the pH benchtop or portable meter you need for your measurements. Orion pH meter selector tool
See our complete list of offerings for your measurement needs
Benchtop
Meters
benchtop meters
Portable
Meters
portable meters
Electrodes and Probes
electrodes and probes
Buffers and Solutions
buffers and solutions
Choose the best electrode for your application
This interactive Orion electrode selection guide will help you to find the perfect electrode for your application based on your requirements.
Find your electrode
Orion meter brochure
The measure of confidence
Download the Orion solutions brochure to learn about our premium, advanced, and basic meters and help identify the best match based on your required features, performance specifications, and budget.
Request brochure
Resources
Support
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.
Find your software