iBind Western System
The original notebook-sized automated western blot processing device
Simply load your primary antibody, secondary antibody, and wash solutions into the device and then walk away. The Invitrogen iBind Western System automatically performs all immunoblotting steps using sequential lateral flow technology, a simple form of capillary action. In less than 3 hours, the blot is ready for final detection. You can continue to use your existing chromogenic, chemiluminescent, or fluorescent western blot detection protocols, along with your choice of primary antibody or secondary antibody conjugates of HRP, AP, or fluorescent dyes.
  • Antibody savings—use up to 80% less primary antibody than with traditional tray-based incubation steps for western blotting
  • Load and go—processes solutions using sequential lateral flow technology, with no batteries, shakers, trays, or timers required
  • Reproducibility—automated processing enables improved consistency among blots
Request a demoOrder nowDownload manual
See what customers have to say about the iBind western system
Watch the iBind video demo
Quick one-time setup
With iBind devices, you simply load all your solutions and walk away!
Sample loading wells for the iBind western system
Row / Well Solution
1 Primary antibody
2 1X iBind / iBind FD Solution
3 Secondary antibody
4 1X iBind / iBind FD Solution
Example data
Robust results with less primary antibody
One of the key elements of a successful western blot is the primary antibody; however, this reagent also contributes to over 90% of the total cost of the blot. Because the iBind Western System is more sensitive than manual processing methods, you can use less antibody and achieve similar results.
*Results may vary. The protocol for primary antibody use is 80% less than a traditional manual method.
less primary antibody
Using chemiluminescent or fluorescence-based detection methods, you can use up to 80% less primary antibody than manual methods, saving you significant cost per blot. A 2-fold dilution series of EGF receptor control cell lysate (30 µg, 15 µg, 7.5 µg, 3.75 µg, and 1.875 µg) was used. Proteins were separated using Bolt Bis-Tris Plus gels on the Mini Gel Tank and transferred to PVDF membranes using the iBlot 2 Dry Blotting System. The blots were probed with an anti–phospho-EGF receptor [Tyr1068] (1H12) mouse monoclonal antibody (1:1,000 dilution, equated to 2 µL antibody for the iBind device method and 10 µL antibody for the manual method) followed by a goat anti–mouse IgG (H+L) peroxidase-conjugated antibody (1:360 for iBind device processing (5.55 µL); 1:1,800 for manual method (5.55 µL)). The standard iBind solution kit was used for the chemiluminescence blot (panel C, manual processing and panel D, iBind system processing); the fluorescence detection (panel A, manual processing and panel B, iBind processing) kit was used for fluorescence detection. These results demonstrate that western blots processed on the iBind device show comparable results to those obtained when western blots are processed manually, with lower overall primary antibody requirements for the iBind device.
Excellent western performance compared to manual blotting
  • The iBind System offers greater sensitivity compared to manual methods for many monoclonal and polyclonal antibodies.
  • Combine the iBind Western System with highly specific primary and secondary antibodies to achieve cleaner western blots.
  • Automated processing enables improved blot-to-blot consistency with CVs typically less than 5% (compared to manual processing that can have CVs of 13%).
 iBind System
Western blots processed on the Invitrogen iBind device show excellent sensitivity compared to western blots processed manually. (A–B) Western blots with phosphorylated Akt (left to right: 30 µg–500 ng cell lysate load) were processed either on the iBind device or using standard manual western processing protocols as specified by the antibody manufacturer (monoclonal anti–phospho-Akt [pT308] (C31E5E) primary antibody; HRP-conjugated anti-rabbit secondary antibody). The blot processed with the iBind device detected phospho-Akt in 500 ng of cell lysate, while the target was detected in 4 µg on the manually processed blot. (C–D) Western blots with cell lysate expressing CREB (left to right: 30 µg–1 µg cell lysate load) were processed either on the iBind device or using standard manual western processing protocols as specified by the antibody manufacturer (polyclonal anti-CREB primary antibody; HRP-conjugated anti-rabbit secondary antibody). The blot processed with the iBind system detected CREB in 6 µg of cell lysate, while 10 µg of lysate was needed to detect CREB on the manually processed blot. For all blots, proteins were separated using Bolt Bis-Tris Plus gels on the Mini Gel Tank and transferred to PVDF membranes using the iBlot 2 Dry Blotting System.
Invitrogen iBind system
Invitrogen iBind system vs. manual western blotting with fluorescence-based detection of c-Jun and phospho-Stat3. Western blots of cell lysates containing phospho-Stat3 and c-Jun (left to right: 30 µg–120 ng lysate protein) were processed either on the iBind device or using standard manual western processing protocols as specified by the manufacturer for each antibody (monoclonal phospho-Stat3 [pY705] (M9C6) mouse primary antibody and monoclonal c-Jun (60A8) rabbit primary antibody). (A), (C) Alexa Fluor 680 goat anti-rabbit secondary antibody and Alexa Fluor 790 goat anti-mouse secondary antibody. (B), (D) IRDye 680LT goat anti-rabbit secondary antibody and IRDye 800CW goat anti-mouse secondary antibody. Blots processed with the iBind device detected both target proteins at lower levels than manually processed blots. For all blots, proteins were separated using the Invitrogen Bolt Gel Electrophoresis System and transferred to NC membranes using the Invitrogen iBlot 2 Dry Blotting System.
Robust results with less primary antibody
One of the key elements of a successful western blot is the primary antibody; however, this reagent also contributes to over 90% of the total cost of the blot. Because the iBind Western System is more sensitive than manual processing methods, you can use less antibody and achieve similar results.
*Results may vary. The protocol for primary antibody use is 80% less than a traditional manual method.
less primary antibody
Using chemiluminescent or fluorescence-based detection methods, you can use up to 80% less primary antibody than manual methods, saving you significant cost per blot. A 2-fold dilution series of EGF receptor control cell lysate (30 µg, 15 µg, 7.5 µg, 3.75 µg, and 1.875 µg) was used. Proteins were separated using Bolt Bis-Tris Plus gels on the Mini Gel Tank and transferred to PVDF membranes using the iBlot 2 Dry Blotting System. The blots were probed with an anti–phospho-EGF receptor [Tyr1068] (1H12) mouse monoclonal antibody (1:1,000 dilution, equated to 2 µL antibody for the iBind device method and 10 µL antibody for the manual method) followed by a goat anti–mouse IgG (H+L) peroxidase-conjugated antibody (1:360 for iBind device processing (5.55 µL); 1:1,800 for manual method (5.55 µL)). The standard iBind solution kit was used for the chemiluminescence blot (panel C, manual processing and panel D, iBind system processing); the fluorescence detection (panel A, manual processing and panel B, iBind processing) kit was used for fluorescence detection. These results demonstrate that western blots processed on the iBind device show comparable results to those obtained when western blots are processed manually, with lower overall primary antibody requirements for the iBind device.
Excellent western performance compared to manual blotting
  • The iBind System offers greater sensitivity compared to manual methods for many monoclonal and polyclonal antibodies.
  • Combine the iBind Western System with highly specific primary and secondary antibodies to achieve cleaner western blots.
  • Automated processing enables improved blot-to-blot consistency with CVs typically less than 5% (compared to manual processing that can have CVs of 13%).
 iBind System
Western blots processed on the Invitrogen iBind device show excellent sensitivity compared to western blots processed manually. (A–B) Western blots with phosphorylated Akt (left to right: 30 µg–500 ng cell lysate load) were processed either on the iBind device or using standard manual western processing protocols as specified by the antibody manufacturer (monoclonal anti–phospho-Akt [pT308] (C31E5E) primary antibody; HRP-conjugated anti-rabbit secondary antibody). The blot processed with the iBind device detected phospho-Akt in 500 ng of cell lysate, while the target was detected in 4 µg on the manually processed blot. (C–D) Western blots with cell lysate expressing CREB (left to right: 30 µg–1 µg cell lysate load) were processed either on the iBind device or using standard manual western processing protocols as specified by the antibody manufacturer (polyclonal anti-CREB primary antibody; HRP-conjugated anti-rabbit secondary antibody). The blot processed with the iBind system detected CREB in 6 µg of cell lysate, while 10 µg of lysate was needed to detect CREB on the manually processed blot. For all blots, proteins were separated using Bolt Bis-Tris Plus gels on the Mini Gel Tank and transferred to PVDF membranes using the iBlot 2 Dry Blotting System.
Invitrogen iBind system
Invitrogen iBind system vs. manual western blotting with fluorescence-based detection of c-Jun and phospho-Stat3. Western blots of cell lysates containing phospho-Stat3 and c-Jun (left to right: 30 µg–120 ng lysate protein) were processed either on the iBind device or using standard manual western processing protocols as specified by the manufacturer for each antibody (monoclonal phospho-Stat3 [pY705] (M9C6) mouse primary antibody and monoclonal c-Jun (60A8) rabbit primary antibody). (A), (C) Alexa Fluor 680 goat anti-rabbit secondary antibody and Alexa Fluor 790 goat anti-mouse secondary antibody. (B), (D) IRDye 680LT goat anti-rabbit secondary antibody and IRDye 800CW goat anti-mouse secondary antibody. Blots processed with the iBind device detected both target proteins at lower levels than manually processed blots. For all blots, proteins were separated using the Invitrogen Bolt Gel Electrophoresis System and transferred to NC membranes using the Invitrogen iBlot 2 Dry Blotting System.
Get your iBind Western System Starter Kit now
wf11827-ibind-image
Begin using a simpler, more elegant western blotting process. The iBind Starter Kit includes a complete iBind Western Device, plus cards and a solution kit, so you can get started right away:
  • iBind Western System
  • iBind Cards (1 box of 10)
  • iBind Solution Kit (Note: fluorescence detection requires FD Solution Kit)
Order starter kit