Complete growth medium
Component Cat. No.
Gibco DMEM, with GlutaMAX Supplement 61870036
10% Gibco FBS A3160401
1.0 mM Gibco Sodium Pyruvate 11360070
Proper culture techniques and procedures are an essential part of ensuring successful transfection. Subculturing, also referred to as passaging, is the removal of medium and transfer of cells from a culture into fresh growth medium, in order to propagate the cells.
See our product list for this protocol
Brightfield image of A549 cells in culture
Brightfield image of A549 cells in culture, prior to transfection
Passaging
  • Maintain cells in T-75 flasks.
  • Use Gibco TrypLE dissociation reagent.
  • Passage cells every 3–4 days to ensure that they do not enter senescence.
  • Transfection of cells should be performed only between passages 5 and 25 post-thaw.
  • If designing an experiment that involves transfection, ensure that setup coincides with a cell passage.
  • Plate cells for transfection only 1 day before the experiment.
Seeding cells for transfection
  • The day before transfection, dissociate cells that are 80–90% confluent in a T-75 flask.
  • Count the cells using standard trypan blue exclusion.
    • ––Important: The cell number and concentration determined can vary significantly depending on what method is used for counting; it is important to be consistent and use a single method throughout an experiment.
  • The cell culture must have >90% viability and be 70% confluent on the day of transfection.
    • ––Important: If cells are not at the right confluence, do not wait until the next day to perform transfection, as this can significantly affect transfection efficiency.
  • Seed 1.05 x 105 cells in 500 μL growth medium for a single well of a 24-well plate.
Transfection protocol
Component Cat. No.
Invitrogen Lipofectamine 3000 Transfection Reagent L3000008
Gibco Opti-MEM I Reduced Serum Medium 31985062
Thermo Scientific Nunc Multidish, 24-Wells, Cell Culture Treated 142475
On the day of transfection, which should be 1 day following cell plating, perform the following steps, which have been optimized for a single well of a 24-well plate using Invitrogen Lipofectamine 3000 Transfection Reagent:
Step Tube Complexation components Amount per well
(24-well)
1 Tube 1 Opti-MEM I medium 25 μL
Lipofectamine 3000 reagent 1.5 μL
2 Tube 2 Opti-MEM I medium 25 μL
DNA amount (DNA concentration should be 0.5–5 μg/μL) 500 ng
P3000™ reagent 1 μL
3 Add tube 2 solution to tube 1 and mix well
4 Incubate mixture from step 3 at room temperature for 10–15 min
5 Add 50 μL of complex from step 4 to cells;
gently swirl plate to ensure homogeneous distribution of complex to the entire well
Transfection efficiency analysis
At 48 hr following transfection of a GFP reporter construct, cells were evaluated via microscopy and flow cytometry. To assess transfection efficiency, cells were first visualized via fluorescence microscopy for qualitative assessment of protein expression, morphology, and viability (Figure 1). Cells were then prepared for flow cytometry by aspirating the medium and replacing it with 250 μL of a 7:3 mixture of TrypLE reagent:1X DPBS. Cells were incubated at 37°C for 10 min and then pipetted up and down to ensure single cells for flow cytometry analysis.
Posttransfection analysis of A549 fluorescence cells 
Posttransfection analysis of A549 bright-field cells 
Figure 1. Posttransfection analysis of cells. (A) Fluorescence and (B) bright-field images demonstrating 57–60% transfection efficiency.
Tips and tricks
  • Decreasing the serum content of the culture medium (to <10%) at the time of transfection is acceptable, but replace with complete growth medium within 4–24 hr post-transfection.
  • Antibiotics can be used during transfection.
  • Prior to flow cytometry, visualize cells under a bright-field microscope to verify dissociation following incubation with TrypLE reagent.
Scaling up or down Lipofectamine 3000 reagent transfections
Use the following table to scale the volumes for your transfection experiment. The most common sizes are listed below.
Culture vessel Multiplication factor* Shared reagents DNA transfection siRNA transfection
Growth medium Opti-MEM medium for complexing DNA P3000 reagent Lipofectamine 3000 reagent** siRNA Lipofectamine 3000 reagent**
96-well 0.2 100 μL 2 x 5 μL 100 ng 0.1 µL 0.3 µL 3 pmol 0.3 µL
48-well 0.5 250 μL 2 x 12.5 μL 0.25 µg 0.5 µL 0.75 µL 7.5 pmol 0.75 µL
24-well 1 500 μL 2 x 25 μL 0.5 µg 1 µL 1.5 µL 15 pmol 1.5 µL
12-well 2 1 mL 2 x 50 μL 1 µg 2 µL 3 µL 30 pmol 3 µL
6-well 5 2 mL 2 x 125 μL 2.5 µg 5 µL 7.5 µL 75 pmol 7.5 µL
60 mm 11.05 5 mL 2 x 250 μL 5.5–11 µg 11–22 µL 16.5 µL 166 pmol 17 µL
10 cm 28.95 10 mL 2 x 500 μL 14–28 µg 28–56 µL 43 µL 434 pmol 43 µL
T-75 39.47 15 mL 2 x 750 μL 20–40 µg 40–80 µL 59 µL 592 pmol 59 µL
T-175 92.11 35 mL 2 x 1.75 mL 46–96 µg 92–180 µL 138 µL 1,382 pmol 138 µL
* After determining the optimum reagent amount, use the multiplication factor to determine the reagent amount needed for your new plate format.
** Optimum amount needed is determined from the protocol for Lipofectamine 3000 Transfection Reagent.
Ordering information
Gibco Lung Cancer Starter Kit
For your convenience, the essential components of this protocol are now available in the Gibco Lung Cancer Starter Kit. The kit includes: basal medium, FBS, Lipofectamine 3000 reagent, Opti-MEM medium, and TrypLE reagent. The kit is available at thermofisher.com/cancercellculture. For additional components required for the protocol see ordering table below.
Find out more at thermofisher.com/3000
Download PDF version