This uniquely designed software from Dynal completes the RELI™ SSO automation package, providing a fast and accurate results interpretation and management system.
Automated Data input
PMP receives strip image directly from Dynal RELI-Scan™ Instrument or a flat bed scanner. Accurate results interpretation with no manual data entry. High throughput analysis with reduced interpretation time.
Positive Probe Assignment
Each probe has an individual signal intensity cut off value for accurate assignment of positive and negative signals.
Hierarchical Result Display
Provides clear and flexible result display with no data loss
Complete and Partial Matched Types
Displays complete matchesOffers alternative partial matches Unknown reactions do not affect analysis.
Results Integration
Software combines data from multiple tests to produce a single result. Software recommends typing kit solutions to resolve ambiguities.