MiniFiler™ Getting Started
To prepare your lab for the use of the MiniFiler™ Kit, you will need:
The ability to run 5-dye applications – The MiniFiler™ Kit uses the same size standard, matrix standard and capillary electrophoresis modules as Identifiler®, Yfiler® and SEfiler™ Kits.
- GeneMapper® ID Software v3.2 – Data from the MiniFiler™ Kit can not be analyzed with GeneScan®/Genotyper® Software
- New panel and bin set for GeneMapper® ID software