Power Snap Image
File Name: update111.zip
Version: 1.0.4 Device, 1.1.1 Camera
Date Published: Sept 2017
Language: English
Download Size: 5.1 MB
Download Upgrade
Requirements
  • Firmware Upgrade File
  • USB thumb drive
  • E-Gel Power Snap Device (G8100) and/or Camera Hood (G8200)
Instructions
This procedure is a step-by-step guide for updating both the E-Gel Power Snap Electrophoresis Device and E-Gel Power Snap Camera firmware to the latest version release. Firmware updates are necessary at times when there is a need to add new features or a critical bug fix is required. The updating process will take around 15-20 minutes to complete.
Preparation
  1. Insert the USB thumb drive to your PC’s USB port.
    Note: The USB disk’s file system should be in FAT32. Please reformat the disk if necessary. Please remove previous firmware upgrade files prior to starting.
  2. Download the software update ZIP file from the hyperlink above (requires internet access).
  3. Copy the downloaded file from step 2 to USB thumb drive. Extract the contents of the ZIP file to USB thumb drive’s root directory as shown in Panel 1.
  4. Updates cannot be executed by connecting instrument directly to a computer, a USB device is required to update firmware.
saving-file
Panel 1
E-Gel Power Snap Electrophoresis Device Software Update
  1. Connect the power adapter to the device and power ON the instrument.
  2. Insert thumb drive in the USB port located at the back of the device, as shown below (Panel 1).
USB-back
Panel 1
  1. From home screen, navigate to ‘Settings > Software update’ as shown in Panels 2A and 2B.
home
Panel 2A
1b_press
Panel 2B
  1. Press and release ‘Software update’ button. The application will start searching for the update files in the USB thumbdrive. The following screen is displayed if a valid update is found. Press and release the ‘Update’ button (Panel 3).
2_searchingUSB
Panel 3. The base module will automatically reboot aftrer copying the update files to the instrument’s internal storage.
3_update_found
4_updating
4b_updating
4c_rebooting
  1. The screen in Panel 4 will be displayed while software installation is in progress. The electophoresis device module software installation will take approximately 1-2 minutes, please wait.
    Important Note: DO NOT POWER OFF THE INSTRUMENT AT THIS TIME.
bootloader
Panel 4. The instrument will restart after software installation is complete.
home
  1. Power cycle the instrument by turning the power switch OFF and then ON again after a few seconds. From the home screen, navigate to software version page by pressing ‘Settings > About instrument’ to check the software version (Panel 5A and 5B).
settings
Panel 5A
version
Panel 5B
  1. Device software update is complete.
E-Gel Power Snap Camera Software Update
  1. Dock the E-Gel Power Snap Camera Device on the E-Gel Power Snap Device.
  2. Insert the update thumb drive (Panel 1A) in the USB port. Wait at least 3-5 seconds for the USB thumb drive to initialize. A complete initialization is indicated by a displayed USB icon at the lower left of the home screen, as shown in the screenshot in Panel 1B.
USB-front
Panel 1A
home_a
Panel 1B
  1. Navigate to ‘Settings’ screen by pressing the gear icon located at the upper right of the home screen.
  2. Press and release ‘Update software’ button. The application will start searching for the update files in the USB thumbdrive. A confirmation screen is displayed when the correct software update files are found (Panel 2).
1_settings_a
Panel 2
4_updating_a
  1. Press and release the ‘Update’ button in the confirmation screen. The application will start copying the update files from the USB thumb drive to the instrument’s internal storage (Panel 3).
3_updates_found
Panel 3
4_updating_a
  1. Press and release ‘Restart’ button to reset the instrument and start the installation process (Panel 4).
5_restart
Panel 4
  1. The following screen will be displayed while software installation is in progress. The camera module software installation will take around 3-5 minutes, please wait (Panel 5).
    Important Note: DO NOT POWER OFF THE INSTRUMENT AT THIS TIME.
cam_bootloader
Panel 5
home_a
  1. Power cycle the device by redocking the camera module. From the home screen, navigate to ‘Settings > About instrument’ and check the software version (Panel 6).
1_settings_a
Panel 6
about_page
  1. Device software update is complete.
Preparation
  1. Insert the USB thumb drive to your PC’s USB port.
    Note: The USB disk’s file system should be in FAT32. Please reformat the disk if necessary. Please remove previous firmware upgrade files prior to starting.
  2. Download the software update ZIP file from the hyperlink above (requires internet access).
  3. Copy the downloaded file from step 2 to USB thumb drive. Extract the contents of the ZIP file to USB thumb drive’s root directory as shown in Panel 1.
  4. Updates cannot be executed by connecting instrument directly to a computer, a USB device is required to update firmware.
saving-file
Panel 1
E-Gel Power Snap Electrophoresis Device Software Update
  1. Connect the power adapter to the device and power ON the instrument.
  2. Insert thumb drive in the USB port located at the back of the device, as shown below (Panel 1).
USB-back
Panel 1
  1. From home screen, navigate to ‘Settings > Software update’ as shown in Panels 2A and 2B.
home
Panel 2A
1b_press
Panel 2B
  1. Press and release ‘Software update’ button. The application will start searching for the update files in the USB thumbdrive. The following screen is displayed if a valid update is found. Press and release the ‘Update’ button (Panel 3).
2_searchingUSB
Panel 3. The base module will automatically reboot aftrer copying the update files to the instrument’s internal storage.
3_update_found
4_updating
4b_updating
4c_rebooting
  1. The screen in Panel 4 will be displayed while software installation is in progress. The electophoresis device module software installation will take approximately 1-2 minutes, please wait.
    Important Note: DO NOT POWER OFF THE INSTRUMENT AT THIS TIME.
bootloader
Panel 4. The instrument will restart after software installation is complete.
home
  1. Power cycle the instrument by turning the power switch OFF and then ON again after a few seconds. From the home screen, navigate to software version page by pressing ‘Settings > About instrument’ to check the software version (Panel 5A and 5B).
settings
Panel 5A
version
Panel 5B
  1. Device software update is complete.
E-Gel Power Snap Camera Software Update
  1. Dock the E-Gel Power Snap Camera Device on the E-Gel Power Snap Device.
  2. Insert the update thumb drive (Panel 1A) in the USB port. Wait at least 3-5 seconds for the USB thumb drive to initialize. A complete initialization is indicated by a displayed USB icon at the lower left of the home screen, as shown in the screenshot in Panel 1B.
USB-front
Panel 1A
home_a
Panel 1B
  1. Navigate to ‘Settings’ screen by pressing the gear icon located at the upper right of the home screen.
  2. Press and release ‘Update software’ button. The application will start searching for the update files in the USB thumbdrive. A confirmation screen is displayed when the correct software update files are found (Panel 2).
1_settings_a
Panel 2
4_updating_a
  1. Press and release the ‘Update’ button in the confirmation screen. The application will start copying the update files from the USB thumb drive to the instrument’s internal storage (Panel 3).
3_updates_found
Panel 3
4_updating_a
  1. Press and release ‘Restart’ button to reset the instrument and start the installation process (Panel 4).
5_restart
Panel 4
  1. The following screen will be displayed while software installation is in progress. The camera module software installation will take around 3-5 minutes, please wait (Panel 5).
    Important Note: DO NOT POWER OFF THE INSTRUMENT AT THIS TIME.
cam_bootloader
Panel 5
home_a
  1. Power cycle the device by redocking the camera module. From the home screen, navigate to ‘Settings > About instrument’ and check the software version (Panel 6).
1_settings_a
Panel 6
about_page
  1. Device software update is complete.
Resources
Nucleic acid education
Learn about gel electrophoresis basics, workflows, considerations, applications, and troubleshooting in separation and analysis of nucleic acids.
Related products
Support
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification and analysis applications.
Product support documentation
Search for manuals, protocols, Material Safety Data Sheets, product literature and certificates by catalog number or product name.
Contact us
Email or call our Technical Application Scientists for additional questions regarding DNA ladders