mySpin mini centrifuge
Mini centrifuge rotors to maximize speed and flexibility
Thermo Scientific mySPIN 12 and 6 mini centrifuge rotors enable fast spins up to 12,500 rpm and are ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2mL) for customized use.
Get more information
Bring increased efficiency and user comfort to your lab
The mySPIN 12 and 6 mini centrifuges, providing 12- or 6-place rotors with tool-free, quick rotor exchange for ease-of-use, are adaptable for microfilter cell separation and HPLC protocols as well as PCR, clinical and diagnostic applications. Spin 0.2mL and 1.5/2.0mL tubes with mySPIN centrifuge rotors, and ensure continued, uninterrupted operation with mySPIN centrifuge replacement parts.
75004083 12 x 1.5/2.0mL Rotor
75004084 32 x 0.2mL Rotor
50144572 Rotor Cover for 12 x 1.5/2.0mL Rotor
75004082 Power Adapter
20190362 Universal Power Cord
50144571 Rotor Knob
50146145 Tube Adapter Inserts for 0.2/0.5mL
75004062 6 x 1.5/2.0mL Circular Rotor
75004063 16 x 0.2mL Rotor
75004064 Power Adapter and Universal Power Cord Set
20290729 Replacement O-ring
50146145 Tube Adapter Inserts for 0.2/0.5mL
mySPIN mini centrifuge series brochure
Download brochure
Fits in.
  • To your space, with its compact footprint
  • To your applications, with its flexibility
  • To your timeline, with quick-spin options
Stands out.
  • Standard safety features provide assurance
  • Efficient and easy to use with ergonomic design, duplicate spin function and tool-free rotors
Related mini centrifuge rotors categories
mySPIN 12 and 6 mini centrifuges are designed for quick and pulse spins, and offer convenience at your personal workstation. Tool-free rotors, duplicate spin function, and ergonomic design promote both efficiency and ease of use.
Use our compact, safe and easy-to-use microcentrifuges to support your micro-volume protocols. They combine power with versatility and convenience, and can accelerate your sample preparation processes with RCF up to 17,000 xg or 21,100 xg.
Choose from a broad range of microcentrifuge tubes designed to meet the most rigorous requirements of centrifugation and daily laboratory use. They're ideal for pure protection and security during sample preparation, transfer and storage.
Resources
Support
Unity Lab Services 
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.