SFCA Membrane Filter
SFCA Membrane Filters do not contain surfactants or wetting agents as compared to CA filters, thereby improving downstream cleanliness for applications requiring low protein binding, like cell culture media and sera. 
Pore size quick reference chart
0.65/0.8μm 0.45μm 0.2μm 0.1μm
Analysis of mold, fungi and yeast, stabilization of fermented product, clarification and prefiltration Analysis of bacteria, sterility testing, clarification and prefiltration
Sterilization, sterility testing, “final filtration”
Reduction of mycoplasma, ultracleaning
SFCA Membrane Materials Reference
Application Material Properties Flow Rate Throughput Extractables Autoclave Protein Binding
Cell Culture media
Serum
Molecular biology solutions
Hydrophilic
Moderate to High Moderate Low Yes, wet, liquid cycle Low
Select the best SFCA Membrane for your application
To find the membrane you need, select by one of the column categories to sort the products. Search is also available for another way to find your membrane. The select button will direct you to the product specifications.
No records were found matching your criteria
Catalog # Pore Size Volume Format  
122-0020 0.2μm 115mL Filter Unit Select
155-0020 0.2μm 150mL Filter Unit Select
156-4020 0.2μm 500mL Filter Unit Select
157-0020 0.2μm 250mL Filter Unit Select
158-0020 0.2μm 1000mL Filter Unit Select
161-0020 0.2μm 1000mL Filter Unit Select
162-0020 0.2μm 500mL Filter Unit Select
290-3320 0.2μm 150mL Bottle Top Filter Select
290-4520 0.2μm 150mL Bottle Top Filter Select
291-3320 0.2μm 500mL Bottle Top Filter Select
291-4520 0.2μm 500mL Bottle Top Filter Select
292-3320 0.2μm 1000mL Bottle Top Filter Select
292-4520 0.2μm 1000mL Bottle Top Filter Select
723-2520  0.2µm 10mL-100mL Syringe Filters  Select
723-9920  0.2µm 10mL-100mL Syringe Filters  Select
724-2020  0.2µm 10mL-100mL Syringe Filters  Select
122-0045  0.45µm 115mL Filter Unit Select
155-0045  0.45µm 150mL Filter Unit Select
156-4045  0.45µm 500mL Filter Unit Select
157-0045  0.45µm 250mL Filter Unit Select
158-0045  0.45µm 1000mL Filter Unit Select
161-0045  0.45µm 1000 mL Filter Unit Select
162-0045  0.45µm 500mL Filter Unit Select
290-3345  0.45µm 150mL Bottle Top Filter Select
290-4545  0.45µm 150mL Bottle Top Filter Select
291-3345  0.45µm 500mL Bottle Top Filter Select
291-4545  0.45µm 500 mL Bottle Top Filter Select
723-2545  0.45µm 10mL-100mL Syringe Filter Select
723-9945  0.45µm 10mL-100mL Syringe Filter Select
724-2045  0.45µm 10mL-100mL Syringe Filter Select