ART Universal Pipette Tips
Tips that fit any pipette in your lab
Thermo Scientific ART pipette tips have been a trusted tool in the lab for decades. The ART portfolio offers a complete selection of barrier, non-filtered, and specialty tips, making it easier than ever to find a tip to fit virtually any brand of pipette in your laboratory and deliver the quality, performance, and reliability you expect.
Introducing SoftFit-L LTS for your Rainin specific needs.
View selection tool ›
What can ART barrier tips do that traditional filter tips can't?
Only ART self-sealing barrier tips seal completely to block the passage of aerosols, liquids, radioactive isotopes, and biological materials, preventing contamination of the pipette and cross contamination of subsequent samples in an experiment. No traditional filter can do that.
No contamination
Traditional filter tips cannot stop the flow of contaminants like the ART barrier. ART provides complete protection.
Low retention technology
Low retention ART tip technology
ART barrier tips are available in low-retention tips that deliver the maximum sample volume for the best results.
Test them for yourself, take the ART challenge 
Green packaging
ART reload green system
 Discover greener alternatives with ART packaging options that save space and generate less waste.
 Learn more   
Uncompromised purity
The ART tip portfolio offers tips with exceptional clarity and uncompromised purity. All ART pipette tips are certified to be free of RNase, DNase, DNA, ATP, and endotoxins.
Choose the right ART tip for your pipette and application
Self-sealing barrier tips
Filtered tip
ART barrier pipette tips are ideal for use with your most valuable samples and sensitive assays to prevent contamination, while protecting your pipettes, too.
Recommended uses:
  • PCR
  • DNA and RNA purification
  • Pipetting radioactive agents
  • Protein biology
  • Cell culture
  • Biobanking
Non-filtered tips
Non-filtered tip
ART non-filtered tips are the same high-quality tips as ART barrier tips.
Recommended uses:
  • Non-critical samples and assays
SoftFit-L filtered tips
SoftFit-L Tips
ART SoftFit-L filtered tips block aerosol particulates, formed during aspiration, from entering the pipette, preventing cross contamination. The positive-stop design reduces tip attachment and ejection forces. For use with Rainin™ LTS pipettes.
Recommended uses:
  • PCR
  • Molecular Biology
  • Cell Culture
Selecting the best ART tip with the right features
Tip type
Benefits
Recommended use
ART Pipette Tips*
Barrier tip
  • Self-sealing barrier prevents contamination from aerosols and liquids
  • Critical assays and samples
  • Pipetting expensive reagents
  • Every day pipetting
Non-filtered tip
  • Economic, high-quality tips
  • Non-critical assays and samples
Low-retention tip
  • Improves sample delivery and conservation of expensive reagents
  • Viscous samples
  • Samples containing detergents
  • Protein and DNA samples
  • Small volumes
Extended-length tip
  • Accessing the bottom of tall or narrow vessels without risk of contamination
  • Cell culture
  • Accessing bottom of narrow tubes
Wide-bore tip
  • Large orifice prevents cell shearing
Pipetting
  • Fragile cell lines
  • Macro-molecules (genomic DNA)
  • Viscous samples
  • Difficult-to-pipette solutions
Micropoint tip
  • Thin wall construction prevents drop formation on the tip
  • Aids in maximum sample delivery
  • Small-volume pipetting
  • Critical samples
Gel-loading tip
  • Easy, precise loading of wells in electrophoresis gels
  • Loading polyacrylamide or agarose gels for electrophoresis
  • Loading sequencing gels
Solvent-safe tip
  • Carbon filter protects pipette when using solvents
  • Pipetting solvents and caustic or corrosive solutions
SoftFit-L Pipette Tips*
SoftFit-L filtered tip
  • Blocks aerosols from entering pipette preventing contamination
  • Positive-stop design reduces attachment and ejection forces
  • Critical assays and samples
  • Every day pipetting
SoftFit-L non-filtered tip
  • Economic, high-quality tips
  • Positive-stop design reduces attachment and ejection forces
  • Non-critical assays and samples
SoftFit-L low-retention tip
  • Hydrophobic surface minimizes sample delivery and conservation of expensive reagents
  • Positive-stop design reduces attachment and ejection forces
  • Viscous samples
  • Samples containing detergents
  • Protein and DNA samples
  • Small volumes
*SoftFit-L tips are compatible with Rainin™ LTS pipettes only. Not for use with other manufacturer's pipettes or packaging.
Selecting the best ART tip with the right packaging
Packaging type
Features & benefits
ART Tips Packaging
Hinged rack
  • One-handed access to tips 
  • Rack transforms from hinged to lift-off lid
  • Flush tip insert for use with multi-channel pipettes
Lift-off lid
  • Independent lift-off lids
  • Ideal for use with multi-channel pipettes
Reload tower
  • Environmentally friendly packaging
  • Contains up to 54% less plastic than hinged-rack tips, and requires up to 55% less storage space
  • Maintains stringent quality and sterility standards
Reload inserts
  • Single, sterile insert of tips in blister cup
  • Reduces plastic use by up to 25%
Bulk
  • Non-sterile, bulk tips used to manually refill empty ART racks 
Individually wrapped
  • Single individually wrapped tips
  • Ideal for the most stringent sterility requirements
SoftFit-L Tips Packaging
SoftFit-L hinged rack
  • One-handed access to tips 
  • Flush tip insert for use with multi-channel pipettes
  • Color-coded for easy volume identification
SoftFit-L reload inserts*
  • Compatible with ART SoftFit-L empty racks
  • Single, sterile insert of filtered tips in blister cup
  • Reduces plastic use by up to 25%
SoftFit-L reload tower*
  • Compatible with ART SoftFit-L empty racks
  • Reduces plastic use by up to 50%
  • Maintains stringent quality and sterility standards
*SoftFit-L reload towers and inserts are compatible with SoftFit-L tip racks only. Not for use with other manufacturer's racks. SoftFit-L racks can be ordered separately.
Selecting the best ART tip with the right features
Tip type
Benefits
Recommended use
ART Pipette Tips*
Barrier tip
  • Self-sealing barrier prevents contamination from aerosols and liquids
  • Critical assays and samples
  • Pipetting expensive reagents
  • Every day pipetting
Non-filtered tip
  • Economic, high-quality tips
  • Non-critical assays and samples
Low-retention tip
  • Improves sample delivery and conservation of expensive reagents
  • Viscous samples
  • Samples containing detergents
  • Protein and DNA samples
  • Small volumes
Extended-length tip
  • Accessing the bottom of tall or narrow vessels without risk of contamination
  • Cell culture
  • Accessing bottom of narrow tubes
Wide-bore tip
  • Large orifice prevents cell shearing
Pipetting
  • Fragile cell lines
  • Macro-molecules (genomic DNA)
  • Viscous samples
  • Difficult-to-pipette solutions
Micropoint tip
  • Thin wall construction prevents drop formation on the tip
  • Aids in maximum sample delivery
  • Small-volume pipetting
  • Critical samples
Gel-loading tip
  • Easy, precise loading of wells in electrophoresis gels
  • Loading polyacrylamide or agarose gels for electrophoresis
  • Loading sequencing gels
Solvent-safe tip
  • Carbon filter protects pipette when using solvents
  • Pipetting solvents and caustic or corrosive solutions
SoftFit-L Pipette Tips*
SoftFit-L filtered tip
  • Blocks aerosols from entering pipette preventing contamination
  • Positive-stop design reduces attachment and ejection forces
  • Critical assays and samples
  • Every day pipetting
SoftFit-L non-filtered tip
  • Economic, high-quality tips
  • Positive-stop design reduces attachment and ejection forces
  • Non-critical assays and samples
SoftFit-L low-retention tip
  • Hydrophobic surface minimizes sample delivery and conservation of expensive reagents
  • Positive-stop design reduces attachment and ejection forces
  • Viscous samples
  • Samples containing detergents
  • Protein and DNA samples
  • Small volumes
*SoftFit-L tips are compatible with Rainin™ LTS pipettes only. Not for use with other manufacturer's pipettes or packaging.
Selecting the best ART tip with the right packaging
Packaging type
Features & benefits
ART Tips Packaging
Hinged rack
  • One-handed access to tips 
  • Rack transforms from hinged to lift-off lid
  • Flush tip insert for use with multi-channel pipettes
Lift-off lid
  • Independent lift-off lids
  • Ideal for use with multi-channel pipettes
Reload tower
  • Environmentally friendly packaging
  • Contains up to 54% less plastic than hinged-rack tips, and requires up to 55% less storage space
  • Maintains stringent quality and sterility standards
Reload inserts
  • Single, sterile insert of tips in blister cup
  • Reduces plastic use by up to 25%
Bulk
  • Non-sterile, bulk tips used to manually refill empty ART racks 
Individually wrapped
  • Single individually wrapped tips
  • Ideal for the most stringent sterility requirements
SoftFit-L Tips Packaging
SoftFit-L hinged rack
  • One-handed access to tips 
  • Flush tip insert for use with multi-channel pipettes
  • Color-coded for easy volume identification
SoftFit-L reload inserts*
  • Compatible with ART SoftFit-L empty racks
  • Single, sterile insert of filtered tips in blister cup
  • Reduces plastic use by up to 25%
SoftFit-L reload tower*
  • Compatible with ART SoftFit-L empty racks
  • Reduces plastic use by up to 50%
  • Maintains stringent quality and sterility standards
*SoftFit-L reload towers and inserts are compatible with SoftFit-L tip racks only. Not for use with other manufacturer's racks. SoftFit-L racks can be ordered separately.
ART barrier vs. traditional filter tips
ART barrier tips differ from traditional filter tips. ART self-sealing barrier tips are the only tips that seal completely to prevent liquid samples and aerosols from coming in contact with the pipette nose cone. When the barrier seals it protects the pipette and subsequent samples from cross contamination. 
Researchers have made ART self-sealing barrier tips their first choice in sample protection. ART barrier tips provide 100% security against aerosol and liquid contamination.
Compare the barrier
The ART barrier prevents cross contamination by blocking the passage of aerosols, liquids, radioactive isotopes, and biological materials to the pipette or to subsequent samples in an experiment.
The result of low retention
  • Less liquid retention to improve liquid handling
  • Improve volume accuracy to increase reliability
  • Precise sample delivery to save expensive reagents
SoftFit-L Tips
The SoftFit-L was designed for optimal performance with Rainin™ LTS pipettes. SoftFit-L tips deliver the accuracy and precision you expect and maintain the ergonomic benefits of the pipette with a positive-stop collar that reduces tip attachment and ejection forces.
Learn more