Thermo Scientific 48, 96 and 384 well polypropylene storage microplates and deepwell plates are an economical and reliable solution for sample storage, high-throughput screening and low-binding assays in drug discovery, molecular biology and genomic applications. They are made with virgin medical grade polypropylene resin to reduce extractables and leachables interacting with samples.
Thermo Scientific Abgene polypropylene storage plates are Class 8 clean room manufactured and human DNA free to ensure the absence of contamination for molecular biology applications. Thermo Scientific Nunc storage plates have over 40 years of quality in plastics manufacturing, offering one of the highest sterility assurance levels in the industry for cell-based applications (SAL 10-6).
Filter Results
Recommended product(s)