Clicktainer showcase
Simply twist the cap to click and seal the vial for leakproof* protection
We understand the value of patient samples. Sample collection needs to be easy, and sample transport needs to be secure. Thermo Scientific Samco Clicktainer vials have a wide opening and leakproof* seal. The “click” seal system indicates when the cap is properly applied so the sample will not leak or cause contamination.
Request a sample   Request a quote
What makes Clicktainer vials different from other sample containers?
Clicktainer click seal
Click Seal closure
The NEW Click Seal closure allows for visual and audible indication of the full cap closure. You will feel it click into place!
Clicktainer internal tabs
Easy sample handling
The smooth internal tabs reduce  the risk of tearing with gloves or the tab breaking off, contaminating the sample.
Clicktainer leakproof design
Leakproof performance guaranteed
Each Clicktainer vial is thoroughly tested:
  • Leak testing in accordance with BS EN 14254
Clicktainer usability features
Knurls on cap and base
Vials are easy to handle with gloved hands.
Stackable
Clicktainer vials are stackable, helping you to optimize sample storage.
Clicktainer audible click seal
Clicktainer family shot
Convenient labels, color caps, and formats for quick identification
Available in a range of label options:
  • Unlabelled
  • Tabbed label sterile
  • Non-tabbed label non-sterile
  • Tabbed temperature strip
  • Temperature strip only
And in a variety of cap colors and packaging formats:
  • Assembled product for general laboratory use, clinical applications, and mice necropsy
  • Dispenser box ideal for use both in smaller laboratories and doctor offices
  • Individual packaged product for sterile environments such as hospital operating rooms and cleanrooms
  • As separate components for manufacturing automation where suppliers add formalin solution, boric acid, sodium thiosulfate, or other reagents for resale into histology, urinalysis, water testing, etc.
Clicktainer label
Unlike other sample containers, Clicktainer labels are clearly designed:
  • Space for full name
  • Multiple languages—
    • English
    • Spanish
    • French
  • Medical number
  • IVD/ CE mark
  • Sterility level
Clicktainer compliance testing banner
Applications
283x195 providing sample protection
With their innovative closure system, Thermo Scientific Samco Clicktainer vials are suited for the collection and transport of fluids, solids, and powders. This makes them ideal for a wide range of customer applications, from the collection of valuable and often hazardous patient samples such as urine, feces, and sputum, to their suitability for use with formalin or fixatives for histology applications.
Thermo Fisher Scientific subjects the Samco Clicktainer Vial to a number of rigorous leak test procedures. We conduct a vacuum test that draws 95 kPa of pressure within a sealed chamber, followed by leak testing in accordance with BS EN 14254.
Clicktainer vial offering
Select the product code of interest and it will appear on the top of the ordering page.
120mL/53mm vials
Product Code Description Sterility Cap color Format Inner pack/ case qty.
120ORG53-1000 Tabbed with label Sterile Orange Preassembled 75/300
120WHT53-1000 Tabbed with label Sterile White Preassembled 75/300
120WHT53-2000 No label Non-sterile White Separate cap & vial 300/300
120YEL53-2000 Non-tabbed with label
Non-sterile Yellow Preassembled 75/300
120WHT53-3000 Midstream urine kit, no funnel
Sterile White Preassembled 1/100
120ORG53-4000 No label, peel pouch Sterile Orange Preassembled 1/100
120WHT53-4000 No label, peel pouch Sterile White Preassembled 1/100
90mL/53mm vials
Product Code Description Sterility Cap color Format Inner pack/ case qty.
90ORG53-1000 Tabbed with label    Sterile Orange Preassembled 100/400
90WHT53-1000 Tabbed with label    Sterile White Preassembled 100/300
90GRN53-1000 Tabbed with label   Sterile Green Preassembled 100/400
90ORG53-1001 Tabbed with label, dispenser pack Sterile Orange Preassembled 20/320
90WHT53-1001 Temperature strip with label 
Sterile White Preassembled 100/300
90WHT53-1002 Temperature strip only Sterile White Preassembled 100/400
90WHT53-2000 Non-tabbed with label Non-sterile White Preassembled 75/300
90YEL53-2000 Non-tabbed with label
Non-sterile Yellow Preassembled 100/400
90WHT53-2001 Temperature strip with label
Non-sterile White Preassembled 300/300
90WHT53-2002 No label
Non-sterile White Separate cap & vial 300/300
90WHT53-2003 No label
Non-sterile White Preassembled 300/300
90WHT53-2004 Temperature strip only Non-sterile White Preassembled 100/400
Caps and uncapped vials
Product Code Description Sterility Cap color Format Inner pack/ case qty.
9000-5300 Vial, no label Non-sterile No cap Vial 300/300
12000-5300 Vial, no label Non-sterile No cap Vial 300/300
CAPWHT53-1000 Clickseal cap Non-sterile White Cap 1500/1500
CAPORG53-1000 Clickseal cap Non-sterile Orange Cap 1500/1500
CAPGRN53-1000 Clickseal cap Non-sterile Green Cap 1500/1500
CAPRED53-1000 Clickseal cap Non-sterile Red Cap 1500/1500
CAPYEL53-1000 Clickseal cap Non-sterile Yellow Cap 1500/1500
120mL/53mm vials
Product Code Description Sterility Cap color Format Inner pack/ case qty.
120ORG53-1000 Tabbed with label Sterile Orange Preassembled 75/300
120WHT53-1000 Tabbed with label Sterile White Preassembled 75/300
120WHT53-2000 No label Non-sterile White Separate cap & vial 300/300
120YEL53-2000 Non-tabbed with label
Non-sterile Yellow Preassembled 75/300
120WHT53-3000 Midstream urine kit, no funnel
Sterile White Preassembled 1/100
120ORG53-4000 No label, peel pouch Sterile Orange Preassembled 1/100
120WHT53-4000 No label, peel pouch Sterile White Preassembled 1/100
90mL/53mm vials
Product Code Description Sterility Cap color Format Inner pack/ case qty.
90ORG53-1000 Tabbed with label    Sterile Orange Preassembled 100/400
90WHT53-1000 Tabbed with label    Sterile White Preassembled 100/300
90GRN53-1000 Tabbed with label   Sterile Green Preassembled 100/400
90ORG53-1001 Tabbed with label, dispenser pack Sterile Orange Preassembled 20/320
90WHT53-1001 Temperature strip with label 
Sterile White Preassembled 100/300
90WHT53-1002 Temperature strip only Sterile White Preassembled 100/400
90WHT53-2000 Non-tabbed with label Non-sterile White Preassembled 75/300
90YEL53-2000 Non-tabbed with label
Non-sterile Yellow Preassembled 100/400
90WHT53-2001 Temperature strip with label
Non-sterile White Preassembled 300/300
90WHT53-2002 No label
Non-sterile White Separate cap & vial 300/300
90WHT53-2003 No label
Non-sterile White Preassembled 300/300
90WHT53-2004 Temperature strip only Non-sterile White Preassembled 100/400
Caps and uncapped vials
Product Code Description Sterility Cap color Format Inner pack/ case qty.
9000-5300 Vial, no label Non-sterile No cap Vial 300/300
12000-5300 Vial, no label Non-sterile No cap Vial 300/300
CAPWHT53-1000 Clickseal cap Non-sterile White Cap 1500/1500
CAPORG53-1000 Clickseal cap Non-sterile Orange Cap 1500/1500
CAPGRN53-1000 Clickseal cap Non-sterile Green Cap 1500/1500
CAPRED53-1000 Clickseal cap Non-sterile Red Cap 1500/1500
CAPYEL53-1000 Clickseal cap Non-sterile Yellow Cap 1500/1500
*The term “leakproof” applies to Thermo Scientific Samco Clicktainer products, 90mL/53 mm and 120mL/53 mm, that meet the following criteria: a) a vacuum test that draws 95kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with BS EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by 1 hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco Clicktainer Vials under conditions of their planned applications. Thermo Scientific Samco Clicktainer Vials are leakproof at ambient temperature and pressure when used with their Clicktainer closures.