Sample preparation for molecular and non-molecular workflows
Sample preparation for molecular and non-molecular workflows
Prepare even the most challenging food samples with options to pool, clean, purify, concentrate and automate your sample preparation for molecular detection and other techniques.
Nucleic acid extraction systems for food samples
Applied Biosystems™ PrepSEQ™ Rapid Spin Sample Preparation Kits are designed for manual sample preparation upfront of the Applied Biosystems™ MicroSEQ™, Applied Biosystems™ RapidFinder™ STEC and Applied Biosystems™ TaqMan™ assays. They are suitable for use with other PCR kits and offer:
  • Fast, efficient sample preparation for PCR-based assays
  • Optimal performance on a wide range of sample types
  • A simple protocol which reduces time-to-results
  • A microcentrifuge-based system — no additional equipment is required
  • An ideal solution for low to medium sample throughput
Automate DNA extraction for high test throughput environments with the Applied Biosystems™ MagMAX™ Express magnetic particle processor. Ideal for automated magnetic-bead-based DNA extraction using Applied Biosystems™ PrepSEQ™ Nucleic Acid Extraction Reagents, upfront of MicroSEQ, RapidFinder STEC and Taqman assays, the system has been independently validated and approved for automating the sample preparation process for a number of Applied Biosystems food application workflows.
Immunomagnetic separation (IMS) and paramagnetic preparation for food samples
Selectively capture and concentrate target organisms from enriched samples by immunomagnetic separation (IMS) technology. After washing, the bead bound target is released for use in downstream applications. Dynabeads™ magnetic beads for food safety testing have been designed for both manual and automated separation using the Applied Biosystems™ BeadRetriever™ Magnetic Bead Processor.
Selectively bind and concentrate target organisms from many complex matrices found in food, animal feed, ingredients and environmental samples with the Applied Biosystems™ Pathatrix™ system.
Food-authenticity-labeling-270x195
Be confident in the authenticity of food ingredients with rapid molecular solutions
Combat food fraud and ensure correct product labeling with effective species detection and quantification solutions, and genetically modified organism (GMO) testing, to help you to protect public health and your brand reputation.
Learn more
Resources
Follow us
Join our food testing community for the latest food safety news and updates.