Fast, reproducible sample injection with minimum carryover
Fast, reproducible sample injection with minimum carryover
These innovative and carefully designed autosamplers ensure reliable, precise, and accurate injection for nanoliter to milliliter sample volumes. They support a wide range of formats and sample throughputs. Choose from rapid separation, nano, capillary, micro, standard, semipreparative, and more.
Popular LC autosamplers
UltiMate 3000 ACC-3000 Autosampler Column Compartments
UltiMate WPS-3000SL/TSL Standard Well Plate Autosampler
Vanquish™ Split Samplers
LC autosamplers and sample injectors
Sample Handling Devices for Vanquish Systems
Vanquish™ Split Samplers Gain confidence in peak identification and quantitation from maximum injection precision and huge sample capacity with high-throughput performance.
Vanquish™ Charger Module Increase sample capacity for extra-long unattended operation. This intelligent robotic automates sample loading in the Vanquish UHPLC system.
Autosampler for UltiMate 3000 Basic Systems
UltiMate™ 3000 ACC-3000 Autosampler Column Compartments Save money and space with this integrated autosampler and column compartment which supports conventional LC and UHPLC at backpressures up to 62MPa.
Autosampler for UltiMate 3000 Standard (SD) Systems
UltiMate™ 3000 WPS-3000SL/TSL Standard Well Plate Autosamplers A flexible, affordable autosampler you can count on.
Autosampler for UltiMate 3000 Rapid Separation (RS) Systems
UltiMate™ 3000 WPS-3000RS/TRS Rapid Separation Well Plate Autosamplers Minimize gradient delay and extra column volume for high-efficiency separations and short chromatographic run times.
Autosamplers for Biocompatible Systems
UltiMate™ 3000 WPS-3000TBPL Thermostatted Biocompatible Standard Well Plate Autosampler Protect your sensitive biomolecules in standard analytical applications. Autosampler supports well plates and vial trays in any combination.
UltiMate™ 3000 WPS-3000TBRS Biocompatible Rapid Separation Well Plate Autosampler Offers an entirely biocompatible flow path for sensitive analyses. Ideal for sensitive electrochemical detection or biomolecule analyses in rapid separation applications (15,000 psi).
Vanquish™ Split Samplers Gain confidence in peak identification and quantitation from maximum injection precision and huge sample capacity with high-throughput performance.
Vanquish™ Charger Module Increase sample capacity for extra-long unattended operation. This intelligent robotic automates sample loading in the Vanquish UHPLC system.
Autosampler for UltiMate 3000 RSLCnano Systems
UltiMate™ 3000 WPS-3000TPL/PL RSLCnano Well Plate Autosamplers Ensure excellent peak shape and resolution for small injection volumes and low flow rates. Autosampler supports pressures to 90 MPa and multiple sample formats.
Autosampler for UltiMate 3000 Semi-Preparative
UltiMate™ 3000 WPS-3000SL/TSL Semi-Preparative Autosamplers Perform reliable, accurate and precise large-volume injections for purifying compounds to increase lower milligram range with this semi-preparative autosampler.
Autosampler with Fraction Collection
UltiMate™ 3000 WPS-3000TFC/BFC Well Plate Autosampler with Fraction Collector Simplify off-line multidimensional and multistep liquid chromatography (LC) in analytical and micro flow rate ranges.
Need Autosampler Accessories?
Need autosampler accessories?
Get reliable performance and productivity with quality parts and accessories designed for your HPLC/UHPLC autosampler.
Resources