Reduce process time and enhance flexibility
We have the hands-on expertise to help you accelerate productivity and ease the burden of regulatory compliance. The quality of our production cell culture platform, Thermo Scientific Nunc Cell Factory systems, reflects the rigorous standards that have made us the world's leading provider of cell culture products.
Obtain faster results and lot-to-lot consistency with Nunc Cell Factory systems. A proven solution for large-scale production of cells, vaccines, and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture products. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling, and harvesting.
Advantages of rigid containment
Rigid containers are available in PC, PETG, PP, LDPE and HDPE, and in volumes ranging from 30 mL to 50 L. Extensive regulatory documentation assures that the products meet your demanding biotechnology requirements and reduces the time and cost to implement them in your process. The Thermo Scientific Nalgene multi-tiered storage portfolio enables you to choose the appropriate product quality for your application. Users of our tiered portfolio benefit from:
  • Low particulates: USP <788> compliance reduces the chance of contamination; decreasing potential loss of high value contents
  • Improved packaging: Triple layer packaging facilitates ease of entry into clean room environments
  • Sterile and ready-to-use: Saves resources by reducing the need for validation, washing, packaging, and sterilization
Featured products
Choose the right bottle for your application. Available in PC, PETG, and HDPE, and in volumes ranging from 30 mL to 20 L. Extensive regulatory documentation assures that the products meet your demanding biotechnology requirements and reduces the time and cost to implement them in your process
When using carboys for critical applications, standard laboratory carboys won't do. Supporting documentation, choices and designs sensitive to the necessary cleaning validations are just a few things to look for.
Container cleanliness is critical. Understanding different cleanliness and validation options, as well as their impact, can help you to identify quality products that will not influence the integrity of your product.
How do you select the best containers for your application?
Container cleanliness is critical. Understanding different cleanliness and validation options, as well as their impact, can help you to identify quality products that will not influence the integrity of your product. Find out about what you need to consider when selecting a container for your biopharmaceutical manufacturing applications.
Download our infographic
* Our guarantee for a leakproof seal is subject to our standard product warranty, as set forth in the Thermo Fisher Scientific Terms and Conditions of Sale. Our products are leakproof at ambient temperature and pressure when used with their corresponding closures. However, to ensure safe usage, customers are advised to test our containers and closures under conditions of their planned applications.