Advanced transmitted-light system
Cell counting and time-lapse imaging
This cell imaging system is the ideal instrument for transmitted-light microscopy. Combining precision components with unique design functionality, the EVOS XL Cell Imaging System is perfect for colorimetric applications like H&E  staining and immunohistochemistry.
Request a demo  Request a quote
Compare imaging instruments
Cell Imaging System is the ideal instrument for transmitted-light microscopy. Combining precision components with unique design functionality, the EVOS® XL Cell Imaging System is perfect for colorimetric applications like H&E  staining and immunohistochemistry.
Request a demo
See the gallery of beautiful images taken with the EVOS XL Cell Imaging System.
See what your peers are publishing with the EVOS XL Cell Imaging System.
Get the full experience by requesting an in-lab demo from one of our trained representatives.
EVOS imaging systems brochure
See the entire portfolio of easy to use cell imaging instruments that can be configured with a variety of objectives, light cubes, and vessel holders to meet your research needs.
Download the EVOS brochure
System Highlights
Attribute EVOS XL Detail
Illumination Adjustable intensity LED (>50,000-hour life per light cube)
Contrast methods Transmitted light (bright field and phase contrast)
Objective turret 5-position, front-mounted control
Condenser Includes 3 positions, with bright-field and phase-contrast annuli
Condenser working distance 53 mm
Stage Mechanical “glide” stage with X-Y axis fine-positioning controls
Interchangeable vessel holders available
LCD display 15” high-resolution (1,024 x 768 pixels) color monitor with adjustable tilt
Cameras Color:
1/2” 2,048 x 1,536, 3.1 megapixels
Captured images Color camera:
24-bit color TIFF or PNG;
JPEG, BMP (2,048 x 1,536 pixels
Output ports 3 USB ports, 1 DVI port (supports direct output to USB and networked stage)
Power supply AC adaptor; input 100–240 V, 50–60 Hz; output 12 V DC/4.15 A
Dimensions Height: 578 mm (22.8 in)
Depth: 470 mm (18.5 in)
Weight 14.2 kg (31.4 lb)
EVOS® XL Cell Imaging System Citations
Thermo Fisher Cloud provides scientists with a secure place to store, share and analyze their data. Upload your files from anywhere you have access to the Internet by simply dragging them to the Cloud folder on your Windows™ desktop. Share the results with your colleagues to observe patterns, validate findings, and develop insights. And it’s not just for data files. Upload a presentation for an upcoming conference or a particularly useful how-to video.  
Get started today with 10 GB of free storage.
Learn more ›
Access it now › 
Use your existing web account username and password or register 
System Highlights
Attribute EVOS XL Detail
Illumination Adjustable intensity LED (>50,000-hour life per light cube)
Contrast methods Transmitted light (bright field and phase contrast)
Objective turret 5-position, front-mounted control
Condenser Includes 3 positions, with bright-field and phase-contrast annuli
Condenser working distance 53 mm
Stage Mechanical “glide” stage with X-Y axis fine-positioning controls
Interchangeable vessel holders available
LCD display 15” high-resolution (1,024 x 768 pixels) color monitor with adjustable tilt
Cameras Color:
1/2” 2,048 x 1,536, 3.1 megapixels
Captured images Color camera:
24-bit color TIFF or PNG;
JPEG, BMP (2,048 x 1,536 pixels
Output ports 3 USB ports, 1 DVI port (supports direct output to USB and networked stage)
Power supply AC adaptor; input 100–240 V, 50–60 Hz; output 12 V DC/4.15 A
Dimensions Height: 578 mm (22.8 in)
Depth: 470 mm (18.5 in)
Weight 14.2 kg (31.4 lb)
EVOS® XL Cell Imaging System Citations
Thermo Fisher Cloud provides scientists with a secure place to store, share and analyze their data. Upload your files from anywhere you have access to the Internet by simply dragging them to the Cloud folder on your Windows™ desktop. Share the results with your colleagues to observe patterns, validate findings, and develop insights. And it’s not just for data files. Upload a presentation for an upcoming conference or a particularly useful how-to video.  
Get started today with 10 GB of free storage.
Learn more ›
Access it now › 
Use your existing web account username and password or register 
EVOS XL Resources
EVOS XL Software Update
EVOS XL User Manual
EVOS XL Quick Start Guide
EVOS Accessories
Webinars
New Cell Analysis Support Center
Find tips, troubleshooting help, and resources for your cell analysis workflow.