AscI nucleotide recognition sequence
Figure color legend
Name derivation: Arthrobacter species
Recognition and Cutting site: GG^CGCG CC
CC GCGC^GG
Resulting DNA Ends: 5′ protruding ends
Prototype: AscI
Isoschizomers: SgsI, PalAI
Sensitivity to methylation:  CpG blocked
Neoschizomers None
Anza RE generating compatible ends Anza™ 28 MluI, Anza™ 45 PteI, Anza™ 95 MauBI
References