PmeI nucleotide recognition sequence
Figure color legend
Name derivation: Pseudomonas mendocina
Recognition and Cutting site: GTTT^AAAC
CAAA^TTG
Resulting DNA Ends: blunt
Prototype: PmeI
Isoschizomers: MssI
Sensitivity to methylation:  EcoKI blocked if overlapped
Neoschizomers None
Anza RE generating compatible ends Any enzyme generating blunt ends
References