Name derivation: Rhodopseudomonas sphaeroides
Recognition and Cutting site: GT^AC
CA^TG
Resulting DNA Ends: blunt
Prototype: RsaI
Isoschizomers: AfaI
Sensitivity to methylation:  CpG impaired if overlapped
Neoschizomers Csp6I, CviQI, RsaNI
Anza RE generating compatible ends Any enzyme generating blunt ends
References