Name derivation: Streptomyces achromogenes
Recognition and Cutting site: G AGCT^C
C^TCGA G
Resulting DNA Ends: 3′ protruding ends
Prototype: SacI
Isoschizomers: Psp124BI, SstI
Sensitivity to methylation:  None
Neoschizomers Ecl136II, Eco53kI, EcoICRI
Anza RE generating compatible ends Anza™ 126 Alw21I* (*if recognized sequence is 5'-GAGCTC-3')
References