SpeI nucleotide recognition sequence
Figure color legend
Name derivation: Sphaerotilus natans
Recognition and Cutting site: A^CTAG T
T GATC^A
Resulting DNA Ends: 5′ protruding ends
Prototype: SpeI
Isoschizomers:
BcuI, AhlI
Sensitivity to methylation:  none
Neoschizomers None
Anza RE generating compatible ends Anza™ 6 NheI, Anza™ 12 XbaI, Anza™ 15 XmaJI
References